Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35076347-35076849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCG
Entrez Gene ID
2189
Gene Name
Fanconi anemia complementation group G
Gene Aliases
FAG, XRCC9
UniGene
Hs.591084
Gene Chromosome Location
Chr.9: 35073838-35080016 on Build GRCh38
Transcripts
NM_004629.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4945101 c.924+8A>C p.? Chr.9: 35076716-35076716 on Build GRCh38
3779990 c.970G>C p.E324Q Chr.9: 35076538-35076538 on Build GRCh38
753540 c.930G>C p.L310F Chr.9: 35076578-35076578 on Build GRCh38
5698594 c.971A>T p.E324V Chr.9: 35076537-35076537 on Build GRCh38
2773923 c.1071G>A p.T357T Chr.9: 35076437-35076437 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774982952 Chr.9: 35076728-35076728 on Build GRCh38
rs397507561 Chr.9: 35076585-35076585 on Build GRCh38
rs112854819 Chr.9: 35076438-35076438 on Build GRCh38
rs771395169 Chr.9: 35076815-35076815 on Build GRCh38
rs762238437 Chr.9: 35076509-35076509 on Build GRCh38
rs772109570 Chr.9: 35076460-35076460 on Build GRCh38
rs147594292 Chr.9: 35076555-35076555 on Build GRCh38
rs121434426 Chr.9: 35076442-35076442 on Build GRCh38
rs755031729 Chr.9: 35076781-35076781 on Build GRCh38
rs966754065 Chr.9: 35076606-35076607 on Build GRCh38
rs896457719 Chr.9: 35076596-35076596 on Build GRCh38
rs774516300 Chr.9: 35076443-35076443 on Build GRCh38
rs774687438 Chr.9: 35076821-35076821 on Build GRCh38
rs587778347 Chr.9: 35076785-35076785 on Build GRCh38
rs373748419 Chr.9: 35076626-35076626 on Build GRCh38
rs762162897 Chr.9: 35076592-35076592 on Build GRCh38
rs753610432 Chr.9: 35076620-35076620 on Build GRCh38
rs745833004 Chr.9: 35076564-35076564 on Build GRCh38
rs758835619 Chr.9: 35076637-35076637 on Build GRCh38
rs761607613 Chr.9: 35076596-35076597 on Build GRCh38
rs1011265736 Chr.9: 35076624-35076624 on Build GRCh38
rs761579255 Chr.9: 35076747-35076747 on Build GRCh38
rs771357395 Chr.9: 35076719-35076719 on Build GRCh38
rs4986940 Chr.9: 35076520-35076520 on Build GRCh38
rs896122445 Chr.9: 35076667-35076667 on Build GRCh38
rs745928033 Chr.9: 35076446-35076446 on Build GRCh38
rs369622138 Chr.9: 35076487-35076487 on Build GRCh38
rs772068491 Chr.9: 35076698-35076698 on Build GRCh38
rs750185901 Chr.9: 35076761-35076761 on Build GRCh38
rs886063897 Chr.9: 35076768-35076768 on Build GRCh38
rs969148942 Chr.9: 35076392-35076392 on Build GRCh38
rs934681493 Chr.9: 35076572-35076572 on Build GRCh38
rs780410457 Chr.9: 35076425-35076429 on Build GRCh38
rs752346718 Chr.9: 35076540-35076540 on Build GRCh38
rs764465471 Chr.9: 35076491-35076491 on Build GRCh38
rs777452126 Chr.9: 35076433-35076433 on Build GRCh38
rs567380452 Chr.9: 35076606-35076606 on Build GRCh38
rs976115806 Chr.9: 35076801-35076801 on Build GRCh38
rs746425749 Chr.9: 35076823-35076823 on Build GRCh38
rs975327872 Chr.9: 35076639-35076639 on Build GRCh38
rs774491251 Chr.9: 35076681-35076681 on Build GRCh38
rs955088655 Chr.9: 35076682-35076682 on Build GRCh38
rs767013203 Chr.9: 35076607-35076607 on Build GRCh38
rs780448016 Chr.9: 35076404-35076404 on Build GRCh38
rs749198217 Chr.9: 35076434-35076434 on Build GRCh38
rs376714786 Chr.9: 35076416-35076416 on Build GRCh38
rs773864987 Chr.9: 35076597-35076597 on Build GRCh38
rs766989205 Chr.9: 35076534-35076534 on Build GRCh38
rs749744400 Chr.9: 35076451-35076464 on Build GRCh38
rs1032767148 Chr.9: 35076630-35076630 on Build GRCh38
rs55802583 Chr.9: 35076398-35076398 on Build GRCh38
rs781363676 Chr.9: 35076689-35076689 on Build GRCh38
rs145092954 Chr.9: 35076551-35076551 on Build GRCh38
rs763451432 Chr.9: 35076713-35076713 on Build GRCh38
rs17883759 Chr.9: 35076627-35076627 on Build GRCh38
rs892370233 Chr.9: 35076623-35076623 on Build GRCh38
rs368982658 Chr.9: 35076420-35076420 on Build GRCh38
rs193268256 Chr.9: 35076693-35076693 on Build GRCh38
rs770187727 Chr.9: 35076605-35076605 on Build GRCh38
rs755919340 Chr.9: 35076432-35076432 on Build GRCh38
rs984958735 Chr.9: 35076506-35076506 on Build GRCh38
rs370002843 Chr.9: 35076581-35076581 on Build GRCh38
rs775450331 Chr.9: 35076578-35076578 on Build GRCh38
rs34504701 Chr.9: 35076750-35076751 on Build GRCh38
rs375991144 Chr.9: 35076524-35076524 on Build GRCh38
rs1034678101 Chr.9: 35076406-35076406 on Build GRCh38
rs755753920 Chr.9: 35076543-35076543 on Build GRCh38
rs1052752319 Chr.9: 35076595-35076595 on Build GRCh38
rs776920780 Chr.9: 35076590-35076590 on Build GRCh38
rs867467817 Chr.9: 35076823-35076824 on Build GRCh38
rs571751302 Chr.9: 35076780-35076780 on Build GRCh38
rs768083289 Chr.9: 35076567-35076567 on Build GRCh38
rs375601834 Chr.9: 35076731-35076731 on Build GRCh38
rs754317012 Chr.9: 35076382-35076382 on Build GRCh38
rs139157408 Chr.9: 35076633-35076633 on Build GRCh38
rs146244724 Chr.9: 35076609-35076609 on Build GRCh38
rs61757386 Chr.9: 35076743-35076743 on Build GRCh38
rs781276482 Chr.9: 35076790-35076790 on Build GRCh38
rs768922471 Chr.9: 35076608-35076608 on Build GRCh38
rs56139991 Chr.9: 35076712-35076712 on Build GRCh38
rs763736360 Chr.9: 35076733-35076733 on Build GRCh38
rs34100177 Chr.9: 35076551-35076552 on Build GRCh38
rs17880082 Chr.9: 35076767-35076767 on Build GRCh38
rs770009374 Chr.9: 35076804-35076804 on Build GRCh38
rs910750230 Chr.9: 35076512-35076512 on Build GRCh38
rs765722724 Chr.9: 35076516-35076516 on Build GRCh38
rs530535955 Chr.9: 35076574-35076574 on Build GRCh38
rs758294567 Chr.9: 35076634-35076634 on Build GRCh38
rs754507676 Chr.9: 35076405-35076405 on Build GRCh38
rs779840229 Chr.9: 35076779-35076779 on Build GRCh38
rs763827052 Chr.9: 35076612-35076612 on Build GRCh38
rs202152417 Chr.9: 35076552-35076552 on Build GRCh38
rs778142751 Chr.9: 35076808-35076808 on Build GRCh38
rs754484649 Chr.9: 35076532-35076532 on Build GRCh38
rs749604746 Chr.9: 35076813-35076813 on Build GRCh38
rs1034785014 Chr.9: 35076661-35076661 on Build GRCh38
rs768793395 Chr.9: 35076688-35076688 on Build GRCh38
rs768761780 Chr.9: 35076589-35076589 on Build GRCh38
rs780110754 Chr.9: 35076678-35076678 on Build GRCh38
rs372808885 Chr.9: 35076492-35076492 on Build GRCh38
rs757780161 Chr.9: 35076384-35076384 on Build GRCh38
rs2237857 Chr.9: 35076758-35076758 on Build GRCh38
rs964852317 Chr.9: 35076768-35076768 on Build GRCh38
rs748334033 Chr.9: 35076791-35076791 on Build GRCh38
rs749657888 Chr.9: 35076478-35076478 on Build GRCh38

View Full Product Details