Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35699843-35700351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLN1
Entrez Gene ID
7094
Gene Name
talin 1
Gene Aliases
ILWEQ, TLN
UniGene
Hs.471014
Gene Chromosome Location
Chr.9: 35697337-35732395 on Build GRCh38
Transcripts
NM_006289.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7160343 c.6598G>A p.D2200N Chr.9: 35700253-35700253 on Build GRCh38
50251 c.6566C>T p.A2189V Chr.9: 35700285-35700285 on Build GRCh38
5998667 c.6629G>A p.R2210H Chr.9: 35700222-35700222 on Build GRCh38
6304357 c.6626G>A p.R2209H Chr.9: 35700225-35700225 on Build GRCh38
3848482 c.6650G>A p.R2217Q Chr.9: 35700201-35700201 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368513332 Chr.9: 35700008-35700008 on Build GRCh38
rs761387809 Chr.9: 35700262-35700262 on Build GRCh38
rs779916121 Chr.9: 35700180-35700180 on Build GRCh38
rs564539167 Chr.9: 35699915-35699915 on Build GRCh38
rs140329770 Chr.9: 35700284-35700284 on Build GRCh38
rs750943300 Chr.9: 35699927-35699927 on Build GRCh38
rs755770682 Chr.9: 35699943-35699943 on Build GRCh38
rs764826141 Chr.9: 35700270-35700270 on Build GRCh38
rs546429149 Chr.9: 35699905-35699905 on Build GRCh38
rs1007964919 Chr.9: 35700225-35700225 on Build GRCh38
rs111273183 Chr.9: 35699897-35699897 on Build GRCh38
rs1028051257 Chr.9: 35699900-35699900 on Build GRCh38
rs747049665 Chr.9: 35700091-35700091 on Build GRCh38
rs2295794 Chr.9: 35700153-35700153 on Build GRCh38
rs770202937 Chr.9: 35700208-35700208 on Build GRCh38
rs763416037 Chr.9: 35700140-35700140 on Build GRCh38
rs752137122 Chr.9: 35700146-35700146 on Build GRCh38
rs879119546 Chr.9: 35700071-35700071 on Build GRCh38
rs374067583 Chr.9: 35700115-35700115 on Build GRCh38
rs374219676 Chr.9: 35700254-35700254 on Build GRCh38
rs879165236 Chr.9: 35700301-35700301 on Build GRCh38
rs897970005 Chr.9: 35700103-35700103 on Build GRCh38
rs889164933 Chr.9: 35700016-35700016 on Build GRCh38
rs372820994 Chr.9: 35700182-35700182 on Build GRCh38
rs770185051 Chr.9: 35700121-35700121 on Build GRCh38
rs201579023 Chr.9: 35700159-35700159 on Build GRCh38
rs144449921 Chr.9: 35700154-35700154 on Build GRCh38
rs756242496 Chr.9: 35700322-35700322 on Build GRCh38
rs759032379 Chr.9: 35700028-35700028 on Build GRCh38
rs116187598 Chr.9: 35699885-35699885 on Build GRCh38
rs35538582 Chr.9: 35700020-35700021 on Build GRCh38
rs376931317 Chr.9: 35699960-35699960 on Build GRCh38
rs138625537 Chr.9: 35700217-35700217 on Build GRCh38
rs763362601 Chr.9: 35700214-35700214 on Build GRCh38
rs767008341 Chr.9: 35700029-35700029 on Build GRCh38
rs748941469 Chr.9: 35700045-35700045 on Build GRCh38
rs749106140 Chr.9: 35700048-35700048 on Build GRCh38
rs369758025 Chr.9: 35700168-35700168 on Build GRCh38
rs760027665 Chr.9: 35700223-35700223 on Build GRCh38
rs529885482 Chr.9: 35699980-35699980 on Build GRCh38
rs750483415 Chr.9: 35699893-35699893 on Build GRCh38
rs769570217 Chr.9: 35700141-35700142 on Build GRCh38
rs768485173 Chr.9: 35700112-35700112 on Build GRCh38
rs374966990 Chr.9: 35699948-35699948 on Build GRCh38
rs780920772 Chr.9: 35699938-35699938 on Build GRCh38
rs777297308 Chr.9: 35699953-35699953 on Build GRCh38
rs760753833 Chr.9: 35699984-35699984 on Build GRCh38
rs750965317 Chr.9: 35700021-35700021 on Build GRCh38
rs148578435 Chr.9: 35700198-35700198 on Build GRCh38
rs780053708 Chr.9: 35700143-35700145 on Build GRCh38
rs779969254 Chr.9: 35700090-35700090 on Build GRCh38
rs758160563 Chr.9: 35700283-35700283 on Build GRCh38
rs769841997 Chr.9: 35700201-35700201 on Build GRCh38
rs149234082 Chr.9: 35699935-35699935 on Build GRCh38
rs369986019 Chr.9: 35699940-35699940 on Build GRCh38
rs551759488 Chr.9: 35700167-35700167 on Build GRCh38
rs778640666 Chr.9: 35700052-35700052 on Build GRCh38
rs371527394 Chr.9: 35700096-35700096 on Build GRCh38
rs574175840 Chr.9: 35699875-35699875 on Build GRCh38
rs981246241 Chr.9: 35699926-35699926 on Build GRCh38
rs200428164 Chr.9: 35700261-35700261 on Build GRCh38
rs962391714 Chr.9: 35699894-35699894 on Build GRCh38
rs749110540 Chr.9: 35700149-35700156 on Build GRCh38
rs745787793 Chr.9: 35700057-35700057 on Build GRCh38
rs745844921 Chr.9: 35699962-35699962 on Build GRCh38
rs181220913 Chr.9: 35699892-35699892 on Build GRCh38
rs752190315 Chr.9: 35700038-35700038 on Build GRCh38
rs201563518 Chr.9: 35700292-35700292 on Build GRCh38
rs747767154 Chr.9: 35700143-35700143 on Build GRCh38
rs199643101 Chr.9: 35699961-35699961 on Build GRCh38
rs1032221970 Chr.9: 35700329-35700329 on Build GRCh38
rs1019489575 Chr.9: 35700249-35700249 on Build GRCh38
rs144879123 Chr.9: 35700020-35700020 on Build GRCh38
rs139866308 Chr.9: 35699904-35699904 on Build GRCh38
rs915658768 Chr.9: 35699901-35699901 on Build GRCh38
rs772133960 Chr.9: 35699963-35699963 on Build GRCh38
rs754818491 Chr.9: 35700293-35700293 on Build GRCh38
rs74431517 Chr.9: 35700241-35700241 on Build GRCh38
rs112672148 Chr.9: 35699973-35699973 on Build GRCh38
rs776747087 Chr.9: 35700005-35700005 on Build GRCh38
rs781194388 Chr.9: 35700299-35700299 on Build GRCh38
rs567897405 Chr.9: 35700302-35700302 on Build GRCh38
rs544154004 Chr.9: 35700044-35700044 on Build GRCh38
rs149034259 Chr.9: 35700075-35700075 on Build GRCh38
rs749073228 Chr.9: 35699959-35699959 on Build GRCh38
rs143073968 Chr.9: 35700202-35700202 on Build GRCh38
rs185621543 Chr.9: 35700187-35700187 on Build GRCh38
rs773653646 Chr.9: 35700123-35700123 on Build GRCh38
rs753524036 Chr.9: 35700158-35700158 on Build GRCh38
rs188918349 Chr.9: 35700230-35700230 on Build GRCh38
rs768271086 Chr.9: 35700161-35700161 on Build GRCh38
rs374151556 Chr.9: 35699997-35699997 on Build GRCh38
rs746990760 Chr.9: 35700181-35700181 on Build GRCh38
rs951788288 Chr.9: 35699895-35699895 on Build GRCh38
rs369648672 Chr.9: 35700222-35700222 on Build GRCh38
rs778587047 Chr.9: 35700116-35700116 on Build GRCh38
rs11541910 Chr.9: 35700199-35700199 on Build GRCh38
rs113763455 Chr.9: 35699972-35699972 on Build GRCh38
rs771935637 Chr.9: 35700081-35700081 on Build GRCh38

View Full Product Details