Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 36163791-36164333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLIPR2
Entrez Gene ID
152007
Gene Name
GLI pathogenesis related 2
Gene Aliases
C9orf19, GAPR-1, GAPR1
UniGene
Hs.493819
Gene Chromosome Location
Chr.9: 36136536-36163913 on Build GRCh38
Transcripts
NM_001287010.1 NM_001287011.1 NM_001287012.1 NM_001287013.1 NM_001287014.1 NM_022343.3 NR_104637.1 NR_104638.1 NR_104639.1 NR_104640.1 NR_104641.1

SNP Detail

SNP ID Chromosome Location
rs546090120 Chr.9: 36164108-36164108 on Build GRCh38
rs1015308603 Chr.9: 36163947-36163947 on Build GRCh38
rs540707204 Chr.9: 36164047-36164047 on Build GRCh38
rs527432636 Chr.9: 36164191-36164191 on Build GRCh38
rs386734481 Chr.9: 36163941-36163942 on Build GRCh38
rs903160721 Chr.9: 36163891-36163891 on Build GRCh38
rs957377902 Chr.9: 36164288-36164288 on Build GRCh38
rs774315607 Chr.9: 36163984-36163984 on Build GRCh38
rs562800328 Chr.9: 36164113-36164113 on Build GRCh38
rs917765318 Chr.9: 36164222-36164222 on Build GRCh38
rs1031939716 Chr.9: 36163919-36163919 on Build GRCh38
rs562363628 Chr.9: 36164069-36164069 on Build GRCh38
rs41277095 Chr.9: 36163907-36163907 on Build GRCh38
rs77267485 Chr.9: 36164261-36164261 on Build GRCh38
rs966023790 Chr.9: 36163901-36163901 on Build GRCh38
rs866832156 Chr.9: 36163845-36163845 on Build GRCh38
rs939671633 Chr.9: 36164025-36164025 on Build GRCh38
rs779299201 Chr.9: 36163819-36163819 on Build GRCh38
rs10814343 Chr.9: 36163942-36163942 on Build GRCh38
rs28669721 Chr.9: 36163941-36163941 on Build GRCh38
rs778683346 Chr.9: 36164008-36164008 on Build GRCh38
rs532024525 Chr.9: 36164143-36164143 on Build GRCh38
rs576795811 Chr.9: 36164077-36164077 on Build GRCh38
rs957824564 Chr.9: 36163928-36163928 on Build GRCh38
rs546777969 Chr.9: 36163958-36163958 on Build GRCh38
rs566779388 Chr.9: 36163883-36163885 on Build GRCh38
rs28683175 Chr.9: 36163932-36163932 on Build GRCh38
rs539760133 Chr.9: 36163818-36163818 on Build GRCh38
rs1007980642 Chr.9: 36164213-36164213 on Build GRCh38
rs576151310 Chr.9: 36164233-36164233 on Build GRCh38
rs879705364 Chr.9: 36164012-36164012 on Build GRCh38
rs562289121 Chr.9: 36164158-36164158 on Build GRCh38
rs74937378 Chr.9: 36164152-36164152 on Build GRCh38
rs758977974 Chr.9: 36163892-36163892 on Build GRCh38
rs535330021 Chr.9: 36163934-36163934 on Build GRCh38
rs77932772 Chr.9: 36164267-36164267 on Build GRCh38
rs889649566 Chr.9: 36164170-36164170 on Build GRCh38
rs919849252 Chr.9: 36164052-36164052 on Build GRCh38
rs541766754 Chr.9: 36164268-36164268 on Build GRCh38
rs746329536 Chr.9: 36163870-36163870 on Build GRCh38
rs1037081486 Chr.9: 36164046-36164046 on Build GRCh38
rs770814958 Chr.9: 36163978-36163978 on Build GRCh38
rs570446639 Chr.9: 36163814-36163814 on Build GRCh38
rs1049663445 Chr.9: 36164164-36164164 on Build GRCh38
rs768947170 Chr.9: 36164299-36164299 on Build GRCh38

View Full Product Details