Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 36339479-36339986 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF38
Entrez Gene ID
152006
Gene Name
ring finger protein 38
Gene Aliases
-
UniGene
Hs.333503
Gene Chromosome Location
Chr.9: 36336398-36487384 on Build GRCh38
Transcripts
NM_022781.4 NM_194328.2 NM_194329.2 NM_194330.2 NM_194332.2 XM_005251366.3 XM_005251367.3 XM_005251368.3 XM_006716721.3 XM_011517712.2 XM_011517713.2 XM_017014294.1 XM_017014295.1 XM_017014296.1 XM_017014297.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6517103 c.1295C>T p.S432L Chr.9: 36339777-36339777 on Build GRCh38
150607 c.1487-10G>T p.? Chr.9: 36339823-36339823 on Build GRCh38
3907334 c.1306C>T p.R436W Chr.9: 36339766-36339766 on Build GRCh38
4722191 c.1282C>T p.R428* Chr.9: 36339790-36339790 on Build GRCh38
4722190 c.1510C>T p.R504* Chr.9: 36339790-36339790 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781609645 Chr.9: 36339718-36339718 on Build GRCh38
rs866628481 Chr.9: 36339526-36339526 on Build GRCh38
rs763571220 Chr.9: 36339759-36339759 on Build GRCh38
rs191461722 Chr.9: 36339823-36339823 on Build GRCh38
rs746361613 Chr.9: 36339857-36339857 on Build GRCh38
rs750685968 Chr.9: 36339639-36339639 on Build GRCh38
rs200646561 Chr.9: 36339741-36339741 on Build GRCh38
rs930958380 Chr.9: 36339556-36339556 on Build GRCh38
rs748363779 Chr.9: 36339719-36339719 on Build GRCh38
rs1026210211 Chr.9: 36339898-36339899 on Build GRCh38
rs1011536384 Chr.9: 36339689-36339689 on Build GRCh38
rs751515740 Chr.9: 36339820-36339820 on Build GRCh38
rs749574071 Chr.9: 36339851-36339851 on Build GRCh38
rs373555424 Chr.9: 36339714-36339714 on Build GRCh38
rs572944003 Chr.9: 36339890-36339890 on Build GRCh38
rs765399702 Chr.9: 36339554-36339554 on Build GRCh38
rs372003346 Chr.9: 36339643-36339643 on Build GRCh38
rs1026821150 Chr.9: 36339875-36339875 on Build GRCh38
rs1007716018 Chr.9: 36339841-36339841 on Build GRCh38
rs765085808 Chr.9: 36339790-36339790 on Build GRCh38
rs892647860 Chr.9: 36339797-36339797 on Build GRCh38
rs943780130 Chr.9: 36339579-36339579 on Build GRCh38
rs571616511 Chr.9: 36339742-36339742 on Build GRCh38
rs183303684 Chr.9: 36339880-36339880 on Build GRCh38
rs534719661 Chr.9: 36339691-36339691 on Build GRCh38
rs1053333510 Chr.9: 36339657-36339657 on Build GRCh38
rs771294973 Chr.9: 36339852-36339852 on Build GRCh38
rs1035233909 Chr.9: 36339527-36339527 on Build GRCh38
rs749811942 Chr.9: 36339737-36339737 on Build GRCh38
rs956829507 Chr.9: 36339537-36339537 on Build GRCh38
rs1035766180 Chr.9: 36339812-36339812 on Build GRCh38
rs767031982 Chr.9: 36339896-36339896 on Build GRCh38
rs567454592 Chr.9: 36339548-36339548 on Build GRCh38
rs774631272 Chr.9: 36339598-36339598 on Build GRCh38
rs1040841789 Chr.9: 36339638-36339638 on Build GRCh38
rs770256448 Chr.9: 36339728-36339728 on Build GRCh38
rs919097121 Chr.9: 36339594-36339594 on Build GRCh38
rs747039180 Chr.9: 36339707-36339707 on Build GRCh38
rs985534903 Chr.9: 36339562-36339562 on Build GRCh38
rs878899923 Chr.9: 36339623-36339623 on Build GRCh38
rs371083999 Chr.9: 36339829-36339829 on Build GRCh38
rs529399904 Chr.9: 36339555-36339555 on Build GRCh38
rs567183971 Chr.9: 36339659-36339659 on Build GRCh38
rs893409105 Chr.9: 36339674-36339674 on Build GRCh38
rs760218308 Chr.9: 36339748-36339748 on Build GRCh38
rs557317526 Chr.9: 36339839-36339839 on Build GRCh38
rs773380104 Chr.9: 36339731-36339731 on Build GRCh38
rs761452413 Chr.9: 36339768-36339768 on Build GRCh38
rs976404347 Chr.9: 36339592-36339592 on Build GRCh38
rs910879454 Chr.9: 36339935-36339935 on Build GRCh38
rs748383941 Chr.9: 36339836-36339836 on Build GRCh38
rs200434728 Chr.9: 36339796-36339796 on Build GRCh38
rs776323171 Chr.9: 36339765-36339765 on Build GRCh38
rs910940946 Chr.9: 36339578-36339578 on Build GRCh38
rs79193527 Chr.9: 36339699-36339699 on Build GRCh38
rs544904345 Chr.9: 36339700-36339700 on Build GRCh38
rs910197636 Chr.9: 36339571-36339571 on Build GRCh38
rs758385415 Chr.9: 36339803-36339803 on Build GRCh38
rs202022976 Chr.9: 36339858-36339858 on Build GRCh38
rs574153653 Chr.9: 36339846-36339846 on Build GRCh38
rs950593021 Chr.9: 36339949-36339950 on Build GRCh38
rs934842682 Chr.9: 36339644-36339644 on Build GRCh38
rs952748658 Chr.9: 36339879-36339879 on Build GRCh38
rs757768045 Chr.9: 36339850-36339863 on Build GRCh38

View Full Product Details