Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 39085545-39086052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTNAP3
Entrez Gene ID
79937
Gene Name
contactin associated protein-like 3
Gene Aliases
CASPR3, CNTNAP3A
UniGene
Hs.128474
Gene Chromosome Location
Chr.9: 39072767-39288303 on Build GRCh38
Transcripts
NM_033655.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2776207 c.3408C>T p.N1136N Chr.9: 39085770-39085770 on Build GRCh38
1109156 c.3390C>A p.S1130S Chr.9: 39085788-39085788 on Build GRCh38
487470 c.3403T>G p.F1135V Chr.9: 39085775-39085775 on Build GRCh38
1109157 c.3370A>C p.K1124Q Chr.9: 39085808-39085808 on Build GRCh38
5427978 c.3367A>G p.T1123A Chr.9: 39085811-39085811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs939033178 Chr.9: 39085975-39085975 on Build GRCh38
rs771568256 Chr.9: 39085700-39085700 on Build GRCh38
rs746636706 Chr.9: 39085797-39085797 on Build GRCh38
rs2150942 Chr.9: 39085767-39085767 on Build GRCh38
rs570590345 Chr.9: 39085706-39085706 on Build GRCh38
rs3011117 Chr.9: 39085984-39085984 on Build GRCh38
rs747693399 Chr.9: 39085821-39085821 on Build GRCh38
rs368875525 Chr.9: 39086013-39086013 on Build GRCh38
rs866274595 Chr.9: 39085674-39085674 on Build GRCh38
rs767541305 Chr.9: 39085769-39085769 on Build GRCh38
rs369994235 Chr.9: 39085705-39085705 on Build GRCh38
rs984302673 Chr.9: 39085927-39085927 on Build GRCh38
rs1025730524 Chr.9: 39085901-39085901 on Build GRCh38
rs892253253 Chr.9: 39085812-39085812 on Build GRCh38
rs939131179 Chr.9: 39085698-39085698 on Build GRCh38
rs765247293 Chr.9: 39085743-39085743 on Build GRCh38
rs757693099 Chr.9: 39085777-39085777 on Build GRCh38
rs758830553 Chr.9: 39085873-39085875 on Build GRCh38
rs761362855 Chr.9: 39085965-39085968 on Build GRCh38
rs1044775448 Chr.9: 39085602-39085602 on Build GRCh38
rs373751434 Chr.9: 39085707-39085707 on Build GRCh38
rs184369252 Chr.9: 39085835-39085835 on Build GRCh38
rs947674161 Chr.9: 39085590-39085590 on Build GRCh38
rs980454892 Chr.9: 39085966-39085966 on Build GRCh38
rs913662062 Chr.9: 39086018-39086018 on Build GRCh38
rs1043592628 Chr.9: 39085862-39085862 on Build GRCh38
rs774157549 Chr.9: 39085866-39085866 on Build GRCh38
rs371184544 Chr.9: 39085770-39085770 on Build GRCh38
rs368796414 Chr.9: 39085766-39085766 on Build GRCh38
rs557397704 Chr.9: 39085924-39085924 on Build GRCh38
rs181242281 Chr.9: 39085771-39085771 on Build GRCh38
rs969667526 Chr.9: 39085962-39085962 on Build GRCh38
rs3071045 Chr.9: 39085706-39085707 on Build GRCh38
rs536768522 Chr.9: 39085635-39085635 on Build GRCh38
rs910154871 Chr.9: 39085578-39085578 on Build GRCh38
rs772812012 Chr.9: 39085754-39085754 on Build GRCh38
rs2183891 Chr.9: 39085936-39085936 on Build GRCh38
rs372421268 Chr.9: 39085813-39085813 on Build GRCh38
rs927732333 Chr.9: 39085629-39085629 on Build GRCh38
rs998645107 Chr.9: 39085899-39085899 on Build GRCh38
rs775269027 Chr.9: 39085702-39085702 on Build GRCh38
rs927584344 Chr.9: 39085973-39085973 on Build GRCh38
rs951502756 Chr.9: 39085920-39085920 on Build GRCh38
rs535709976 Chr.9: 39085890-39085890 on Build GRCh38
rs757245093 Chr.9: 39085671-39085671 on Build GRCh38
rs777959942 Chr.9: 39085696-39085696 on Build GRCh38
rs749820526 Chr.9: 39085775-39085775 on Build GRCh38
rs773184117 Chr.9: 39085846-39085846 on Build GRCh38
rs775372621 Chr.9: 39086009-39086009 on Build GRCh38
rs749668640 Chr.9: 39085697-39085697 on Build GRCh38
rs757315598 Chr.9: 39085702-39085703 on Build GRCh38
rs562669127 Chr.9: 39085926-39085926 on Build GRCh38
rs746724228 Chr.9: 39085703-39085703 on Build GRCh38
rs368138929 Chr.9: 39085874-39085874 on Build GRCh38
rs910123016 Chr.9: 39085951-39085951 on Build GRCh38
rs779458691 Chr.9: 39085783-39085783 on Build GRCh38
rs1018733060 Chr.9: 39085883-39085883 on Build GRCh38
rs376717524 Chr.9: 39085864-39085864 on Build GRCh38
rs1412361 Chr.9: 39085858-39085858 on Build GRCh38
rs1412360 Chr.9: 39085811-39085811 on Build GRCh38
rs1029068 Chr.9: 39085570-39085570 on Build GRCh38

View Full Product Details