Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 112079936-112080384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUSD1
Entrez Gene ID
64420
Gene Name
sushi domain containing 1
Gene Aliases
-
UniGene
Hs.494827
Gene Chromosome Location
Chr.9: 112040781-112175520 on Build GRCh38
Transcripts
NM_001282640.1 NM_001282643.1 NM_022486.4 XM_011518915.1 XM_011518916.2 XM_011518917.1 XM_011518918.1 XM_011518919.1 XM_017015037.1 XM_017015038.1 XM_017015039.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7143725 c.1536C>G p.I512M Chr.9: 112080104-112080104 on Build GRCh38
6793377 c.1541C>T p.T514I Chr.9: 112080099-112080099 on Build GRCh38
7143724 c.1536C>G p.I512M Chr.9: 112080104-112080104 on Build GRCh38
6793376 c.1541C>T p.T514I Chr.9: 112080099-112080099 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1014029271 Chr.9: 112080246-112080246 on Build GRCh38
rs60956063 Chr.9: 112080173-112080173 on Build GRCh38
rs374502979 Chr.9: 112080145-112080145 on Build GRCh38
rs369464770 Chr.9: 112080336-112080336 on Build GRCh38
rs775294819 Chr.9: 112080105-112080105 on Build GRCh38
rs919359977 Chr.9: 112080015-112080015 on Build GRCh38
rs373500000 Chr.9: 112080205-112080205 on Build GRCh38
rs569585463 Chr.9: 112080361-112080361 on Build GRCh38
rs757506490 Chr.9: 112080209-112080209 on Build GRCh38
rs550703909 Chr.9: 112080207-112080208 on Build GRCh38
rs771321740 Chr.9: 112080080-112080080 on Build GRCh38
rs749420214 Chr.9: 112080206-112080206 on Build GRCh38
rs562166060 Chr.9: 112080009-112080010 on Build GRCh38
rs777970899 Chr.9: 112080066-112080066 on Build GRCh38
rs774250276 Chr.9: 112080090-112080090 on Build GRCh38
rs201153383 Chr.9: 112080183-112080183 on Build GRCh38
rs539603383 Chr.9: 112080278-112080278 on Build GRCh38
rs375682807 Chr.9: 112080019-112080020 on Build GRCh38
rs548531031 Chr.9: 112080042-112080042 on Build GRCh38
rs199528391 Chr.9: 112080053-112080053 on Build GRCh38
rs763488837 Chr.9: 112080142-112080142 on Build GRCh38
rs1009836551 Chr.9: 112080357-112080357 on Build GRCh38
rs756638372 Chr.9: 112080270-112080270 on Build GRCh38
rs556342510 Chr.9: 112080298-112080298 on Build GRCh38
rs755170369 Chr.9: 112080348-112080348 on Build GRCh38
rs550964139 Chr.9: 112080297-112080297 on Build GRCh38
rs1056655573 Chr.9: 112080339-112080339 on Build GRCh38
rs773979503 Chr.9: 112080055-112080056 on Build GRCh38
rs926810105 Chr.9: 112080321-112080321 on Build GRCh38
rs748188299 Chr.9: 112080055-112080055 on Build GRCh38
rs1002834649 Chr.9: 112080310-112080310 on Build GRCh38
rs377318056 Chr.9: 112080216-112080216 on Build GRCh38
rs892698259 Chr.9: 112080345-112080345 on Build GRCh38
rs762068940 Chr.9: 112080104-112080104 on Build GRCh38
rs756697131 Chr.9: 112080196-112080196 on Build GRCh38
rs201785262 Chr.9: 112080113-112080113 on Build GRCh38
rs759427914 Chr.9: 112080101-112080101 on Build GRCh38
rs569329796 Chr.9: 112079995-112079995 on Build GRCh38
rs753923208 Chr.9: 112080304-112080304 on Build GRCh38
rs200677918 Chr.9: 112080154-112080154 on Build GRCh38
rs755336246 Chr.9: 112080164-112080164 on Build GRCh38
rs751950178 Chr.9: 112080051-112080051 on Build GRCh38
rs933305703 Chr.9: 112080026-112080026 on Build GRCh38
rs769762461 Chr.9: 112080056-112080056 on Build GRCh38
rs531736492 Chr.9: 112080003-112080003 on Build GRCh38
rs570703173 Chr.9: 112080236-112080236 on Build GRCh38
rs750097284 Chr.9: 112079989-112079989 on Build GRCh38
rs201054428 Chr.9: 112080103-112080103 on Build GRCh38
rs905389759 Chr.9: 112079979-112079979 on Build GRCh38
rs534304613 Chr.9: 112079966-112079966 on Build GRCh38
rs369751150 Chr.9: 112080160-112080160 on Build GRCh38
rs749451680 Chr.9: 112080075-112080075 on Build GRCh38
rs761812350 Chr.9: 112080083-112080085 on Build GRCh38

View Full Product Details