Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 120600800-120601344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEGF9
Entrez Gene ID
1955
Gene Name
multiple EGF like domains 9
Gene Aliases
EGFL5
UniGene
Hs.744903
Gene Chromosome Location
Chr.9: 120600918-120714487 on Build GRCh38
Transcripts
NM_001080497.2

SNP Detail

SNP ID Chromosome Location
rs916097536 Chr.9: 120601313-120601313 on Build GRCh38
rs555183263 Chr.9: 120600890-120600890 on Build GRCh38
rs1035984904 Chr.9: 120600922-120600922 on Build GRCh38
rs1047596281 Chr.9: 120600955-120600955 on Build GRCh38
rs938760447 Chr.9: 120601009-120601009 on Build GRCh38
rs952544409 Chr.9: 120601020-120601022 on Build GRCh38
rs536849712 Chr.9: 120600860-120600860 on Build GRCh38
rs973337990 Chr.9: 120601222-120601222 on Build GRCh38
rs928606376 Chr.9: 120601283-120601283 on Build GRCh38
rs1055777037 Chr.9: 120601307-120601307 on Build GRCh38
rs910385385 Chr.9: 120600894-120600894 on Build GRCh38
rs921211368 Chr.9: 120601040-120601040 on Build GRCh38
rs982554101 Chr.9: 120601024-120601024 on Build GRCh38
rs74602937 Chr.9: 120601020-120601020 on Build GRCh38
rs1057382790 Chr.9: 120601196-120601196 on Build GRCh38
rs1007706014 Chr.9: 120601133-120601133 on Build GRCh38
rs569251482 Chr.9: 120601016-120601019 on Build GRCh38
rs1017480072 Chr.9: 120601135-120601135 on Build GRCh38
rs10616 Chr.9: 120601255-120601255 on Build GRCh38
rs1003119629 Chr.9: 120600901-120600901 on Build GRCh38
rs560017664 Chr.9: 120601223-120601223 on Build GRCh38
rs973305612 Chr.9: 120600899-120600899 on Build GRCh38
rs568189519 Chr.9: 120601229-120601232 on Build GRCh38
rs540997830 Chr.9: 120601151-120601151 on Build GRCh38
rs144096339 Chr.9: 120601055-120601055 on Build GRCh38
rs902858939 Chr.9: 120601100-120601100 on Build GRCh38
rs368811308 Chr.9: 120600874-120600874 on Build GRCh38
rs963316333 Chr.9: 120600895-120600895 on Build GRCh38
rs976632884 Chr.9: 120601038-120601038 on Build GRCh38
rs576556071 Chr.9: 120600903-120600903 on Build GRCh38
rs148160212 Chr.9: 120600915-120600915 on Build GRCh38
rs903347855 Chr.9: 120601157-120601157 on Build GRCh38
rs998498805 Chr.9: 120601117-120601117 on Build GRCh38
rs36002962 Chr.9: 120601055-120601056 on Build GRCh38
rs1030403414 Chr.9: 120601118-120601118 on Build GRCh38
rs12347416 Chr.9: 120601033-120601033 on Build GRCh38
rs963180934 Chr.9: 120601191-120601191 on Build GRCh38
rs372486888 Chr.9: 120601129-120601129 on Build GRCh38
rs894701093 Chr.9: 120601202-120601202 on Build GRCh38
rs192207077 Chr.9: 120601030-120601030 on Build GRCh38
rs79854335 Chr.9: 120600847-120600847 on Build GRCh38
rs1043264893 Chr.9: 120601069-120601069 on Build GRCh38
rs1004360763 Chr.9: 120601274-120601274 on Build GRCh38
rs982841702 Chr.9: 120601257-120601257 on Build GRCh38
rs774339957 Chr.9: 120600938-120600938 on Build GRCh38
rs938808653 Chr.9: 120601299-120601299 on Build GRCh38
rs928768413 Chr.9: 120600969-120600969 on Build GRCh38
rs894634304 Chr.9: 120601032-120601032 on Build GRCh38
rs771508779 Chr.9: 120601086-120601086 on Build GRCh38
rs774945740 Chr.9: 120601249-120601249 on Build GRCh38

View Full Product Details