Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 120824233-120824737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMD5
Entrez Gene ID
5711
Gene Name
proteasome 26S subunit, non-ATPase 5
Gene Aliases
S5B
UniGene
Hs.193725
Gene Chromosome Location
Chr.9: 120798318-120843021 on Build GRCh38
Transcripts
NM_001270427.1 NM_005047.3 XM_011518866.2 XM_017014927.1 XM_017014928.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3929781 c.150C>A p.N50K Chr.9: 120824663-120824663 on Build GRCh38
3745801 c.149A>C p.N50T Chr.9: 120824664-120824664 on Build GRCh38
4718497 c.144_145insT p.G49fs*8 Chr.9: 120824668-120824669 on Build GRCh38
4588411 c.143T>A p.F48Y Chr.9: 120824670-120824670 on Build GRCh38
3745800 c.836A>C p.N279T Chr.9: 120824664-120824664 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs869269639 Chr.9: 120824319-120824320 on Build GRCh38
rs1000996779 Chr.9: 120824303-120824303 on Build GRCh38
rs189500645 Chr.9: 120824523-120824523 on Build GRCh38
rs773069672 Chr.9: 120824687-120824687 on Build GRCh38
rs746572807 Chr.9: 120824596-120824596 on Build GRCh38
rs757201696 Chr.9: 120824386-120824386 on Build GRCh38
rs980393345 Chr.9: 120824311-120824311 on Build GRCh38
rs200818857 Chr.9: 120824449-120824449 on Build GRCh38
rs759788128 Chr.9: 120824522-120824522 on Build GRCh38
rs757397524 Chr.9: 120824562-120824562 on Build GRCh38
rs755449750 Chr.9: 120824329-120824329 on Build GRCh38
rs773400263 Chr.9: 120824304-120824306 on Build GRCh38
rs750413180 Chr.9: 120824566-120824566 on Build GRCh38
rs149552567 Chr.9: 120824680-120824680 on Build GRCh38
rs912252355 Chr.9: 120824584-120824584 on Build GRCh38
rs766977934 Chr.9: 120824460-120824460 on Build GRCh38
rs775378883 Chr.9: 120824626-120824626 on Build GRCh38
rs752822667 Chr.9: 120824525-120824525 on Build GRCh38
rs559333970 Chr.9: 120824534-120824534 on Build GRCh38
rs764023748 Chr.9: 120824529-120824529 on Build GRCh38
rs35260594 Chr.9: 120824635-120824635 on Build GRCh38
rs758356338 Chr.9: 120824676-120824676 on Build GRCh38
rs544079478 Chr.9: 120824516-120824516 on Build GRCh38
rs771946655 Chr.9: 120824416-120824416 on Build GRCh38
rs550137285 Chr.9: 120824304-120824305 on Build GRCh38
rs1052647426 Chr.9: 120824655-120824655 on Build GRCh38
rs572450000 Chr.9: 120824445-120824445 on Build GRCh38
rs539127458 Chr.9: 120824328-120824328 on Build GRCh38
rs371296805 Chr.9: 120824602-120824602 on Build GRCh38
rs150588580 Chr.9: 120824625-120824625 on Build GRCh38
rs963115132 Chr.9: 120824340-120824340 on Build GRCh38
rs746693947 Chr.9: 120824446-120824446 on Build GRCh38
rs866608724 Chr.9: 120824383-120824383 on Build GRCh38
rs760120806 Chr.9: 120824621-120824621 on Build GRCh38
rs113401299 Chr.9: 120824456-120824456 on Build GRCh38
rs971961397 Chr.9: 120824368-120824368 on Build GRCh38
rs10760117 Chr.9: 120824459-120824459 on Build GRCh38
rs35903891 Chr.9: 120824304-120824304 on Build GRCh38
rs372310762 Chr.9: 120824681-120824681 on Build GRCh38
rs774975928 Chr.9: 120824608-120824608 on Build GRCh38
rs535194298 Chr.9: 120824303-120824303 on Build GRCh38
rs747654438 Chr.9: 120824607-120824607 on Build GRCh38
rs775063677 Chr.9: 120824475-120824475 on Build GRCh38
rs778795627 Chr.9: 120824565-120824565 on Build GRCh38
rs761863729 Chr.9: 120824653-120824653 on Build GRCh38
rs753904860 Chr.9: 120824556-120824556 on Build GRCh38
rs781047713 Chr.9: 120824606-120824606 on Build GRCh38
rs79329938 Chr.9: 120824306-120824306 on Build GRCh38
rs777940231 Chr.9: 120824371-120824371 on Build GRCh38
rs760851585 Chr.9: 120824304-120824307 on Build GRCh38
rs753003023 Chr.9: 120824342-120824342 on Build GRCh38
rs956347132 Chr.9: 120824563-120824563 on Build GRCh38
rs765327655 Chr.9: 120824654-120824654 on Build GRCh38
rs756099935 Chr.9: 120824353-120824353 on Build GRCh38
rs942711989 Chr.9: 120824555-120824555 on Build GRCh38
rs376146984 Chr.9: 120824437-120824437 on Build GRCh38
rs756365476 Chr.9: 120824526-120824526 on Build GRCh38
rs969592118 Chr.9: 120824506-120824506 on Build GRCh38
rs908967895 Chr.9: 120824327-120824327 on Build GRCh38
rs749599444 Chr.9: 120824431-120824431 on Build GRCh38
rs921063046 Chr.9: 120824639-120824639 on Build GRCh38
rs771295108 Chr.9: 120824435-120824435 on Build GRCh38
rs749158783 Chr.9: 120824372-120824372 on Build GRCh38
rs761837817 Chr.9: 120824634-120824634 on Build GRCh38
rs778232658 Chr.9: 120824423-120824423 on Build GRCh38
rs368077721 Chr.9: 120824573-120824573 on Build GRCh38
rs778883387 Chr.9: 120824395-120824395 on Build GRCh38
rs750317182 Chr.9: 120824669-120824669 on Build GRCh38
rs554256815 Chr.9: 120824333-120824333 on Build GRCh38
rs780959709 Chr.9: 120824714-120824714 on Build GRCh38
rs759978461 Chr.9: 120824467-120824467 on Build GRCh38
rs764298853 Chr.9: 120824643-120824643 on Build GRCh38
rs777257491 Chr.9: 120824448-120824449 on Build GRCh38
rs756894174 Chr.9: 120824421-120824421 on Build GRCh38
rs370927809 Chr.9: 120824583-120824583 on Build GRCh38
rs374398274 Chr.9: 120824652-120824652 on Build GRCh38
rs748336939 Chr.9: 120824303-120824304 on Build GRCh38
rs745660870 Chr.9: 120824409-120824409 on Build GRCh38
rs771341026 Chr.9: 120824323-120824324 on Build GRCh38

View Full Product Details