Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 120909038-120909538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAF1
Entrez Gene ID
7185
Gene Name
TNF receptor associated factor 1
Gene Aliases
EBI6, MGC:10353
UniGene
Hs.531251
Gene Chromosome Location
Chr.9: 120902393-120929173 on Build GRCh38
Transcripts
NM_001190945.1 NM_001190947.1 NM_005658.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1104699 c.969G>A p.S323S Chr.9: 120909293-120909293 on Build GRCh38
112041 c.885_886insC p.F296fs*39 Chr.9: 120909376-120909377 on Build GRCh38
3926088 c.919C>T p.L307L Chr.9: 120909343-120909343 on Build GRCh38
6807192 c.885C>A p.A295A Chr.9: 120909377-120909377 on Build GRCh38
1744606 c.953A>G p.K318R Chr.9: 120909309-120909309 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372991526 Chr.9: 120909386-120909386 on Build GRCh38
rs1036295518 Chr.9: 120909493-120909493 on Build GRCh38
rs200187170 Chr.9: 120909294-120909294 on Build GRCh38
rs529690461 Chr.9: 120909169-120909169 on Build GRCh38
rs762688850 Chr.9: 120909251-120909251 on Build GRCh38
rs61749211 Chr.9: 120909237-120909237 on Build GRCh38
rs12377786 Chr.9: 120909481-120909481 on Build GRCh38
rs139440050 Chr.9: 120909350-120909350 on Build GRCh38
rs779613531 Chr.9: 120909185-120909185 on Build GRCh38
rs2239657 Chr.9: 120909242-120909242 on Build GRCh38
rs1029754909 Chr.9: 120909196-120909196 on Build GRCh38
rs201073899 Chr.9: 120909284-120909284 on Build GRCh38
rs758964655 Chr.9: 120909422-120909422 on Build GRCh38
rs771544516 Chr.9: 120909209-120909209 on Build GRCh38
rs576606864 Chr.9: 120909453-120909453 on Build GRCh38
rs750167887 Chr.9: 120909179-120909179 on Build GRCh38
rs796697504 Chr.9: 120909070-120909070 on Build GRCh38
rs766369886 Chr.9: 120909364-120909364 on Build GRCh38
rs762846377 Chr.9: 120909353-120909353 on Build GRCh38
rs1052073386 Chr.9: 120909505-120909505 on Build GRCh38
rs929169236 Chr.9: 120909137-120909137 on Build GRCh38
rs897622388 Chr.9: 120909501-120909501 on Build GRCh38
rs201090941 Chr.9: 120909181-120909181 on Build GRCh38
rs540711208 Chr.9: 120909123-120909123 on Build GRCh38
rs531083712 Chr.9: 120909197-120909197 on Build GRCh38
rs1036587839 Chr.9: 120909172-120909172 on Build GRCh38
rs757480964 Chr.9: 120909184-120909184 on Build GRCh38
rs897644283 Chr.9: 120909128-120909128 on Build GRCh38
rs1046227072 Chr.9: 120909141-120909141 on Build GRCh38
rs144954439 Chr.9: 120909468-120909468 on Build GRCh38
rs1044747284 Chr.9: 120909149-120909149 on Build GRCh38
rs746395490 Chr.9: 120909293-120909293 on Build GRCh38
rs149068020 Chr.9: 120909503-120909503 on Build GRCh38
rs568283719 Chr.9: 120909248-120909248 on Build GRCh38
rs112060909 Chr.9: 120909335-120909335 on Build GRCh38
rs941924108 Chr.9: 120909068-120909068 on Build GRCh38
rs34536586 Chr.9: 120909258-120909258 on Build GRCh38
rs773602200 Chr.9: 120909513-120909513 on Build GRCh38
rs182675772 Chr.9: 120909249-120909249 on Build GRCh38
rs780627684 Chr.9: 120909425-120909425 on Build GRCh38
rs143118846 Chr.9: 120909514-120909514 on Build GRCh38
rs769885544 Chr.9: 120909349-120909349 on Build GRCh38
rs541831117 Chr.9: 120909487-120909487 on Build GRCh38
rs781310877 Chr.9: 120909411-120909411 on Build GRCh38
rs779587375 Chr.9: 120909226-120909226 on Build GRCh38
rs967528095 Chr.9: 120909202-120909202 on Build GRCh38
rs747468741 Chr.9: 120909303-120909303 on Build GRCh38
rs754581552 Chr.9: 120909280-120909280 on Build GRCh38
rs75115807 Chr.9: 120909403-120909403 on Build GRCh38
rs775727698 Chr.9: 120909235-120909235 on Build GRCh38
rs761843237 Chr.9: 120909347-120909347 on Build GRCh38
rs759387426 Chr.9: 120909368-120909368 on Build GRCh38
rs200389282 Chr.9: 120909221-120909221 on Build GRCh38
rs553024344 Chr.9: 120909469-120909469 on Build GRCh38
rs376238472 Chr.9: 120909414-120909414 on Build GRCh38
rs999112477 Chr.9: 120909479-120909479 on Build GRCh38
rs751410769 Chr.9: 120909263-120909263 on Build GRCh38
rs908907268 Chr.9: 120909457-120909457 on Build GRCh38
rs780281725 Chr.9: 120909300-120909300 on Build GRCh38
rs779519689 Chr.9: 120909291-120909291 on Build GRCh38
rs941715413 Chr.9: 120909250-120909250 on Build GRCh38
rs547111635 Chr.9: 120909283-120909283 on Build GRCh38
rs369428051 Chr.9: 120909238-120909238 on Build GRCh38
rs116944279 Chr.9: 120909385-120909385 on Build GRCh38
rs907271743 Chr.9: 120909151-120909151 on Build GRCh38
rs772573228 Chr.9: 120909229-120909229 on Build GRCh38
rs559115469 Chr.9: 120909159-120909159 on Build GRCh38
rs903413901 Chr.9: 120909470-120909470 on Build GRCh38
rs947839657 Chr.9: 120909423-120909423 on Build GRCh38
rs776918483 Chr.9: 120909340-120909340 on Build GRCh38
rs766140398 Chr.9: 120909257-120909257 on Build GRCh38
rs1021910430 Chr.9: 120909194-120909194 on Build GRCh38
rs894499348 Chr.9: 120909171-120909171 on Build GRCh38
rs569958951 Chr.9: 120909365-120909365 on Build GRCh38
rs376078303 Chr.9: 120909199-120909199 on Build GRCh38
rs201867327 Chr.9: 120909193-120909193 on Build GRCh38
rs767009422 Chr.9: 120909374-120909374 on Build GRCh38
rs938769594 Chr.9: 120909316-120909316 on Build GRCh38
rs755686930 Chr.9: 120909287-120909287 on Build GRCh38

View Full Product Details