Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 124482309-124482815 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR5A1
Entrez Gene ID
2516
Gene Name
nuclear receptor subfamily 5 group A member 1
Gene Aliases
AD4BP, ELP, FTZ1, FTZF1, POF7, SF-1, SF1, SPGF8, SRXY3, hSF-1
UniGene
Hs.495108
Gene Chromosome Location
Chr.9: 124481234-124507420 on Build GRCh38
Transcripts
NM_004959.4 XM_005251871.3 XM_011518455.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4708049 c.707T>C p.L236P Chr.9: 124482789-124482789 on Build GRCh38
4708048 c.1355T>C p.L452P Chr.9: 124482789-124482789 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs950903307 Chr.9: 124482515-124482515 on Build GRCh38
rs5900617 Chr.9: 124482585-124482585 on Build GRCh38
rs903016039 Chr.9: 124482444-124482444 on Build GRCh38
rs972209072 Chr.9: 124482505-124482505 on Build GRCh38
rs930127350 Chr.9: 124482522-124482522 on Build GRCh38
rs893191829 Chr.9: 124482433-124482433 on Build GRCh38
rs1052883677 Chr.9: 124482403-124482403 on Build GRCh38
rs963098275 Chr.9: 124482500-124482500 on Build GRCh38
rs766512961 Chr.9: 124482742-124482742 on Build GRCh38
rs754099590 Chr.9: 124482727-124482728 on Build GRCh38
rs756976922 Chr.9: 124482486-124482486 on Build GRCh38
rs904046758 Chr.9: 124482708-124482708 on Build GRCh38
rs988924070 Chr.9: 124482647-124482647 on Build GRCh38
rs570774964 Chr.9: 124482710-124482721 on Build GRCh38
rs759904693 Chr.9: 124482734-124482745 on Build GRCh38
rs1046008206 Chr.9: 124482340-124482340 on Build GRCh38
rs201169860 Chr.9: 124482584-124482584 on Build GRCh38
rs1030165651 Chr.9: 124482455-124482455 on Build GRCh38
rs947440291 Chr.9: 124482654-124482654 on Build GRCh38
rs867106298 Chr.9: 124482771-124482771 on Build GRCh38
rs773528915 Chr.9: 124482719-124482719 on Build GRCh38
rs915034 Chr.9: 124482676-124482676 on Build GRCh38
rs1016359943 Chr.9: 124482497-124482497 on Build GRCh38
rs928839553 Chr.9: 124482529-124482529 on Build GRCh38
rs759390100 Chr.9: 124482756-124482756 on Build GRCh38
rs922898885 Chr.9: 124482567-124482567 on Build GRCh38
rs919044760 Chr.9: 124482517-124482517 on Build GRCh38
rs77800186 Chr.9: 124482665-124482665 on Build GRCh38
rs903277804 Chr.9: 124482673-124482673 on Build GRCh38
rs1030134357 Chr.9: 124482681-124482681 on Build GRCh38
rs1043869829 Chr.9: 124482696-124482706 on Build GRCh38
rs767255033 Chr.9: 124482719-124482721 on Build GRCh38
rs891575406 Chr.9: 124482409-124482409 on Build GRCh38
rs953303810 Chr.9: 124482483-124482483 on Build GRCh38
rs1002421521 Chr.9: 124482476-124482476 on Build GRCh38
rs146454575 Chr.9: 124482765-124482765 on Build GRCh38
rs961782970 Chr.9: 124482554-124482554 on Build GRCh38
rs367957279 Chr.9: 124482741-124482741 on Build GRCh38
rs1026390658 Chr.9: 124482507-124482507 on Build GRCh38
rs1055310823 Chr.9: 124482749-124482749 on Build GRCh38
rs907495226 Chr.9: 124482391-124482391 on Build GRCh38
rs755859743 Chr.9: 124482768-124482768 on Build GRCh38
rs774409128 Chr.9: 124482746-124482746 on Build GRCh38
rs79833327 Chr.9: 124482791-124482791 on Build GRCh38
rs796517206 Chr.9: 124482586-124482586 on Build GRCh38
rs984623194 Chr.9: 124482521-124482521 on Build GRCh38
rs770164832 Chr.9: 124482713-124482713 on Build GRCh38
rs563606221 Chr.9: 124482717-124482717 on Build GRCh38
rs763737945 Chr.9: 124482775-124482775 on Build GRCh38
rs1020392354 Chr.9: 124482440-124482440 on Build GRCh38
rs1055775159 Chr.9: 124482560-124482560 on Build GRCh38
rs997342835 Chr.9: 124482452-124482452 on Build GRCh38
rs1056208474 Chr.9: 124482401-124482401 on Build GRCh38
rs554329124 Chr.9: 124482434-124482434 on Build GRCh38
rs984392900 Chr.9: 124482731-124482731 on Build GRCh38
rs1042191255 Chr.9: 124482672-124482672 on Build GRCh38
rs889006988 Chr.9: 124482683-124482683 on Build GRCh38
rs35584614 Chr.9: 124482417-124482418 on Build GRCh38
rs1038690991 Chr.9: 124482370-124482370 on Build GRCh38
rs76802269 Chr.9: 124482394-124482394 on Build GRCh38
rs893109403 Chr.9: 124482563-124482563 on Build GRCh38
rs574309764 Chr.9: 124482656-124482656 on Build GRCh38
rs201030479 Chr.9: 124482586-124482586 on Build GRCh38
rs937603175 Chr.9: 124482558-124482558 on Build GRCh38
rs763130061 Chr.9: 124482737-124482737 on Build GRCh38
rs981611957 Chr.9: 124482528-124482528 on Build GRCh38
rs933018728 Chr.9: 124482395-124482395 on Build GRCh38
rs1033615712 Chr.9: 124482480-124482480 on Build GRCh38

View Full Product Details