Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 16760835-16761339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYAP1
Entrez Gene ID
94056
Gene Name
synapse associated protein 1
Gene Aliases
PRO3113
UniGene
Hs.744256
Gene Chromosome Location
Chr.X: 16719584-16762684 on Build GRCh38
Transcripts
NM_032796.3 NR_033181.1

SNP Detail

SNP ID Chromosome Location
rs1020790314 Chr.X: 16761087-16761087 on Build GRCh38
rs753064146 Chr.X: 16760991-16760991 on Build GRCh38
rs193069657 Chr.X: 16761238-16761238 on Build GRCh38
rs1054216070 Chr.X: 16761278-16761278 on Build GRCh38
rs551285976 Chr.X: 16761009-16761009 on Build GRCh38
rs933354918 Chr.X: 16760927-16760927 on Build GRCh38
rs1026159652 Chr.X: 16761244-16761244 on Build GRCh38
rs868825548 Chr.X: 16761268-16761268 on Build GRCh38
rs923227042 Chr.X: 16760915-16760915 on Build GRCh38
rs866625981 Chr.X: 16761210-16761210 on Build GRCh38
rs1044519418 Chr.X: 16760985-16760985 on Build GRCh38
rs182554926 Chr.X: 16761043-16761043 on Build GRCh38
rs563038610 Chr.X: 16761033-16761033 on Build GRCh38
rs934604026 Chr.X: 16761250-16761250 on Build GRCh38
rs891769427 Chr.X: 16761023-16761023 on Build GRCh38
rs187978227 Chr.X: 16761154-16761154 on Build GRCh38
rs924520703 Chr.X: 16761226-16761226 on Build GRCh38
rs760028997 Chr.X: 16760872-16760872 on Build GRCh38
rs1009140832 Chr.X: 16761171-16761171 on Build GRCh38
rs914444230 Chr.X: 16761280-16761280 on Build GRCh38
rs751745280 Chr.X: 16761103-16761103 on Build GRCh38
rs191579859 Chr.X: 16761008-16761008 on Build GRCh38
rs948438750 Chr.X: 16761287-16761288 on Build GRCh38
rs71662093 Chr.X: 16761211-16761212 on Build GRCh38
rs184563735 Chr.X: 16761279-16761279 on Build GRCh38
rs986653987 Chr.X: 16761123-16761123 on Build GRCh38
rs891210434 Chr.X: 16761248-16761248 on Build GRCh38
rs765542027 Chr.X: 16760948-16760948 on Build GRCh38
rs1017384348 Chr.X: 16761176-16761176 on Build GRCh38
rs34332618 Chr.X: 16761210-16761211 on Build GRCh38

View Full Product Details