Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 18892999-18893508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHKA2
Entrez Gene ID
5256
Gene Name
phosphorylase kinase regulatory subunit alpha 2
Gene Aliases
GSD9A, PHK, PYK, PYKL, XLG, XLG2
UniGene
Hs.54941
Gene Chromosome Location
Chr.X: 18892298-18984362 on Build GRCh38
Transcripts
NM_000292.2 XM_005274548.4 XM_005274550.4 XM_006724496.3 XM_006724498.3 XM_011545537.2 XM_011545538.2 XM_017029580.1 NR_029379.1

Gene Symbol
PHKA2-AS1
Entrez Gene ID
100132163
Gene Name
PHKA2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.607628
Gene Chromosome Location
Chr.X: 18890296-18894975 on Build GRCh38
Transcripts
NM_000292.2 XM_005274548.4 XM_005274550.4 XM_006724496.3 XM_006724498.3 XM_011545537.2 XM_011545538.2 XM_017029580.1 NR_029379.1

SNP Detail

SNP ID Chromosome Location
rs988693219 Chr.X: 18893454-18893454 on Build GRCh38
rs375034922 Chr.X: 18893444-18893444 on Build GRCh38
rs1014082862 Chr.X: 18893345-18893345 on Build GRCh38
rs988745890 Chr.X: 18893132-18893132 on Build GRCh38
rs1000297236 Chr.X: 18893420-18893420 on Build GRCh38
rs374849324 Chr.X: 18893483-18893483 on Build GRCh38
rs984384672 Chr.X: 18893122-18893123 on Build GRCh38
rs1055668477 Chr.X: 18893336-18893336 on Build GRCh38
rs147641162 Chr.X: 18893393-18893393 on Build GRCh38
rs934535420 Chr.X: 18893064-18893064 on Build GRCh38
rs371654294 Chr.X: 18893472-18893472 on Build GRCh38
rs368335914 Chr.X: 18893467-18893467 on Build GRCh38
rs1014413130 Chr.X: 18893367-18893367 on Build GRCh38
rs773062340 Chr.X: 18893479-18893479 on Build GRCh38
rs925877649 Chr.X: 18893039-18893039 on Build GRCh38
rs750634059 Chr.X: 18893302-18893303 on Build GRCh38
rs768718632 Chr.X: 18893363-18893363 on Build GRCh38
rs771329811 Chr.X: 18893342-18893342 on Build GRCh38
rs769235463 Chr.X: 18893465-18893465 on Build GRCh38
rs750498601 Chr.X: 18893445-18893445 on Build GRCh38
rs748052446 Chr.X: 18893272-18893272 on Build GRCh38
rs911462857 Chr.X: 18893126-18893126 on Build GRCh38
rs895978008 Chr.X: 18893360-18893360 on Build GRCh38
rs181609896 Chr.X: 18893177-18893177 on Build GRCh38
rs376602692 Chr.X: 18893453-18893453 on Build GRCh38
rs769262697 Chr.X: 18893476-18893476 on Build GRCh38
rs1033402707 Chr.X: 18893440-18893440 on Build GRCh38
rs201847825 Chr.X: 18893481-18893481 on Build GRCh38
rs372416461 Chr.X: 18893212-18893212 on Build GRCh38
rs762504306 Chr.X: 18893450-18893451 on Build GRCh38
rs941019487 Chr.X: 18893265-18893265 on Build GRCh38
rs774734878 Chr.X: 18893354-18893355 on Build GRCh38
rs1023029634 Chr.X: 18893406-18893406 on Build GRCh38
rs952703951 Chr.X: 18893109-18893109 on Build GRCh38
rs970574694 Chr.X: 18893416-18893416 on Build GRCh38
rs779795628 Chr.X: 18893316-18893316 on Build GRCh38
rs922390647 Chr.X: 18893153-18893153 on Build GRCh38
rs202246833 Chr.X: 18893462-18893462 on Build GRCh38
rs1038474983 Chr.X: 18893279-18893279 on Build GRCh38
rs971218108 Chr.X: 18893026-18893026 on Build GRCh38
rs746781873 Chr.X: 18893352-18893352 on Build GRCh38
rs769448282 Chr.X: 18893436-18893437 on Build GRCh38
rs922408501 Chr.X: 18893175-18893176 on Build GRCh38
rs894380741 Chr.X: 18893343-18893343 on Build GRCh38
rs918591511 Chr.X: 18893089-18893089 on Build GRCh38
rs749062601 Chr.X: 18893329-18893330 on Build GRCh38
rs745509660 Chr.X: 18893168-18893168 on Build GRCh38
rs754481107 Chr.X: 18893096-18893096 on Build GRCh38
rs942868632 Chr.X: 18893138-18893138 on Build GRCh38
rs185981642 Chr.X: 18893292-18893292 on Build GRCh38
rs775265688 Chr.X: 18893450-18893453 on Build GRCh38
rs562270151 Chr.X: 18893458-18893458 on Build GRCh38

View Full Product Details