Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 19370879-19371382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP3K15
Entrez Gene ID
389840
Gene Name
mitogen-activated protein kinase kinase kinase 15
Gene Aliases
ASK3, bA723P2.3
UniGene
Hs.713701
Gene Chromosome Location
Chr.X: 19360058-19515459 on Build GRCh38
Transcripts
NM_001001671.3 XM_011545507.2 XM_011545508.2 XM_011545510.2 XM_011545511.2

SNP Detail

SNP ID Chromosome Location
rs754871081 Chr.X: 19371303-19371303 on Build GRCh38
rs61740714 Chr.X: 19370985-19370985 on Build GRCh38
rs764726431 Chr.X: 19371065-19371065 on Build GRCh38
rs776067462 Chr.X: 19371231-19371231 on Build GRCh38
rs762237886 Chr.X: 19371359-19371359 on Build GRCh38
rs767744127 Chr.X: 19371042-19371042 on Build GRCh38
rs750354797 Chr.X: 19370947-19370947 on Build GRCh38
rs760453265 Chr.X: 19370921-19370921 on Build GRCh38
rs919064713 Chr.X: 19371258-19371258 on Build GRCh38
rs887751274 Chr.X: 19370991-19370991 on Build GRCh38
rs761211361 Chr.X: 19371054-19371054 on Build GRCh38
rs770527486 Chr.X: 19371242-19371242 on Build GRCh38
rs775422791 Chr.X: 19370920-19370920 on Build GRCh38
rs144031211 Chr.X: 19371003-19371003 on Build GRCh38
rs757572684 Chr.X: 19371076-19371076 on Build GRCh38
rs1003404693 Chr.X: 19371081-19371081 on Build GRCh38
rs779965473 Chr.X: 19370994-19370994 on Build GRCh38
rs191405175 Chr.X: 19370952-19370952 on Build GRCh38
rs372545551 Chr.X: 19371066-19371067 on Build GRCh38
rs200779059 Chr.X: 19371078-19371079 on Build GRCh38
rs771883623 Chr.X: 19371184-19371184 on Build GRCh38
rs779897096 Chr.X: 19371067-19371069 on Build GRCh38
rs557957228 Chr.X: 19371141-19371141 on Build GRCh38
rs147323806 Chr.X: 19371344-19371344 on Build GRCh38
rs768124794 Chr.X: 19370966-19370966 on Build GRCh38
rs775471170 Chr.X: 19371275-19371275 on Build GRCh38
rs190335333 Chr.X: 19371117-19371117 on Build GRCh38
rs112872738 Chr.X: 19371054-19371055 on Build GRCh38
rs754172834 Chr.X: 19370979-19370979 on Build GRCh38
rs766532413 Chr.X: 19370951-19370951 on Build GRCh38
rs749602392 Chr.X: 19371163-19371163 on Build GRCh38
rs200671095 Chr.X: 19371011-19371011 on Build GRCh38
rs945615334 Chr.X: 19371251-19371251 on Build GRCh38
rs759683487 Chr.X: 19371023-19371023 on Build GRCh38
rs140041575 Chr.X: 19371345-19371345 on Build GRCh38
rs750112388 Chr.X: 19371085-19371085 on Build GRCh38
rs186865221 Chr.X: 19371289-19371289 on Build GRCh38
rs140104197 Chr.X: 19370995-19370995 on Build GRCh38
rs757876398 Chr.X: 19371092-19371092 on Build GRCh38
rs776984201 Chr.X: 19371089-19371093 on Build GRCh38
rs761332697 Chr.X: 19371358-19371358 on Build GRCh38
rs188757568 Chr.X: 19371360-19371360 on Build GRCh38
rs768678555 Chr.X: 19371342-19371342 on Build GRCh38
rs370305346 Chr.X: 19370953-19370953 on Build GRCh38
rs755921760 Chr.X: 19371067-19371067 on Build GRCh38
rs756514149 Chr.X: 19370978-19370978 on Build GRCh38
rs769276695 Chr.X: 19371343-19371343 on Build GRCh38
rs764600201 Chr.X: 19371350-19371350 on Build GRCh38
rs774495074 Chr.X: 19371014-19371014 on Build GRCh38
rs368230655 Chr.X: 19370940-19370940 on Build GRCh38
rs948646636 Chr.X: 19371322-19371322 on Build GRCh38
rs375784052 Chr.X: 19371312-19371312 on Build GRCh38
rs773277747 Chr.X: 19370930-19370930 on Build GRCh38
rs749537426 Chr.X: 19371170-19371170 on Build GRCh38
rs182259394 Chr.X: 19371138-19371138 on Build GRCh38
rs759836039 Chr.X: 19371297-19371309 on Build GRCh38
rs73193525 Chr.X: 19371078-19371078 on Build GRCh38
rs765475538 Chr.X: 19371340-19371341 on Build GRCh38
rs769776445 Chr.X: 19371151-19371151 on Build GRCh38
rs746547349 Chr.X: 19371103-19371103 on Build GRCh38
rs776117021 Chr.X: 19371053-19371053 on Build GRCh38
rs780997212 Chr.X: 19371324-19371324 on Build GRCh38
rs925272958 Chr.X: 19371296-19371296 on Build GRCh38
rs1002234962 Chr.X: 19370911-19370911 on Build GRCh38
rs367742905 Chr.X: 19370990-19370990 on Build GRCh38
rs373928304 Chr.X: 19370980-19370980 on Build GRCh38
rs773637857 Chr.X: 19371079-19371081 on Build GRCh38
rs754694626 Chr.X: 19371067-19371068 on Build GRCh38
rs1011802904 Chr.X: 19371131-19371131 on Build GRCh38
rs747701628 Chr.X: 19371308-19371308 on Build GRCh38
rs779590671 Chr.X: 19371097-19371097 on Build GRCh38
rs771265515 Chr.X: 19371082-19371086 on Build GRCh38
rs747536307 Chr.X: 19371081-19371081 on Build GRCh38
rs780351262 Chr.X: 19370998-19370998 on Build GRCh38
rs985744798 Chr.X: 19371154-19371154 on Build GRCh38
rs143117337 Chr.X: 19370981-19370981 on Build GRCh38
rs777983210 Chr.X: 19371166-19371166 on Build GRCh38
rs764898842 Chr.X: 19370977-19370977 on Build GRCh38
rs771118792 Chr.X: 19371012-19371012 on Build GRCh38
rs976241042 Chr.X: 19371250-19371250 on Build GRCh38
rs929147284 Chr.X: 19371290-19371290 on Build GRCh38
rs984703565 Chr.X: 19371304-19371304 on Build GRCh38
rs752049081 Chr.X: 19371006-19371006 on Build GRCh38
rs1031799089 Chr.X: 19371113-19371113 on Build GRCh38
rs754160059 Chr.X: 19371069-19371069 on Build GRCh38

View Full Product Details