Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 19537614-19538126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3KBP1
Entrez Gene ID
30011
Gene Name
SH3 domain containing kinase binding protein 1
Gene Aliases
CD2BP3, CIN85, GIG10, HSB-1, HSB1, MIG18
UniGene
Hs.726365
Gene Chromosome Location
Chr.X: 19533965-19887626 on Build GRCh38
Transcripts
NM_001024666.2 NM_001184960.1 NM_031892.2 XM_005274494.2 XM_011545498.2 XM_011545499.2 XM_011545500.2 XM_011545502.2 XM_017029459.1 XM_017029460.1 XM_017029461.1 XM_017029462.1 XM_017029463.1 XM_017029464.1 XM_017029465.1 XM_017029466.1 XM_017029467.1 XM_017029468.1 XM_017029469.1 XM_017029470.1

SNP Detail

SNP ID Chromosome Location
rs192252658 Chr.X: 19537903-19537903 on Build GRCh38
rs910447682 Chr.X: 19538080-19538080 on Build GRCh38
rs762739943 Chr.X: 19537821-19537822 on Build GRCh38
rs772126704 Chr.X: 19537699-19537699 on Build GRCh38
rs777105719 Chr.X: 19537744-19537744 on Build GRCh38
rs751537283 Chr.X: 19537884-19537884 on Build GRCh38
rs769338873 Chr.X: 19537729-19537729 on Build GRCh38
rs770229859 Chr.X: 19537730-19537730 on Build GRCh38
rs748353385 Chr.X: 19537725-19537725 on Build GRCh38
rs58739461 Chr.X: 19537852-19537854 on Build GRCh38
rs749818093 Chr.X: 19537869-19537869 on Build GRCh38
rs765872916 Chr.X: 19537807-19537807 on Build GRCh38
rs966919013 Chr.X: 19538006-19538006 on Build GRCh38
rs796115021 Chr.X: 19537706-19537706 on Build GRCh38
rs1045040286 Chr.X: 19537826-19537826 on Build GRCh38
rs968285180 Chr.X: 19537990-19537990 on Build GRCh38
rs747153720 Chr.X: 19537707-19537707 on Build GRCh38
rs762337293 Chr.X: 19537752-19537752 on Build GRCh38
rs904878725 Chr.X: 19537842-19537842 on Build GRCh38
rs773402668 Chr.X: 19537731-19537731 on Build GRCh38
rs142403021 Chr.X: 19537724-19537724 on Build GRCh38
rs772861991 Chr.X: 19537799-19537799 on Build GRCh38
rs956417872 Chr.X: 19537860-19537860 on Build GRCh38
rs780185986 Chr.X: 19537696-19537696 on Build GRCh38
rs71975583 Chr.X: 19537848-19537850 on Build GRCh38
rs185038608 Chr.X: 19538009-19538009 on Build GRCh38
rs1032635588 Chr.X: 19538039-19538039 on Build GRCh38
rs757386213 Chr.X: 19537666-19537666 on Build GRCh38
rs767090677 Chr.X: 19537904-19537904 on Build GRCh38
rs184872839 Chr.X: 19537691-19537691 on Build GRCh38
rs371401883 Chr.X: 19537670-19537670 on Build GRCh38
rs201924840 Chr.X: 19537853-19537855 on Build GRCh38
rs779259642 Chr.X: 19537690-19537690 on Build GRCh38
rs986091490 Chr.X: 19538010-19538010 on Build GRCh38
rs990477501 Chr.X: 19538081-19538081 on Build GRCh38
rs1013200370 Chr.X: 19537735-19537735 on Build GRCh38
rs1034779287 Chr.X: 19537857-19537857 on Build GRCh38
rs762514885 Chr.X: 19537801-19537801 on Build GRCh38
rs959079152 Chr.X: 19538061-19538061 on Build GRCh38
rs1020733202 Chr.X: 19537945-19537945 on Build GRCh38
rs887801302 Chr.X: 19537788-19537788 on Build GRCh38
rs755097961 Chr.X: 19537720-19537720 on Build GRCh38
rs1003194965 Chr.X: 19537849-19537849 on Build GRCh38
rs764785952 Chr.X: 19537797-19537797 on Build GRCh38

View Full Product Details