Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 20153394-20153943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS6KA3
Entrez Gene ID
6197
Gene Name
ribosomal protein S6 kinase A3
Gene Aliases
CLS, HU-3, ISPK-1, MAPKAPK1B, MRX19, RSK, RSK2, S6K-alpha3, p90-RSK2, pp90RSK2
UniGene
Hs.445387
Gene Chromosome Location
Chr.X: 20149911-20267514 on Build GRCh38
Transcripts
NM_004586.2 XM_005274573.2 XM_005274577.2 XM_006724507.3 XM_011545555.1 XM_011545556.1 XM_011545557.2 XM_011545558.2 XM_011545559.1 XM_011545560.1 XM_011545561.1 XM_011545562.1 XM_011545563.2 XM_017029713.1 XM_017029714.1 XM_017029715.1 XM_017029716.1 XM_017029717.1 XM_017029718.1 XM_017029719.1

SNP Detail

SNP ID Chromosome Location
rs111612110 Chr.X: 20153915-20153915 on Build GRCh38
rs774999202 Chr.X: 20153863-20153863 on Build GRCh38
rs1035517287 Chr.X: 20153727-20153727 on Build GRCh38
rs979970460 Chr.X: 20153700-20153700 on Build GRCh38
rs911225394 Chr.X: 20153811-20153811 on Build GRCh38
rs959873078 Chr.X: 20153734-20153734 on Build GRCh38
rs752717737 Chr.X: 20153912-20153912 on Build GRCh38
rs749078121 Chr.X: 20153556-20153556 on Build GRCh38
rs987340222 Chr.X: 20153710-20153710 on Build GRCh38
rs930118737 Chr.X: 20153892-20153892 on Build GRCh38
rs969985851 Chr.X: 20153603-20153603 on Build GRCh38
rs181963682 Chr.X: 20153810-20153810 on Build GRCh38
rs751458765 Chr.X: 20153725-20153725 on Build GRCh38
rs746478108 Chr.X: 20153900-20153900 on Build GRCh38
rs953378873 Chr.X: 20153617-20153617 on Build GRCh38
rs759384648 Chr.X: 20153769-20153769 on Build GRCh38
rs913277426 Chr.X: 20153785-20153785 on Build GRCh38
rs114655077 Chr.X: 20153916-20153916 on Build GRCh38
rs1041287820 Chr.X: 20153872-20153872 on Build GRCh38
rs947385172 Chr.X: 20153809-20153809 on Build GRCh38
rs757055754 Chr.X: 20153722-20153722 on Build GRCh38
rs771841973 Chr.X: 20153837-20153837 on Build GRCh38
rs745817380 Chr.X: 20153836-20153836 on Build GRCh38
rs1052157629 Chr.X: 20153876-20153876 on Build GRCh38
rs935163964 Chr.X: 20153756-20153756 on Build GRCh38
rs922782920 Chr.X: 20153884-20153884 on Build GRCh38

View Full Product Details