Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 21655800-21656306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL34
Entrez Gene ID
257240
Gene Name
kelch like family member 34
Gene Aliases
-
UniGene
Hs.448572
Gene Chromosome Location
Chr.X: 21655349-21658387 on Build GRCh38
Transcripts
NM_153270.2

SNP Detail

SNP ID Chromosome Location
rs757823821 Chr.X: 21656042-21656042 on Build GRCh38
rs1046366483 Chr.X: 21656079-21656079 on Build GRCh38
rs768286102 Chr.X: 21656187-21656187 on Build GRCh38
rs371922567 Chr.X: 21656014-21656014 on Build GRCh38
rs780891644 Chr.X: 21656191-21656191 on Build GRCh38
rs764969010 Chr.X: 21655850-21655850 on Build GRCh38
rs369817754 Chr.X: 21655987-21655987 on Build GRCh38
rs867244485 Chr.X: 21656201-21656201 on Build GRCh38
rs750222185 Chr.X: 21655988-21655988 on Build GRCh38
rs779629965 Chr.X: 21656043-21656043 on Build GRCh38
rs1008449809 Chr.X: 21656215-21656215 on Build GRCh38
rs778014244 Chr.X: 21655910-21655910 on Build GRCh38
rs760384364 Chr.X: 21655956-21655956 on Build GRCh38
rs755208784 Chr.X: 21655872-21655872 on Build GRCh38
rs781331086 Chr.X: 21655876-21655876 on Build GRCh38
rs141001805 Chr.X: 21655970-21655970 on Build GRCh38
rs146742477 Chr.X: 21656050-21656050 on Build GRCh38
rs771027431 Chr.X: 21656174-21656176 on Build GRCh38
rs769236865 Chr.X: 21656196-21656196 on Build GRCh38
rs918531434 Chr.X: 21656108-21656108 on Build GRCh38
rs759468695 Chr.X: 21656250-21656250 on Build GRCh38
rs758975840 Chr.X: 21656183-21656183 on Build GRCh38
rs761373282 Chr.X: 21656088-21656088 on Build GRCh38
rs772960516 Chr.X: 21656106-21656106 on Build GRCh38
rs771599605 Chr.X: 21655926-21655926 on Build GRCh38
rs1020221216 Chr.X: 21656271-21656271 on Build GRCh38
rs148422098 Chr.X: 21655871-21655871 on Build GRCh38
rs766205595 Chr.X: 21656001-21656001 on Build GRCh38
rs143430991 Chr.X: 21655991-21655991 on Build GRCh38
rs753712882 Chr.X: 21655841-21655841 on Build GRCh38
rs757336667 Chr.X: 21656151-21656151 on Build GRCh38
rs61736674 Chr.X: 21656093-21656093 on Build GRCh38
rs767421393 Chr.X: 21656255-21656255 on Build GRCh38
rs761704504 Chr.X: 21655977-21655977 on Build GRCh38
rs766043231 Chr.X: 21656118-21656118 on Build GRCh38
rs1006979758 Chr.X: 21656153-21656153 on Build GRCh38
rs767057176 Chr.X: 21656256-21656256 on Build GRCh38
rs891297386 Chr.X: 21656138-21656138 on Build GRCh38
rs754052481 Chr.X: 21656282-21656282 on Build GRCh38
rs981462686 Chr.X: 21656029-21656029 on Build GRCh38
rs776095007 Chr.X: 21656083-21656083 on Build GRCh38
rs753960095 Chr.X: 21656137-21656137 on Build GRCh38
rs751404704 Chr.X: 21656122-21656122 on Build GRCh38
rs770945037 Chr.X: 21656222-21656222 on Build GRCh38
rs762961753 Chr.X: 21656111-21656111 on Build GRCh38
rs763542363 Chr.X: 21655960-21655960 on Build GRCh38
rs773410775 Chr.X: 21655958-21655959 on Build GRCh38
rs189111785 Chr.X: 21656184-21656184 on Build GRCh38
rs780486374 Chr.X: 21656069-21656069 on Build GRCh38
rs768335028 Chr.X: 21656082-21656082 on Build GRCh38
rs917780067 Chr.X: 21655927-21655927 on Build GRCh38
rs4451438 Chr.X: 21655959-21655959 on Build GRCh38
rs756428221 Chr.X: 21656021-21656021 on Build GRCh38
rs750846579 Chr.X: 21656166-21656166 on Build GRCh38
rs748193988 Chr.X: 21656161-21656187 on Build GRCh38
rs376948597 Chr.X: 21656131-21656131 on Build GRCh38
rs1054417495 Chr.X: 21656070-21656070 on Build GRCh38
rs747804174 Chr.X: 21656195-21656195 on Build GRCh38
rs771373294 Chr.X: 21656132-21656132 on Build GRCh38
rs772804950 Chr.X: 21656206-21656206 on Build GRCh38
rs777962151 Chr.X: 21656036-21656036 on Build GRCh38
rs763599026 Chr.X: 21655827-21655827 on Build GRCh38
rs142552568 Chr.X: 21655902-21655902 on Build GRCh38
rs769721888 Chr.X: 21656094-21656094 on Build GRCh38
rs200087075 Chr.X: 21656186-21656186 on Build GRCh38
rs779060186 Chr.X: 21656165-21656165 on Build GRCh38
rs748259656 Chr.X: 21655901-21655901 on Build GRCh38
rs774832814 Chr.X: 21655933-21655933 on Build GRCh38
rs749765670 Chr.X: 21655914-21655914 on Build GRCh38
rs757205780 Chr.X: 21655842-21655842 on Build GRCh38
rs146464429 Chr.X: 21655939-21655939 on Build GRCh38
rs762763527 Chr.X: 21656214-21656214 on Build GRCh38
rs746599130 Chr.X: 21656081-21656081 on Build GRCh38
rs746274321 Chr.X: 21655934-21655934 on Build GRCh38
rs61733995 Chr.X: 21656238-21656238 on Build GRCh38
rs914654948 Chr.X: 21656126-21656126 on Build GRCh38

View Full Product Details