Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 27822632-27823139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGEB10
Entrez Gene ID
139422
Gene Name
MAGE family member B10
Gene Aliases
-
UniGene
Hs.447363
Gene Chromosome Location
Chr.X: 27807990-27823014 on Build GRCh38
Transcripts
NM_182506.3

SNP Detail

SNP ID Chromosome Location
rs376185711 Chr.X: 27822791-27822791 on Build GRCh38
rs903328677 Chr.X: 27822904-27822904 on Build GRCh38
rs937651310 Chr.X: 27822843-27822843 on Build GRCh38
rs955339535 Chr.X: 27822758-27822758 on Build GRCh38
rs896951545 Chr.X: 27822953-27822953 on Build GRCh38
rs922504467 Chr.X: 27822738-27822738 on Build GRCh38
rs754498551 Chr.X: 27822790-27822790 on Build GRCh38
rs755795376 Chr.X: 27822884-27822884 on Build GRCh38
rs964131342 Chr.X: 27822974-27822974 on Build GRCh38
rs1000322499 Chr.X: 27822932-27822932 on Build GRCh38
rs771300153 Chr.X: 27823022-27823022 on Build GRCh38
rs778523874 Chr.X: 27822816-27822816 on Build GRCh38
rs926397100 Chr.X: 27822842-27822842 on Build GRCh38
rs924079251 Chr.X: 27822756-27822756 on Build GRCh38
rs1011872999 Chr.X: 27822717-27822717 on Build GRCh38
rs866519289 Chr.X: 27822889-27822889 on Build GRCh38
rs780072930 Chr.X: 27822743-27822743 on Build GRCh38
rs922181344 Chr.X: 27822802-27822802 on Build GRCh38
rs747862519 Chr.X: 27822923-27822923 on Build GRCh38
rs1036467542 Chr.X: 27822947-27822947 on Build GRCh38
rs183813413 Chr.X: 27822700-27822700 on Build GRCh38
rs1029638910 Chr.X: 27823001-27823001 on Build GRCh38
rs968196341 Chr.X: 27822735-27822735 on Build GRCh38
rs1049082862 Chr.X: 27822851-27822851 on Build GRCh38
rs866491641 Chr.X: 27823022-27823022 on Build GRCh38
rs975480439 Chr.X: 27822729-27822729 on Build GRCh38
rs1042376403 Chr.X: 27822899-27822899 on Build GRCh38
rs896039355 Chr.X: 27822876-27822876 on Build GRCh38
rs750355024 Chr.X: 27822657-27822657 on Build GRCh38
rs966247432 Chr.X: 27822778-27822778 on Build GRCh38
rs369464647 Chr.X: 27823031-27823032 on Build GRCh38
rs747420260 Chr.X: 27823021-27823021 on Build GRCh38
rs1056273943 Chr.X: 27823007-27823007 on Build GRCh38
rs753221298 Chr.X: 27822788-27822788 on Build GRCh38
rs983122446 Chr.X: 27822765-27822765 on Build GRCh38
rs976295373 Chr.X: 27822800-27822800 on Build GRCh38
rs908461588 Chr.X: 27823077-27823077 on Build GRCh38
rs958999228 Chr.X: 27822659-27822661 on Build GRCh38
rs977890366 Chr.X: 27822737-27822737 on Build GRCh38
rs66539971 Chr.X: 27823065-27823065 on Build GRCh38
rs1038085294 Chr.X: 27822840-27822840 on Build GRCh38
rs955569999 Chr.X: 27823026-27823026 on Build GRCh38
rs45612836 Chr.X: 27822984-27822984 on Build GRCh38
rs1056161149 Chr.X: 27822847-27822847 on Build GRCh38
rs146765483 Chr.X: 27822973-27822973 on Build GRCh38

View Full Product Details