Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 30236905-30237401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGEB3
Entrez Gene ID
4114
Gene Name
MAGE family member B3
Gene Aliases
CT3.5
UniGene
Hs.113290
Gene Chromosome Location
Chr.X: 30230436-30237493 on Build GRCh38
Transcripts
NM_002365.4 XM_011545513.2 XM_011545514.2

SNP Detail

SNP ID Chromosome Location
rs7063368 Chr.X: 30237198-30237198 on Build GRCh38
rs1004152502 Chr.X: 30237308-30237308 on Build GRCh38
rs936387952 Chr.X: 30237179-30237179 on Build GRCh38
rs750900622 Chr.X: 30236942-30236942 on Build GRCh38
rs1017697161 Chr.X: 30237093-30237093 on Build GRCh38
rs775329509 Chr.X: 30237240-30237240 on Build GRCh38
rs899160262 Chr.X: 30237251-30237251 on Build GRCh38
rs762978435 Chr.X: 30237253-30237253 on Build GRCh38
rs935874120 Chr.X: 30237347-30237347 on Build GRCh38
rs1026743395 Chr.X: 30237266-30237266 on Build GRCh38
rs754593997 Chr.X: 30236969-30236969 on Build GRCh38
rs866306553 Chr.X: 30237306-30237306 on Build GRCh38
rs950405290 Chr.X: 30237224-30237227 on Build GRCh38
rs759114389 Chr.X: 30237145-30237145 on Build GRCh38
rs147318419 Chr.X: 30237331-30237331 on Build GRCh38
rs945631615 Chr.X: 30237210-30237210 on Build GRCh38
rs969934697 Chr.X: 30237255-30237255 on Build GRCh38
rs755676801 Chr.X: 30237017-30237017 on Build GRCh38
rs529811313 Chr.X: 30237321-30237321 on Build GRCh38
rs753044248 Chr.X: 30236992-30236992 on Build GRCh38
rs531764575 Chr.X: 30236941-30236941 on Build GRCh38
rs187833904 Chr.X: 30237117-30237117 on Build GRCh38
rs914004762 Chr.X: 30237373-30237373 on Build GRCh38
rs776655301 Chr.X: 30236928-30236928 on Build GRCh38
rs751110422 Chr.X: 30237217-30237217 on Build GRCh38
rs938084565 Chr.X: 30237235-30237235 on Build GRCh38
rs780911680 Chr.X: 30237286-30237286 on Build GRCh38
rs920918705 Chr.X: 30237175-30237175 on Build GRCh38
rs975156208 Chr.X: 30237359-30237359 on Build GRCh38
rs369618374 Chr.X: 30236959-30236959 on Build GRCh38
rs193281834 Chr.X: 30237227-30237227 on Build GRCh38
rs989552851 Chr.X: 30237183-30237183 on Build GRCh38
rs965102721 Chr.X: 30237102-30237102 on Build GRCh38
rs906733201 Chr.X: 30237219-30237219 on Build GRCh38
rs761939953 Chr.X: 30236930-30236930 on Build GRCh38
rs769757850 Chr.X: 30237155-30237155 on Build GRCh38
rs751914299 Chr.X: 30236960-30236960 on Build GRCh38
rs755825726 Chr.X: 30237366-30237366 on Build GRCh38
rs144072080 Chr.X: 30236970-30236970 on Build GRCh38
rs1007780972 Chr.X: 30237040-30237040 on Build GRCh38
rs577770423 Chr.X: 30237013-30237014 on Build GRCh38
rs914123199 Chr.X: 30237185-30237185 on Build GRCh38

View Full Product Details