Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 37729484-37730017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XK
Entrez Gene ID
7504
Gene Name
X-linked Kx blood group
Gene Aliases
KX, MCLDS, NA, NAC, X1k, XKR1
UniGene
Hs.78919
Gene Chromosome Location
Chr.X: 37685766-37732130 on Build GRCh38
Transcripts
NM_021083.2

SNP Detail

SNP ID Chromosome Location
rs951004777 Chr.X: 37729669-37729669 on Build GRCh38
rs964989299 Chr.X: 37729967-37729967 on Build GRCh38
rs782730171 Chr.X: 37729987-37729987 on Build GRCh38
rs782076396 Chr.X: 37729645-37729645 on Build GRCh38
rs142405495 Chr.X: 37729520-37729520 on Build GRCh38
rs1054995865 Chr.X: 37729869-37729869 on Build GRCh38
rs782293580 Chr.X: 37729556-37729556 on Build GRCh38
rs782756152 Chr.X: 37729928-37729928 on Build GRCh38
rs17146127 Chr.X: 37729930-37729930 on Build GRCh38
rs144198509 Chr.X: 37729845-37729845 on Build GRCh38
rs1016519544 Chr.X: 37729964-37729964 on Build GRCh38
rs180937360 Chr.X: 37729835-37729835 on Build GRCh38
rs781968093 Chr.X: 37729638-37729638 on Build GRCh38
rs782465046 Chr.X: 37729981-37729981 on Build GRCh38
rs1026699326 Chr.X: 37729900-37729900 on Build GRCh38
rs781795511 Chr.X: 37729796-37729796 on Build GRCh38
rs964873132 Chr.X: 37729739-37729739 on Build GRCh38
rs1006266734 Chr.X: 37729949-37729949 on Build GRCh38
rs147778350 Chr.X: 37729860-37729860 on Build GRCh38
rs368185731 Chr.X: 37729950-37729950 on Build GRCh38
rs150893835 Chr.X: 37729566-37729566 on Build GRCh38
rs140233842 Chr.X: 37729605-37729605 on Build GRCh38
rs56296545 Chr.X: 37729936-37729936 on Build GRCh38
rs184091517 Chr.X: 37729661-37729661 on Build GRCh38
rs181428605 Chr.X: 37729519-37729519 on Build GRCh38
rs996329146 Chr.X: 37729968-37729968 on Build GRCh38
rs937837375 Chr.X: 37729868-37729868 on Build GRCh38
rs945312135 Chr.X: 37729574-37729574 on Build GRCh38
rs985049180 Chr.X: 37729722-37729722 on Build GRCh38
rs1002203769 Chr.X: 37729614-37729614 on Build GRCh38
rs782139819 Chr.X: 37729872-37729872 on Build GRCh38
rs782720430 Chr.X: 37729646-37729646 on Build GRCh38
rs1043625610 Chr.X: 37729856-37729856 on Build GRCh38
rs539603265 Chr.X: 37729723-37729723 on Build GRCh38
rs188429260 Chr.X: 37729816-37729816 on Build GRCh38
rs1043509596 Chr.X: 37729599-37729599 on Build GRCh38
rs913579892 Chr.X: 37729844-37729844 on Build GRCh38
rs974943269 Chr.X: 37729755-37729755 on Build GRCh38
rs782348948 Chr.X: 37729700-37729700 on Build GRCh38
rs921001123 Chr.X: 37729784-37729784 on Build GRCh38
rs933645165 Chr.X: 37729538-37729538 on Build GRCh38
rs1013685139 Chr.X: 37729891-37729891 on Build GRCh38
rs185849854 Chr.X: 37729988-37729988 on Build GRCh38
rs1050813273 Chr.X: 37729562-37729562 on Build GRCh38
rs896601789 Chr.X: 37729643-37729643 on Build GRCh38

View Full Product Details