Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 38054074-38054567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYTL5
Entrez Gene ID
94122
Gene Name
synaptotagmin like 5
Gene Aliases
slp5
UniGene
Hs.662334
Gene Chromosome Location
Chr.X: 37888915-38128820 on Build GRCh38
Transcripts
NM_001163334.1 NM_001163335.1 NM_138780.2 XM_011544001.2 XM_011544002.2 XM_017029972.1 XM_017029973.1 XM_017029974.1

SNP Detail

SNP ID Chromosome Location
rs747441779 Chr.X: 38054435-38054435 on Build GRCh38
rs781133865 Chr.X: 38054379-38054379 on Build GRCh38
rs981972921 Chr.X: 38054363-38054363 on Build GRCh38
rs145581759 Chr.X: 38054421-38054421 on Build GRCh38
rs199690787 Chr.X: 38054245-38054245 on Build GRCh38
rs1054607828 Chr.X: 38054534-38054534 on Build GRCh38
rs764303335 Chr.X: 38054225-38054225 on Build GRCh38
rs1029900815 Chr.X: 38054367-38054367 on Build GRCh38
rs941335835 Chr.X: 38054445-38054445 on Build GRCh38
rs752190115 Chr.X: 38054406-38054406 on Build GRCh38
rs775419067 Chr.X: 38054331-38054332 on Build GRCh38
rs202135180 Chr.X: 38054464-38054465 on Build GRCh38
rs939444958 Chr.X: 38054511-38054511 on Build GRCh38
rs751482809 Chr.X: 38054287-38054287 on Build GRCh38
rs767510948 Chr.X: 38054405-38054405 on Build GRCh38
rs746286212 Chr.X: 38054197-38054197 on Build GRCh38
rs775879263 Chr.X: 38054199-38054199 on Build GRCh38
rs747494621 Chr.X: 38054330-38054330 on Build GRCh38
rs147486310 Chr.X: 38054313-38054313 on Build GRCh38
rs944941922 Chr.X: 38054453-38054453 on Build GRCh38
rs754450459 Chr.X: 38054181-38054181 on Build GRCh38
rs769484187 Chr.X: 38054188-38054188 on Build GRCh38
rs920716673 Chr.X: 38054396-38054396 on Build GRCh38
rs144672652 Chr.X: 38054252-38054252 on Build GRCh38
rs776842401 Chr.X: 38054356-38054356 on Build GRCh38
rs780574469 Chr.X: 38054323-38054323 on Build GRCh38
rs180793831 Chr.X: 38054458-38054458 on Build GRCh38
rs769270948 Chr.X: 38054451-38054451 on Build GRCh38
rs1023289689 Chr.X: 38054337-38054337 on Build GRCh38
rs751418650 Chr.X: 38054402-38054402 on Build GRCh38
rs376656787 Chr.X: 38054456-38054456 on Build GRCh38
rs866455377 Chr.X: 38054422-38054422 on Build GRCh38
rs755760925 Chr.X: 38054304-38054304 on Build GRCh38
rs1040613709 Chr.X: 38054514-38054514 on Build GRCh38
rs754433512 Chr.X: 38054293-38054293 on Build GRCh38
rs200888867 Chr.X: 38054388-38054388 on Build GRCh38
rs780783319 Chr.X: 38054184-38054184 on Build GRCh38
rs373682306 Chr.X: 38054404-38054404 on Build GRCh38
rs376473841 Chr.X: 38054340-38054340 on Build GRCh38
rs749705865 Chr.X: 38054543-38054543 on Build GRCh38
rs144103691 Chr.X: 38054386-38054386 on Build GRCh38
rs375472063 Chr.X: 38054164-38054164 on Build GRCh38
rs762039196 Chr.X: 38054370-38054370 on Build GRCh38
rs766272918 Chr.X: 38054286-38054286 on Build GRCh38
rs903426904 Chr.X: 38054526-38054526 on Build GRCh38
rs770079186 Chr.X: 38054378-38054378 on Build GRCh38
rs747818546 Chr.X: 38054185-38054185 on Build GRCh38
rs779804254 Chr.X: 38054176-38054177 on Build GRCh38
rs776948296 Chr.X: 38054231-38054231 on Build GRCh38
rs758138832 Chr.X: 38054254-38054254 on Build GRCh38
rs762093952 Chr.X: 38054244-38054244 on Build GRCh38
rs1057036764 Chr.X: 38054117-38054117 on Build GRCh38
rs774669772 Chr.X: 38054189-38054189 on Build GRCh38
rs772330806 Chr.X: 38054322-38054322 on Build GRCh38
rs778757702 Chr.X: 38054197-38054198 on Build GRCh38
rs777421527 Chr.X: 38054420-38054420 on Build GRCh38
rs752245362 Chr.X: 38054303-38054303 on Build GRCh38
rs780263191 Chr.X: 38054424-38054424 on Build GRCh38
rs868016391 Chr.X: 38054318-38054318 on Build GRCh38
rs750345460 Chr.X: 38054198-38054198 on Build GRCh38
rs770024710 Chr.X: 38054472-38054472 on Build GRCh38
rs748981437 Chr.X: 38054189-38054190 on Build GRCh38
rs758743506 Chr.X: 38054423-38054423 on Build GRCh38
rs754661192 Chr.X: 38054190-38054190 on Build GRCh38
rs999061801 Chr.X: 38054528-38054528 on Build GRCh38
rs867175501 Chr.X: 38054315-38054315 on Build GRCh38
rs746187137 Chr.X: 38054319-38054319 on Build GRCh38
rs368712030 Chr.X: 38054210-38054210 on Build GRCh38
rs760745364 Chr.X: 38054218-38054218 on Build GRCh38
rs754976703 Chr.X: 38054176-38054176 on Build GRCh38
rs755636022 Chr.X: 38054408-38054408 on Build GRCh38
rs910528933 Chr.X: 38054436-38054436 on Build GRCh38

View Full Product Details