Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 38284311-38284843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPGR
Entrez Gene ID
6103
Gene Name
retinitis pigmentosa GTPase regulator
Gene Aliases
COD1, CORDX1, CRD, PCDX, RP15, RP3, XLRP3, orf15
UniGene
Hs.61438
Gene Chromosome Location
Chr.X: 38269163-38327564 on Build GRCh38
Transcripts
NM_000328.2 NM_001034853.1 XM_005272633.2 XM_011543940.2 XM_017029710.1 XM_017029711.1 XM_017029712.1

SNP Detail

SNP ID Chromosome Location
rs762362189 Chr.X: 38284557-38284557 on Build GRCh38
rs1004005838 Chr.X: 38284776-38284776 on Build GRCh38
rs914048429 Chr.X: 38284402-38284402 on Build GRCh38
rs1016764036 Chr.X: 38284799-38284799 on Build GRCh38
rs986571279 Chr.X: 38284386-38284386 on Build GRCh38
rs968395088 Chr.X: 38284411-38284411 on Build GRCh38
rs1018017713 Chr.X: 38284484-38284484 on Build GRCh38
rs992431161 Chr.X: 38284473-38284473 on Build GRCh38
rs149408606 Chr.X: 38284819-38284819 on Build GRCh38
rs764526118 Chr.X: 38284447-38284447 on Build GRCh38
rs1009794868 Chr.X: 38284374-38284375 on Build GRCh38
rs191558770 Chr.X: 38284663-38284663 on Build GRCh38
rs925559073 Chr.X: 38284550-38284550 on Build GRCh38
rs55711031 Chr.X: 38284812-38284812 on Build GRCh38
rs1022582752 Chr.X: 38284376-38284376 on Build GRCh38
rs929838168 Chr.X: 38284701-38284701 on Build GRCh38
rs978876908 Chr.X: 38284416-38284416 on Build GRCh38
rs982643002 Chr.X: 38284771-38284771 on Build GRCh38
rs185996057 Chr.X: 38284554-38284554 on Build GRCh38
rs1013602165 Chr.X: 38284626-38284626 on Build GRCh38
rs896503928 Chr.X: 38284583-38284583 on Build GRCh38
rs886910164 Chr.X: 38284682-38284682 on Build GRCh38
rs922409334 Chr.X: 38284692-38284692 on Build GRCh38
rs945540772 Chr.X: 38284404-38284404 on Build GRCh38
rs1055123209 Chr.X: 38284556-38284556 on Build GRCh38
rs76689439 Chr.X: 38284375-38284375 on Build GRCh38
rs753612093 Chr.X: 38284640-38284640 on Build GRCh38
rs1043964924 Chr.X: 38284470-38284470 on Build GRCh38
rs964102877 Chr.X: 38284606-38284606 on Build GRCh38

View Full Product Details