Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 46604827-46605327 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC9A7
Entrez Gene ID
84679
Gene Name
solute carrier family 9 member A7
Gene Aliases
NHE-7, NHE7, SLC9A6
UniGene
Hs.91389
Gene Chromosome Location
Chr.X: 46599251-46759172 on Build GRCh38
Transcripts
NM_001257291.1 NM_032591.2 XM_005272681.4 XM_005272682.4 XM_006724564.3 XM_011543990.2 XM_017029905.1 XM_017029906.1

SNP Detail

SNP ID Chromosome Location
rs904014014 Chr.X: 46604888-46604888 on Build GRCh38
rs758355430 Chr.X: 46604956-46604956 on Build GRCh38
rs925317719 Chr.X: 46605235-46605235 on Build GRCh38
rs1005013260 Chr.X: 46605219-46605219 on Build GRCh38
rs753599247 Chr.X: 46605061-46605061 on Build GRCh38
rs921839706 Chr.X: 46604983-46604983 on Build GRCh38
rs951880949 Chr.X: 46605037-46605037 on Build GRCh38
rs935409818 Chr.X: 46605236-46605236 on Build GRCh38
rs868458501 Chr.X: 46605225-46605225 on Build GRCh38
rs757296940 Chr.X: 46605185-46605185 on Build GRCh38
rs1001198430 Chr.X: 46605290-46605290 on Build GRCh38
rs780301566 Chr.X: 46604986-46604986 on Build GRCh38
rs1027397706 Chr.X: 46605018-46605018 on Build GRCh38
rs1031171328 Chr.X: 46604930-46604930 on Build GRCh38
rs933180669 Chr.X: 46605051-46605051 on Build GRCh38
rs760338867 Chr.X: 46605015-46605015 on Build GRCh38
rs142265439 Chr.X: 46605263-46605263 on Build GRCh38
rs898789534 Chr.X: 46605248-46605249 on Build GRCh38
rs1008877680 Chr.X: 46605261-46605261 on Build GRCh38
rs57402003 Chr.X: 46604914-46604914 on Build GRCh38
rs752587690 Chr.X: 46605178-46605178 on Build GRCh38
rs747043130 Chr.X: 46605059-46605059 on Build GRCh38
rs907712090 Chr.X: 46605068-46605068 on Build GRCh38
rs190050173 Chr.X: 46605096-46605096 on Build GRCh38
rs916570533 Chr.X: 46605135-46605135 on Build GRCh38
rs766064658 Chr.X: 46605060-46605060 on Build GRCh38
rs1043829909 Chr.X: 46605216-46605216 on Build GRCh38
rs1019003644 Chr.X: 46604949-46604949 on Build GRCh38
rs1008493774 Chr.X: 46604946-46604946 on Build GRCh38
rs371345358 Chr.X: 46605260-46605260 on Build GRCh38
rs948069222 Chr.X: 46605140-46605140 on Build GRCh38
rs1053003725 Chr.X: 46605237-46605237 on Build GRCh38
rs960881919 Chr.X: 46605086-46605086 on Build GRCh38
rs925922550 Chr.X: 46604913-46604913 on Build GRCh38
rs983652608 Chr.X: 46605062-46605062 on Build GRCh38
rs994364889 Chr.X: 46605190-46605190 on Build GRCh38
rs891338387 Chr.X: 46604938-46604938 on Build GRCh38
rs754795922 Chr.X: 46605090-46605090 on Build GRCh38
rs202048216 Chr.X: 46605191-46605191 on Build GRCh38

View Full Product Details