Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47448100-47448595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF41
Entrez Gene ID
7592
Gene Name
zinc finger protein 41
Gene Aliases
MRX89
UniGene
Hs.496074
Gene Chromosome Location
Chr.X: 47444691-47483671 on Build GRCh38
Transcripts
NM_001324139.1 NM_001324140.1 NM_001324141.1 NM_001324142.1 NM_001324143.1 NM_001324144.1 NM_001324145.1 NM_001324147.1 NM_001324148.1 NM_001324149.1 NM_001324150.1 NM_001324151.1 NM_001324152.1 NM_001324153.1 NM_001324154.1 NM_001324155.1 NM_001324156.1 NM_001324157.1 NM_007130.3 NM_153380.3 XM_006724550.3 XM_006724555.3 XM_017029810.1 XM_017029811.1 XM_017029812.1 XM_017029813.1 XM_017029814.1 XM_017029815.1 XM_017029816.1 XM_017029817.1

SNP Detail

SNP ID Chromosome Location
rs775346171 Chr.X: 47448277-47448277 on Build GRCh38
rs868761000 Chr.X: 47448441-47448441 on Build GRCh38
rs746942789 Chr.X: 47448468-47448468 on Build GRCh38
rs774159620 Chr.X: 47448254-47448254 on Build GRCh38
rs199832810 Chr.X: 47448300-47448300 on Build GRCh38
rs769418347 Chr.X: 47448495-47448495 on Build GRCh38
rs750080807 Chr.X: 47448303-47448303 on Build GRCh38
rs745701586 Chr.X: 47448261-47448261 on Build GRCh38
rs776158914 Chr.X: 47448289-47448289 on Build GRCh38
rs867926388 Chr.X: 47448278-47448278 on Build GRCh38
rs763525540 Chr.X: 47448286-47448286 on Build GRCh38
rs144790101 Chr.X: 47448496-47448496 on Build GRCh38
rs767782187 Chr.X: 47448534-47448534 on Build GRCh38
rs753911808 Chr.X: 47448394-47448394 on Build GRCh38
rs144063485 Chr.X: 47448299-47448299 on Build GRCh38
rs993919615 Chr.X: 47448442-47448442 on Build GRCh38
rs757293995 Chr.X: 47448439-47448439 on Build GRCh38
rs866714414 Chr.X: 47448250-47448250 on Build GRCh38
rs764699249 Chr.X: 47448552-47448552 on Build GRCh38
rs779841634 Chr.X: 47448453-47448453 on Build GRCh38
rs759993530 Chr.X: 47448353-47448354 on Build GRCh38
rs761460937 Chr.X: 47448492-47448492 on Build GRCh38
rs777734489 Chr.X: 47448373-47448373 on Build GRCh38
rs762631416 Chr.X: 47448514-47448514 on Build GRCh38
rs140982109 Chr.X: 47448469-47448469 on Build GRCh38
rs770494529 Chr.X: 47448322-47448322 on Build GRCh38
rs1050476033 Chr.X: 47448476-47448476 on Build GRCh38
rs771921199 Chr.X: 47448452-47448452 on Build GRCh38
rs898068187 Chr.X: 47448239-47448239 on Build GRCh38
rs746076782 Chr.X: 47448447-47448447 on Build GRCh38
rs1025552540 Chr.X: 47448318-47448318 on Build GRCh38
rs868329425 Chr.X: 47448541-47448541 on Build GRCh38
rs958367790 Chr.X: 47448385-47448385 on Build GRCh38
rs772357367 Chr.X: 47448334-47448334 on Build GRCh38
rs756218850 Chr.X: 47448166-47448166 on Build GRCh38
rs771831156 Chr.X: 47448271-47448271 on Build GRCh38
rs760656339 Chr.X: 47448285-47448285 on Build GRCh38
rs942073915 Chr.X: 47448555-47448555 on Build GRCh38
rs398124548 Chr.X: 47448225-47448225 on Build GRCh38
rs148848609 Chr.X: 47448192-47448192 on Build GRCh38
rs777217099 Chr.X: 47448203-47448203 on Build GRCh38
rs868597650 Chr.X: 47448262-47448262 on Build GRCh38
rs762030062 Chr.X: 47448324-47448324 on Build GRCh38
rs200254551 Chr.X: 47448177-47448177 on Build GRCh38
rs748678938 Chr.X: 47448163-47448163 on Build GRCh38
rs753145512 Chr.X: 47448362-47448362 on Build GRCh38
rs61739400 Chr.X: 47448372-47448372 on Build GRCh38
rs186696242 Chr.X: 47448446-47448446 on Build GRCh38
rs776404091 Chr.X: 47448473-47448473 on Build GRCh38
rs766957386 Chr.X: 47448551-47448551 on Build GRCh38

View Full Product Details