Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48003567-48004063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF182
Entrez Gene ID
7569
Gene Name
zinc finger protein 182
Gene Aliases
HHZ150, KOX14, ZNF21, Zfp182
UniGene
Hs.189690
Gene Chromosome Location
Chr.X: 47974851-48003995 on Build GRCh38
Transcripts
NM_001007088.1 NM_001178099.1 NM_006962.1

SNP Detail

SNP ID Chromosome Location
rs782639546 Chr.X: 48003770-48003770 on Build GRCh38
rs782229625 Chr.X: 48003881-48003881 on Build GRCh38
rs930300519 Chr.X: 48003677-48003677 on Build GRCh38
rs895277854 Chr.X: 48003780-48003780 on Build GRCh38
rs181972272 Chr.X: 48003589-48003589 on Build GRCh38
rs782584596 Chr.X: 48004036-48004036 on Build GRCh38
rs35278028 Chr.X: 48003687-48003687 on Build GRCh38
rs34490808 Chr.X: 48004011-48004011 on Build GRCh38
rs782708235 Chr.X: 48004038-48004038 on Build GRCh38
rs1012923080 Chr.X: 48003807-48003807 on Build GRCh38
rs782255180 Chr.X: 48003786-48003786 on Build GRCh38
rs907548066 Chr.X: 48004014-48004014 on Build GRCh38
rs983111970 Chr.X: 48003980-48003980 on Build GRCh38
rs974653349 Chr.X: 48003649-48003649 on Build GRCh38
rs1030996294 Chr.X: 48003894-48003894 on Build GRCh38
rs186393841 Chr.X: 48003963-48003963 on Build GRCh38
rs955425324 Chr.X: 48003609-48003609 on Build GRCh38
rs955476385 Chr.X: 48003910-48003910 on Build GRCh38
rs1043957655 Chr.X: 48003813-48003813 on Build GRCh38
rs781951222 Chr.X: 48003624-48003624 on Build GRCh38
rs939107965 Chr.X: 48004016-48004016 on Build GRCh38
rs939094723 Chr.X: 48003721-48003721 on Build GRCh38
rs920267633 Chr.X: 48003655-48003655 on Build GRCh38
rs1047364896 Chr.X: 48003687-48003687 on Build GRCh38
rs538208887 Chr.X: 48003673-48003673 on Build GRCh38
rs964988409 Chr.X: 48003645-48003645 on Build GRCh38
rs556594257 Chr.X: 48003792-48003792 on Build GRCh38
rs1008275863 Chr.X: 48003921-48003921 on Build GRCh38
rs999550652 Chr.X: 48003879-48003879 on Build GRCh38
rs782024600 Chr.X: 48003962-48003962 on Build GRCh38
rs991838636 Chr.X: 48004017-48004017 on Build GRCh38
rs570768081 Chr.X: 48003613-48003613 on Build GRCh38
rs1056598566 Chr.X: 48003747-48003747 on Build GRCh38
rs782515679 Chr.X: 48003635-48003635 on Build GRCh38
rs964547764 Chr.X: 48003952-48003952 on Build GRCh38
rs907657578 Chr.X: 48003712-48003712 on Build GRCh38
rs903871854 Chr.X: 48003864-48003864 on Build GRCh38
rs951949262 Chr.X: 48003974-48003974 on Build GRCh38

View Full Product Details