Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48258145-48258677 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SSX1
Entrez Gene ID
6756
Gene Name
SSX family member 1
Gene Aliases
CT5.1, SSRC
UniGene
Hs.434142
Gene Chromosome Location
Chr.X: 48255317-48267444 on Build GRCh38
Transcripts
NM_001278691.1 NM_005635.3

SNP Detail

SNP ID Chromosome Location
rs782109188 Chr.X: 48258647-48258647 on Build GRCh38
rs782345088 Chr.X: 48258531-48258531 on Build GRCh38
rs782454897 Chr.X: 48258488-48258488 on Build GRCh38
rs782094672 Chr.X: 48258553-48258553 on Build GRCh38
rs782793205 Chr.X: 48258634-48258634 on Build GRCh38
rs145959932 Chr.X: 48258562-48258562 on Build GRCh38
rs782692902 Chr.X: 48258556-48258556 on Build GRCh38
rs782811751 Chr.X: 48258491-48258491 on Build GRCh38
rs782073930 Chr.X: 48258620-48258620 on Build GRCh38
rs12557174 Chr.X: 48258328-48258328 on Build GRCh38
rs782768254 Chr.X: 48258582-48258582 on Build GRCh38
rs782439169 Chr.X: 48258507-48258507 on Build GRCh38
rs782412485 Chr.X: 48258519-48258519 on Build GRCh38
rs781818520 Chr.X: 48258545-48258545 on Build GRCh38
rs372463561 Chr.X: 48258640-48258640 on Build GRCh38
rs782596185 Chr.X: 48258564-48258564 on Build GRCh38
rs782479670 Chr.X: 48258539-48258539 on Build GRCh38
rs782332611 Chr.X: 48258535-48258535 on Build GRCh38
rs782059118 Chr.X: 48258534-48258534 on Build GRCh38
rs782479044 Chr.X: 48258636-48258637 on Build GRCh38
rs782286940 Chr.X: 48258589-48258589 on Build GRCh38
rs781809573 Chr.X: 48258557-48258557 on Build GRCh38
rs782594887 Chr.X: 48258578-48258578 on Build GRCh38
rs782574666 Chr.X: 48258508-48258508 on Build GRCh38
rs782225333 Chr.X: 48258506-48258506 on Build GRCh38
rs184701342 Chr.X: 48258336-48258336 on Build GRCh38
rs782797795 Chr.X: 48258394-48258394 on Build GRCh38
rs148641027 Chr.X: 48258563-48258563 on Build GRCh38
rs369220749 Chr.X: 48258527-48258527 on Build GRCh38
rs782594508 Chr.X: 48258502-48258502 on Build GRCh38
rs782422642 Chr.X: 48258612-48258612 on Build GRCh38
rs781913147 Chr.X: 48258622-48258622 on Build GRCh38
rs781848875 Chr.X: 48258493-48258493 on Build GRCh38
rs782766177 Chr.X: 48258544-48258544 on Build GRCh38
rs782271895 Chr.X: 48258509-48258509 on Build GRCh38
rs781817244 Chr.X: 48258590-48258590 on Build GRCh38
rs782647012 Chr.X: 48258522-48258522 on Build GRCh38
rs782167339 Chr.X: 48258632-48258632 on Build GRCh38
rs375162928 Chr.X: 48258495-48258495 on Build GRCh38
rs111617084 Chr.X: 48258191-48258191 on Build GRCh38
rs202098049 Chr.X: 48258568-48258568 on Build GRCh38
rs782311385 Chr.X: 48258577-48258577 on Build GRCh38
rs548890992 Chr.X: 48258277-48258277 on Build GRCh38
rs782165372 Chr.X: 48258537-48258537 on Build GRCh38
rs781997232 Chr.X: 48258614-48258614 on Build GRCh38
rs782060083 Chr.X: 48258185-48258185 on Build GRCh38

View Full Product Details