Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49165106-49165600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGIX
Entrez Gene ID
79917
Gene Name
MAGI family member, X-linked
Gene Aliases
JM10, PDZX
UniGene
Hs.193170
Gene Chromosome Location
Chr.X: 49162569-49168157 on Build GRCh38
Transcripts
NM_001099681.2 NM_001099682.2 NM_024859.3 XM_005278062.4 XM_005278063.4 XM_005278064.4 XM_005278065.3 XM_006724560.3 XM_011543985.2 XM_011543986.2 XM_011543987.2

SNP Detail

SNP ID Chromosome Location
rs782405681 Chr.X: 49165201-49165201 on Build GRCh38
rs376354074 Chr.X: 49165408-49165408 on Build GRCh38
rs782763931 Chr.X: 49165260-49165260 on Build GRCh38
rs183982494 Chr.X: 49165301-49165301 on Build GRCh38
rs782365332 Chr.X: 49165208-49165208 on Build GRCh38
rs782689357 Chr.X: 49165163-49165163 on Build GRCh38
rs5905720 Chr.X: 49165199-49165199 on Build GRCh38
rs782129744 Chr.X: 49165215-49165215 on Build GRCh38
rs782187079 Chr.X: 49165205-49165205 on Build GRCh38
rs782242586 Chr.X: 49165372-49165372 on Build GRCh38
rs782673082 Chr.X: 49165390-49165390 on Build GRCh38
rs370810266 Chr.X: 49165216-49165216 on Build GRCh38
rs781896904 Chr.X: 49165164-49165164 on Build GRCh38
rs192451933 Chr.X: 49165265-49165265 on Build GRCh38
rs782297286 Chr.X: 49165332-49165332 on Build GRCh38
rs782173193 Chr.X: 49165188-49165188 on Build GRCh38
rs782312974 Chr.X: 49165434-49165434 on Build GRCh38
rs781878851 Chr.X: 49165148-49165148 on Build GRCh38
rs781989177 Chr.X: 49165331-49165331 on Build GRCh38
rs782155937 Chr.X: 49165222-49165222 on Build GRCh38
rs373871202 Chr.X: 49165252-49165252 on Build GRCh38
rs781934019 Chr.X: 49165202-49165202 on Build GRCh38
rs782429615 Chr.X: 49165166-49165166 on Build GRCh38
rs932124332 Chr.X: 49165253-49165253 on Build GRCh38
rs45505792 Chr.X: 49165310-49165310 on Build GRCh38
rs781946206 Chr.X: 49165355-49165355 on Build GRCh38
rs896134220 Chr.X: 49165305-49165305 on Build GRCh38
rs147071787 Chr.X: 49165385-49165385 on Build GRCh38
rs182456662 Chr.X: 49165150-49165150 on Build GRCh38
rs782724568 Chr.X: 49165383-49165383 on Build GRCh38
rs782038103 Chr.X: 49165217-49165217 on Build GRCh38
rs782406333 Chr.X: 49165354-49165354 on Build GRCh38
rs367797587 Chr.X: 49165288-49165288 on Build GRCh38
rs782388460 Chr.X: 49165189-49165189 on Build GRCh38
rs781837628 Chr.X: 49165250-49165250 on Build GRCh38
rs782468533 Chr.X: 49165298-49165298 on Build GRCh38
rs781937531 Chr.X: 49165523-49165523 on Build GRCh38
rs782720804 Chr.X: 49165245-49165245 on Build GRCh38
rs782394182 Chr.X: 49165328-49165328 on Build GRCh38
rs1009471471 Chr.X: 49165455-49165455 on Build GRCh38
rs976069886 Chr.X: 49165573-49165573 on Build GRCh38
rs782082892 Chr.X: 49165539-49165539 on Build GRCh38
rs781848626 Chr.X: 49165261-49165261 on Build GRCh38
rs187168174 Chr.X: 49165544-49165544 on Build GRCh38
rs782621434 Chr.X: 49165184-49165184 on Build GRCh38
rs949090213 Chr.X: 49165320-49165320 on Build GRCh38
rs782624034 Chr.X: 49165194-49165194 on Build GRCh38
rs781955378 Chr.X: 49165210-49165210 on Build GRCh38

View Full Product Details