Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49243031-49243527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC22
Entrez Gene ID
28952
Gene Name
coiled-coil domain containing 22
Gene Aliases
CXorf37, JM1, RTSC2
UniGene
Hs.26333
Gene Chromosome Location
Chr.X: 49235467-49250526 on Build GRCh38
Transcripts
NM_014008.4 XM_005272599.3

SNP Detail

SNP ID Chromosome Location
rs781890102 Chr.X: 49243293-49243293 on Build GRCh38
rs376665486 Chr.X: 49243240-49243240 on Build GRCh38
rs951690249 Chr.X: 49243226-49243226 on Build GRCh38
rs782617947 Chr.X: 49243413-49243413 on Build GRCh38
rs782768379 Chr.X: 49243194-49243194 on Build GRCh38
rs781846259 Chr.X: 49243303-49243303 on Build GRCh38
rs782616322 Chr.X: 49243233-49243233 on Build GRCh38
rs1027661399 Chr.X: 49243225-49243225 on Build GRCh38
rs782519523 Chr.X: 49243231-49243231 on Build GRCh38
rs959695845 Chr.X: 49243454-49243454 on Build GRCh38
rs782486077 Chr.X: 49243211-49243211 on Build GRCh38
rs782294535 Chr.X: 49243483-49243483 on Build GRCh38
rs782468983 Chr.X: 49243310-49243310 on Build GRCh38
rs781852596 Chr.X: 49243208-49243208 on Build GRCh38
rs369264021 Chr.X: 49243123-49243123 on Build GRCh38
rs141259252 Chr.X: 49243289-49243289 on Build GRCh38
rs370496191 Chr.X: 49243444-49243444 on Build GRCh38
rs781872351 Chr.X: 49243374-49243374 on Build GRCh38
rs782001304 Chr.X: 49243223-49243223 on Build GRCh38
rs1035321189 Chr.X: 49243453-49243453 on Build GRCh38
rs138289992 Chr.X: 49243448-49243448 on Build GRCh38
rs200591905 Chr.X: 49243161-49243161 on Build GRCh38
rs1012646131 Chr.X: 49243496-49243496 on Build GRCh38
rs782109659 Chr.X: 49243272-49243272 on Build GRCh38
rs376936166 Chr.X: 49243261-49243261 on Build GRCh38
rs782588505 Chr.X: 49243217-49243217 on Build GRCh38
rs782265569 Chr.X: 49243229-49243229 on Build GRCh38
rs899976632 Chr.X: 49243412-49243412 on Build GRCh38
rs781995736 Chr.X: 49243397-49243397 on Build GRCh38
rs150796681 Chr.X: 49243363-49243363 on Build GRCh38
rs782369027 Chr.X: 49243222-49243223 on Build GRCh38
rs782591281 Chr.X: 49243471-49243471 on Build GRCh38
rs782522722 Chr.X: 49243301-49243301 on Build GRCh38
rs782108051 Chr.X: 49243406-49243406 on Build GRCh38
rs782445678 Chr.X: 49243419-49243419 on Build GRCh38
rs201088755 Chr.X: 49243407-49243407 on Build GRCh38
rs912598985 Chr.X: 49243401-49243401 on Build GRCh38
rs782397485 Chr.X: 49243253-49243253 on Build GRCh38
rs782546800 Chr.X: 49243330-49243330 on Build GRCh38
rs782037688 Chr.X: 49243369-49243369 on Build GRCh38
rs782376528 Chr.X: 49243353-49243353 on Build GRCh38
rs374091309 Chr.X: 49243258-49243258 on Build GRCh38
rs186703174 Chr.X: 49243241-49243241 on Build GRCh38
rs782762060 Chr.X: 49243302-49243302 on Build GRCh38
rs781791796 Chr.X: 49243250-49243250 on Build GRCh38
rs1008306443 Chr.X: 49243051-49243051 on Build GRCh38
rs782145074 Chr.X: 49243166-49243166 on Build GRCh38
rs782660215 Chr.X: 49243334-49243334 on Build GRCh38
rs139220754 Chr.X: 49243442-49243442 on Build GRCh38
rs782598913 Chr.X: 49243321-49243321 on Build GRCh38
rs782778894 Chr.X: 49243388-49243388 on Build GRCh38
rs782797020 Chr.X: 49243421-49243421 on Build GRCh38
rs782815473 Chr.X: 49243290-49243290 on Build GRCh38
rs782150980 Chr.X: 49243380-49243380 on Build GRCh38
rs782735673 Chr.X: 49243056-49243056 on Build GRCh38
rs991242969 Chr.X: 49243251-49243251 on Build GRCh38
rs782262499 Chr.X: 49243348-49243348 on Build GRCh38
rs148652964 Chr.X: 49243160-49243160 on Build GRCh38
rs781948477 Chr.X: 49243280-49243280 on Build GRCh38
rs782693747 Chr.X: 49243159-49243159 on Build GRCh38
rs781821935 Chr.X: 49243440-49243440 on Build GRCh38
rs782461322 Chr.X: 49243449-49243449 on Build GRCh38
rs781915959 Chr.X: 49243247-49243247 on Build GRCh38
rs782176188 Chr.X: 49243324-49243324 on Build GRCh38
rs191605685 Chr.X: 49243277-49243277 on Build GRCh38

View Full Product Details