Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49250059-49250556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC22
Entrez Gene ID
28952
Gene Name
coiled-coil domain containing 22
Gene Aliases
CXorf37, JM1, RTSC2
UniGene
Hs.26333
Gene Chromosome Location
Chr.X: 49235467-49250526 on Build GRCh38
Transcripts
NM_014008.4 XM_005272599.3 NM_001114377.1 NM_014009.3 XM_006724533.2 XM_011543916.2 XM_017029565.1 XM_017029566.1 XM_017029567.1

Gene Symbol
FOXP3
Entrez Gene ID
50943
Gene Name
forkhead box P3
Gene Aliases
AIID, DIETER, IPEX, JM2, PIDX, XPID
UniGene
Hs.247700
Gene Chromosome Location
Chr.X: 49250436-49266505 on Build GRCh38
Transcripts
NM_014008.4 XM_005272599.3 NM_001114377.1 NM_014009.3 XM_006724533.2 XM_011543916.2 XM_017029565.1 XM_017029566.1 XM_017029567.1

SNP Detail

SNP ID Chromosome Location
rs999703101 Chr.X: 49250228-49250228 on Build GRCh38
rs782082339 Chr.X: 49250085-49250085 on Build GRCh38
rs782173426 Chr.X: 49250419-49250419 on Build GRCh38
rs782228512 Chr.X: 49250271-49250271 on Build GRCh38
rs973823127 Chr.X: 49250329-49250329 on Build GRCh38
rs782009113 Chr.X: 49250514-49250514 on Build GRCh38
rs181219991 Chr.X: 49250467-49250467 on Build GRCh38
rs904046877 Chr.X: 49250217-49250217 on Build GRCh38
rs797044661 Chr.X: 49250268-49250268 on Build GRCh38
rs781811940 Chr.X: 49250374-49250374 on Build GRCh38
rs782529954 Chr.X: 49250210-49250210 on Build GRCh38
rs955673389 Chr.X: 49250255-49250255 on Build GRCh38
rs782431385 Chr.X: 49250375-49250375 on Build GRCh38
rs782147414 Chr.X: 49250280-49250280 on Build GRCh38
rs982667170 Chr.X: 49250510-49250510 on Build GRCh38
rs782490982 Chr.X: 49250406-49250406 on Build GRCh38
rs398124266 Chr.X: 49250151-49250151 on Build GRCh38
rs199701533 Chr.X: 49250286-49250286 on Build GRCh38
rs782409676 Chr.X: 49250451-49250451 on Build GRCh38
rs782609786 Chr.X: 49250352-49250352 on Build GRCh38
rs139328507 Chr.X: 49250196-49250196 on Build GRCh38
rs951208241 Chr.X: 49250385-49250385 on Build GRCh38
rs781860071 Chr.X: 49250319-49250319 on Build GRCh38
rs782171344 Chr.X: 49250447-49250447 on Build GRCh38
rs781905938 Chr.X: 49250245-49250245 on Build GRCh38
rs1031657486 Chr.X: 49250244-49250244 on Build GRCh38
rs377286894 Chr.X: 49250197-49250197 on Build GRCh38
rs1012291874 Chr.X: 49250209-49250209 on Build GRCh38
rs782495769 Chr.X: 49250334-49250334 on Build GRCh38
rs150850672 Chr.X: 49250187-49250187 on Build GRCh38

View Full Product Details