Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49257583-49258079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXP3
Entrez Gene ID
50943
Gene Name
forkhead box P3
Gene Aliases
AIID, DIETER, IPEX, JM2, PIDX, XPID
UniGene
Hs.247700
Gene Chromosome Location
Chr.X: 49250436-49266505 on Build GRCh38
Transcripts
NM_001114377.1 NM_014009.3 XM_006724533.2 XM_011543916.2 XM_017029565.1 XM_017029566.1 XM_017029567.1

SNP Detail

SNP ID Chromosome Location
rs376715533 Chr.X: 49257726-49257726 on Build GRCh38
rs138970962 Chr.X: 49257744-49257744 on Build GRCh38
rs782172663 Chr.X: 49257643-49257643 on Build GRCh38
rs368483472 Chr.X: 49257733-49257733 on Build GRCh38
rs782457232 Chr.X: 49257631-49257631 on Build GRCh38
rs1054922263 Chr.X: 49257867-49257867 on Build GRCh38
rs782273002 Chr.X: 49257718-49257718 on Build GRCh38
rs782171326 Chr.X: 49257614-49257614 on Build GRCh38
rs781982592 Chr.X: 49257788-49257788 on Build GRCh38
rs782142042 Chr.X: 49257754-49257754 on Build GRCh38
rs1046113897 Chr.X: 49257808-49257808 on Build GRCh38
rs782587759 Chr.X: 49257635-49257635 on Build GRCh38
rs906321986 Chr.X: 49257833-49257833 on Build GRCh38
rs782628979 Chr.X: 49257725-49257725 on Build GRCh38
rs954839294 Chr.X: 49257616-49257616 on Build GRCh38
rs781841655 Chr.X: 49257630-49257630 on Build GRCh38
rs11465468 Chr.X: 49257999-49257999 on Build GRCh38
rs782558220 Chr.X: 49257656-49257656 on Build GRCh38
rs35705769 Chr.X: 49257878-49257879 on Build GRCh38
rs782262514 Chr.X: 49257886-49257886 on Build GRCh38
rs11091251 Chr.X: 49257842-49257842 on Build GRCh38
rs942280481 Chr.X: 49257691-49257691 on Build GRCh38
rs910806219 Chr.X: 49257682-49257682 on Build GRCh38
rs782412792 Chr.X: 49257658-49257658 on Build GRCh38
rs781921052 Chr.X: 49257753-49257753 on Build GRCh38
rs893685324 Chr.X: 49257882-49257882 on Build GRCh38
rs782789618 Chr.X: 49257782-49257782 on Build GRCh38
rs782763524 Chr.X: 49257615-49257615 on Build GRCh38
rs782411665 Chr.X: 49258052-49258052 on Build GRCh38
rs782373558 Chr.X: 49257936-49257936 on Build GRCh38
rs782210800 Chr.X: 49257730-49257730 on Build GRCh38
rs903198768 Chr.X: 49257942-49257942 on Build GRCh38

View Full Product Details