Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 102713691-102714193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMCX5-GPRASP2
Entrez Gene ID
100528062
Gene Name
ARMCX5-GPRASP2 readthrough
Gene Aliases
-
UniGene
Hs.522729
Gene Chromosome Location
Chr.X: 102599348-102717733 on Build GRCh38
Transcripts
NM_001199818.1 NM_001004051.3 NM_001184874.2 NM_001184875.2 NM_001184876.2 NM_138437.5

Gene Symbol
GPRASP2
Entrez Gene ID
114928
Gene Name
G protein-coupled receptor associated sorting protein 2
Gene Aliases
GASP2
UniGene
Hs.522729
Gene Chromosome Location
Chr.X: 102712176-102717733 on Build GRCh38
Transcripts
NM_001199818.1 NM_001004051.3 NM_001184874.2 NM_001184875.2 NM_001184876.2 NM_138437.5

SNP Detail

SNP ID Chromosome Location
rs41310725 Chr.X: 102713841-102713841 on Build GRCh38
rs1016619613 Chr.X: 102714106-102714106 on Build GRCh38
rs906432350 Chr.X: 102714013-102714013 on Build GRCh38
rs184090689 Chr.X: 102713768-102713768 on Build GRCh38
rs922810067 Chr.X: 102713774-102713774 on Build GRCh38
rs756114817 Chr.X: 102714003-102714003 on Build GRCh38
rs779010094 Chr.X: 102713818-102713819 on Build GRCh38
rs1003816427 Chr.X: 102714041-102714041 on Build GRCh38
rs757067005 Chr.X: 102713802-102713802 on Build GRCh38
rs781051501 Chr.X: 102713837-102713837 on Build GRCh38
rs576701889 Chr.X: 102714072-102714072 on Build GRCh38
rs139006702 Chr.X: 102713959-102713959 on Build GRCh38
rs983834700 Chr.X: 102714092-102714092 on Build GRCh38
rs750366376 Chr.X: 102713900-102713900 on Build GRCh38
rs973363509 Chr.X: 102713747-102713750 on Build GRCh38
rs973030557 Chr.X: 102714118-102714118 on Build GRCh38
rs188641020 Chr.X: 102714151-102714151 on Build GRCh38
rs763855253 Chr.X: 102713742-102713742 on Build GRCh38
rs779987375 Chr.X: 102714073-102714073 on Build GRCh38
rs373096911 Chr.X: 102714120-102714120 on Build GRCh38
rs889900489 Chr.X: 102713810-102713810 on Build GRCh38
rs369183982 Chr.X: 102713777-102713777 on Build GRCh38
rs762500217 Chr.X: 102713722-102713722 on Build GRCh38
rs376702099 Chr.X: 102714116-102714116 on Build GRCh38
rs1009499180 Chr.X: 102713961-102713961 on Build GRCh38

View Full Product Details