Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 102716498-102717001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMCX5-GPRASP2
Entrez Gene ID
100528062
Gene Name
ARMCX5-GPRASP2 readthrough
Gene Aliases
-
UniGene
Hs.522729
Gene Chromosome Location
Chr.X: 102599348-102717733 on Build GRCh38
Transcripts
NM_001199818.1 NM_001004051.3 NM_001184874.2 NM_001184875.2 NM_001184876.2 NM_138437.5

Gene Symbol
GPRASP2
Entrez Gene ID
114928
Gene Name
G protein-coupled receptor associated sorting protein 2
Gene Aliases
GASP2
UniGene
Hs.522729
Gene Chromosome Location
Chr.X: 102712176-102717733 on Build GRCh38
Transcripts
NM_001199818.1 NM_001004051.3 NM_001184874.2 NM_001184875.2 NM_001184876.2 NM_138437.5

SNP Detail

SNP ID Chromosome Location
rs769510587 Chr.X: 102716726-102716726 on Build GRCh38
rs763213194 Chr.X: 102716903-102716903 on Build GRCh38
rs17284956 Chr.X: 102716553-102716553 on Build GRCh38
rs769424276 Chr.X: 102716541-102716541 on Build GRCh38
rs758853963 Chr.X: 102716904-102716904 on Build GRCh38
rs755861123 Chr.X: 102716941-102716941 on Build GRCh38
rs6621549 Chr.X: 102716828-102716828 on Build GRCh38
rs867965336 Chr.X: 102716956-102716956 on Build GRCh38
rs772961604 Chr.X: 102716821-102716821 on Build GRCh38
rs868759566 Chr.X: 102716548-102716548 on Build GRCh38
rs886318081 Chr.X: 102716603-102716603 on Build GRCh38
rs918524118 Chr.X: 102716832-102716832 on Build GRCh38
rs372423590 Chr.X: 102716868-102716868 on Build GRCh38
rs769537243 Chr.X: 102716623-102716623 on Build GRCh38
rs771754041 Chr.X: 102716840-102716840 on Build GRCh38
rs776120842 Chr.X: 102716529-102716529 on Build GRCh38
rs745501234 Chr.X: 102716702-102716702 on Build GRCh38
rs755507410 Chr.X: 102716660-102716660 on Build GRCh38
rs138144817 Chr.X: 102716618-102716618 on Build GRCh38
rs745419497 Chr.X: 102716536-102716536 on Build GRCh38
rs753933427 Chr.X: 102716865-102716865 on Build GRCh38
rs774126365 Chr.X: 102716817-102716817 on Build GRCh38
rs368065140 Chr.X: 102716612-102716612 on Build GRCh38
rs764084491 Chr.X: 102716554-102716554 on Build GRCh38
rs972451861 Chr.X: 102716624-102716624 on Build GRCh38
rs761444224 Chr.X: 102716887-102716887 on Build GRCh38
rs912392409 Chr.X: 102716936-102716936 on Build GRCh38
rs773918934 Chr.X: 102716823-102716823 on Build GRCh38
rs199667642 Chr.X: 102716593-102716593 on Build GRCh38
rs766463571 Chr.X: 102716622-102716622 on Build GRCh38
rs375571887 Chr.X: 102716694-102716694 on Build GRCh38
rs748760002 Chr.X: 102716760-102716760 on Build GRCh38
rs372332241 Chr.X: 102716683-102716683 on Build GRCh38
rs747996065 Chr.X: 102716621-102716621 on Build GRCh38
rs768764070 Chr.X: 102716796-102716796 on Build GRCh38
rs931798164 Chr.X: 102716572-102716572 on Build GRCh38
rs779714407 Chr.X: 102716746-102716746 on Build GRCh38
rs770341893 Chr.X: 102716839-102716840 on Build GRCh38
rs753150242 Chr.X: 102716687-102716687 on Build GRCh38
rs766552046 Chr.X: 102716862-102716862 on Build GRCh38
rs912019428 Chr.X: 102716689-102716689 on Build GRCh38
rs1036348853 Chr.X: 102716753-102716753 on Build GRCh38
rs759586484 Chr.X: 102716870-102716870 on Build GRCh38
rs765811522 Chr.X: 102716881-102716881 on Build GRCh38
rs774219851 Chr.X: 102716563-102716563 on Build GRCh38
rs760895109 Chr.X: 102716859-102716859 on Build GRCh38
rs1049269148 Chr.X: 102716940-102716940 on Build GRCh38
rs751906238 Chr.X: 102716926-102716926 on Build GRCh38
rs778010561 Chr.X: 102716912-102716912 on Build GRCh38
rs773004253 Chr.X: 102716854-102716854 on Build GRCh38
rs779831055 Chr.X: 102716972-102716972 on Build GRCh38
rs201059510 Chr.X: 102716836-102716836 on Build GRCh38
rs369955160 Chr.X: 102716839-102716839 on Build GRCh38

View Full Product Details