Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 105895293-105895803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NRK
Entrez Gene ID
203447
Gene Name
Nik related kinase
Gene Aliases
NESK
UniGene
Hs.209527
Gene Chromosome Location
Chr.X: 105821944-105958610 on Build GRCh38
Transcripts
NM_198465.3 XM_006724632.3 XM_006724633.3 XM_006724634.3 XM_011530887.2

SNP Detail

SNP ID Chromosome Location
rs756932967 Chr.X: 105895347-105895347 on Build GRCh38
rs780996676 Chr.X: 105895387-105895387 on Build GRCh38
rs749769117 Chr.X: 105895420-105895420 on Build GRCh38
rs367709538 Chr.X: 105895758-105895758 on Build GRCh38
rs752083400 Chr.X: 105895391-105895393 on Build GRCh38
rs938696714 Chr.X: 105895408-105895408 on Build GRCh38
rs750759404 Chr.X: 105895395-105895395 on Build GRCh38
rs373870974 Chr.X: 105895516-105895516 on Build GRCh38
rs997872051 Chr.X: 105895695-105895695 on Build GRCh38
rs867486342 Chr.X: 105895397-105895397 on Build GRCh38
rs780855894 Chr.X: 105895604-105895604 on Build GRCh38
rs542672948 Chr.X: 105895363-105895364 on Build GRCh38
rs886879039 Chr.X: 105895707-105895707 on Build GRCh38
rs761422287 Chr.X: 105895571-105895571 on Build GRCh38
rs111844831 Chr.X: 105895364-105895367 on Build GRCh38
rs866567687 Chr.X: 105895393-105895393 on Build GRCh38
rs764433147 Chr.X: 105895387-105895389 on Build GRCh38
rs760958200 Chr.X: 105895458-105895458 on Build GRCh38
rs368040725 Chr.X: 105895533-105895533 on Build GRCh38
rs968497000 Chr.X: 105895644-105895644 on Build GRCh38
rs779823528 Chr.X: 105895364-105895375 on Build GRCh38
rs866382686 Chr.X: 105895630-105895630 on Build GRCh38
rs769090949 Chr.X: 105895426-105895426 on Build GRCh38
rs765397189 Chr.X: 105895364-105895371 on Build GRCh38
rs966184929 Chr.X: 105895600-105895600 on Build GRCh38
rs1042933235 Chr.X: 105895588-105895588 on Build GRCh38
rs769770801 Chr.X: 105895725-105895727 on Build GRCh38
rs869288013 Chr.X: 105895374-105895375 on Build GRCh38
rs763970804 Chr.X: 105895562-105895562 on Build GRCh38
rs922035968 Chr.X: 105895608-105895608 on Build GRCh38
rs1019147197 Chr.X: 105895577-105895577 on Build GRCh38
rs757744428 Chr.X: 105895392-105895393 on Build GRCh38
rs748067968 Chr.X: 105895699-105895699 on Build GRCh38
rs773413744 Chr.X: 105895451-105895451 on Build GRCh38
rs751698508 Chr.X: 105895367-105895368 on Build GRCh38
rs868566889 Chr.X: 105895466-105895466 on Build GRCh38
rs780632637 Chr.X: 105895402-105895402 on Build GRCh38
rs756532690 Chr.X: 105895400-105895400 on Build GRCh38
rs1039369282 Chr.X: 105895624-105895624 on Build GRCh38
rs562119457 Chr.X: 105895393-105895396 on Build GRCh38
rs763641422 Chr.X: 105895324-105895324 on Build GRCh38
rs748203697 Chr.X: 105895436-105895436 on Build GRCh38
rs927322983 Chr.X: 105895350-105895350 on Build GRCh38
rs768442390 Chr.X: 105895527-105895527 on Build GRCh38
rs904428759 Chr.X: 105895603-105895603 on Build GRCh38
rs992878178 Chr.X: 105895424-105895424 on Build GRCh38
rs778999118 Chr.X: 105895435-105895435 on Build GRCh38
rs374466713 Chr.X: 105895335-105895335 on Build GRCh38
rs145939345 Chr.X: 105895450-105895450 on Build GRCh38
rs751015202 Chr.X: 105895565-105895565 on Build GRCh38
rs979466080 Chr.X: 105895660-105895660 on Build GRCh38
rs923910667 Chr.X: 105895694-105895694 on Build GRCh38

View Full Product Details