Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 107152990-107153492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUP62CL
Entrez Gene ID
54830
Gene Name
nucleoporin 62 C-terminal like
Gene Aliases
-
UniGene
Hs.163629
Gene Chromosome Location
Chr.X: 107123427-107206440 on Build GRCh38
Transcripts
NM_017681.2 NR_033676.1

SNP Detail

SNP ID Chromosome Location
rs567327601 Chr.X: 107153123-107153123 on Build GRCh38
rs755754269 Chr.X: 107153228-107153228 on Build GRCh38
rs137855042 Chr.X: 107153206-107153206 on Build GRCh38
rs376685992 Chr.X: 107153265-107153265 on Build GRCh38
rs896747537 Chr.X: 107153361-107153361 on Build GRCh38
rs933862190 Chr.X: 107153204-107153204 on Build GRCh38
rs751403228 Chr.X: 107153224-107153224 on Build GRCh38
rs972953407 Chr.X: 107153100-107153100 on Build GRCh38
rs1001172608 Chr.X: 107153456-107153456 on Build GRCh38
rs376831639 Chr.X: 107153129-107153129 on Build GRCh38
rs1001587552 Chr.X: 107153434-107153434 on Build GRCh38
rs777705058 Chr.X: 107153379-107153379 on Build GRCh38
rs369367995 Chr.X: 107153130-107153130 on Build GRCh38
rs947195691 Chr.X: 107153422-107153422 on Build GRCh38
rs941227108 Chr.X: 107153080-107153080 on Build GRCh38
rs35155882 Chr.X: 107153276-107153276 on Build GRCh38
rs192513305 Chr.X: 107153065-107153065 on Build GRCh38
rs766572573 Chr.X: 107153205-107153205 on Build GRCh38
rs745411675 Chr.X: 107153138-107153138 on Build GRCh38
rs773241667 Chr.X: 107153198-107153198 on Build GRCh38
rs12845856 Chr.X: 107153091-107153091 on Build GRCh38
rs202006097 Chr.X: 107153217-107153217 on Build GRCh38
rs368485783 Chr.X: 107153328-107153328 on Build GRCh38
rs189019419 Chr.X: 107153027-107153027 on Build GRCh38
rs1013828125 Chr.X: 107153375-107153375 on Build GRCh38
rs373592209 Chr.X: 107153227-107153227 on Build GRCh38
rs769072083 Chr.X: 107153171-107153171 on Build GRCh38
rs1298577 Chr.X: 107153172-107153172 on Build GRCh38
rs772541581 Chr.X: 107153406-107153406 on Build GRCh38
rs749667576 Chr.X: 107153162-107153162 on Build GRCh38
rs1285590 Chr.X: 107153225-107153225 on Build GRCh38
rs749037186 Chr.X: 107153417-107153417 on Build GRCh38
rs772009525 Chr.X: 107153180-107153180 on Build GRCh38
rs748987388 Chr.X: 107153245-107153245 on Build GRCh38
rs369794420 Chr.X: 107153424-107153424 on Build GRCh38
rs762587661 Chr.X: 107153213-107153213 on Build GRCh38
rs748050029 Chr.X: 107153130-107153131 on Build GRCh38
rs780870058 Chr.X: 107153136-107153136 on Build GRCh38
rs953471412 Chr.X: 107153092-107153092 on Build GRCh38
rs1777493 Chr.X: 107153307-107153307 on Build GRCh38
rs758293272 Chr.X: 107153285-107153285 on Build GRCh38
rs779788011 Chr.X: 107153229-107153229 on Build GRCh38
rs763330355 Chr.X: 107153154-107153154 on Build GRCh38
rs760788723 Chr.X: 107153201-107153201 on Build GRCh38
rs999713774 Chr.X: 107153269-107153269 on Build GRCh38
rs769548234 Chr.X: 107153152-107153152 on Build GRCh38
rs905534806 Chr.X: 107153426-107153426 on Build GRCh38
rs145959014 Chr.X: 107153226-107153226 on Build GRCh38

View Full Product Details