Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 107222608-107223144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIH1D3
Entrez Gene ID
139212
Gene Name
PIH1 domain containing 3
Gene Aliases
CXorf41, NYSAR97
UniGene
Hs.128836
Gene Chromosome Location
Chr.X: 107206629-107244243 on Build GRCh38
Transcripts
NM_001169154.1 NM_173494.1 XM_011530855.2

SNP Detail

SNP ID Chromosome Location
rs768075890 Chr.X: 107222734-107222734 on Build GRCh38
rs946126419 Chr.X: 107222765-107222765 on Build GRCh38
rs746947706 Chr.X: 107222870-107222870 on Build GRCh38
rs866273979 Chr.X: 107222753-107222753 on Build GRCh38
rs139957631 Chr.X: 107222958-107222958 on Build GRCh38
rs750645114 Chr.X: 107222831-107222831 on Build GRCh38
rs773857701 Chr.X: 107222893-107222896 on Build GRCh38
rs185104509 Chr.X: 107223103-107223103 on Build GRCh38
rs746244853 Chr.X: 107222872-107222872 on Build GRCh38
rs925297972 Chr.X: 107222894-107222894 on Build GRCh38
rs905464604 Chr.X: 107222973-107222973 on Build GRCh38
rs763547239 Chr.X: 107222796-107222796 on Build GRCh38
rs753613810 Chr.X: 107222711-107222711 on Build GRCh38
rs897961356 Chr.X: 107223105-107223106 on Build GRCh38
rs759265594 Chr.X: 107222835-107222835 on Build GRCh38
rs942558807 Chr.X: 107222705-107222705 on Build GRCh38
rs1057519569 Chr.X: 107222767-107222767 on Build GRCh38
rs776460216 Chr.X: 107222938-107222938 on Build GRCh38
rs774587694 Chr.X: 107223058-107223058 on Build GRCh38
rs775811861 Chr.X: 107222794-107222794 on Build GRCh38
rs976595444 Chr.X: 107222732-107222732 on Build GRCh38
rs746560816 Chr.X: 107222763-107222763 on Build GRCh38
rs1045209972 Chr.X: 107222967-107222967 on Build GRCh38
rs752698361 Chr.X: 107222721-107222721 on Build GRCh38
rs993816443 Chr.X: 107223066-107223066 on Build GRCh38
rs144636752 Chr.X: 107222802-107222802 on Build GRCh38
rs1037917262 Chr.X: 107223019-107223019 on Build GRCh38
rs1056485944 Chr.X: 107222905-107222905 on Build GRCh38
rs879146103 Chr.X: 107222728-107222728 on Build GRCh38
rs762286393 Chr.X: 107222733-107222734 on Build GRCh38
rs778644970 Chr.X: 107222718-107222718 on Build GRCh38
rs370962888 Chr.X: 107222848-107222848 on Build GRCh38
rs770686335 Chr.X: 107222766-107222766 on Build GRCh38
rs935316355 Chr.X: 107222900-107222900 on Build GRCh38
rs746231619 Chr.X: 107222785-107222785 on Build GRCh38
rs757801779 Chr.X: 107222726-107222726 on Build GRCh38
rs754830479 Chr.X: 107222717-107222717 on Build GRCh38
rs780028940 Chr.X: 107222819-107222819 on Build GRCh38
rs898203297 Chr.X: 107223032-107223032 on Build GRCh38
rs778690495 Chr.X: 107222966-107222966 on Build GRCh38
rs979331203 Chr.X: 107222881-107222881 on Build GRCh38
rs267606305 Chr.X: 107222773-107222773 on Build GRCh38
rs759624145 Chr.X: 107223060-107223060 on Build GRCh38
rs752540608 Chr.X: 107222869-107222869 on Build GRCh38
rs756896064 Chr.X: 107222887-107222887 on Build GRCh38
rs777377615 Chr.X: 107222752-107222752 on Build GRCh38
rs955393448 Chr.X: 107222640-107222640 on Build GRCh38
rs1057519568 Chr.X: 107222769-107222775 on Build GRCh38
rs192214441 Chr.X: 107222989-107222989 on Build GRCh38
rs781628085 Chr.X: 107222724-107222724 on Build GRCh38
rs767719280 Chr.X: 107222822-107222822 on Build GRCh38
rs751102925 Chr.X: 107222883-107222883 on Build GRCh38
rs770287553 Chr.X: 107222787-107222787 on Build GRCh38
rs768389578 Chr.X: 107222917-107222917 on Build GRCh38

View Full Product Details