Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 180975632-180976141 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STX6
Entrez Gene ID
10228
Gene Name
syntaxin 6
Gene Aliases
-
UniGene
Hs.518417
Gene Chromosome Location
Chr.1: 180949690-181023121 on Build GRCh38
Transcripts
NM_001286210.1 NM_005819.5 XM_011509058.1 XM_011509059.1 XM_017000006.1 XM_017000007.1

SNP Detail

SNP ID Chromosome Location
rs751833956 Chr.1: 180976108-180976108 on Build GRCh38
rs937424943 Chr.1: 180975872-180975872 on Build GRCh38
rs967310208 Chr.1: 180975819-180975819 on Build GRCh38
rs980585619 Chr.1: 180975993-180975993 on Build GRCh38
rs373631724 Chr.1: 180975789-180975789 on Build GRCh38
rs1006650237 Chr.1: 180976012-180976012 on Build GRCh38
rs989164622 Chr.1: 180975957-180975958 on Build GRCh38
rs898075372 Chr.1: 180975741-180975741 on Build GRCh38
rs939206750 Chr.1: 180976110-180976110 on Build GRCh38
rs560139017 Chr.1: 180975815-180975815 on Build GRCh38
rs865888698 Chr.1: 180975716-180975716 on Build GRCh38
rs776303201 Chr.1: 180975874-180975874 on Build GRCh38
rs142936250 Chr.1: 180975871-180975871 on Build GRCh38
rs915007154 Chr.1: 180975719-180975719 on Build GRCh38
rs1051221055 Chr.1: 180975941-180975941 on Build GRCh38
rs1029316927 Chr.1: 180975764-180975764 on Build GRCh38
rs530087658 Chr.1: 180975980-180975980 on Build GRCh38
rs956913913 Chr.1: 180975945-180975945 on Build GRCh38
rs928632719 Chr.1: 180975856-180975856 on Build GRCh38
rs989112481 Chr.1: 180975698-180975698 on Build GRCh38
rs934988955 Chr.1: 180975688-180975694 on Build GRCh38
rs184179179 Chr.1: 180976079-180976079 on Build GRCh38
rs899151158 Chr.1: 180975858-180975858 on Build GRCh38
rs561977062 Chr.1: 180975801-180975801 on Build GRCh38
rs981864738 Chr.1: 180975838-180975838 on Build GRCh38
rs192180010 Chr.1: 180976058-180976058 on Build GRCh38
rs1011446240 Chr.1: 180976081-180976081 on Build GRCh38
rs1044820341 Chr.1: 180975740-180975740 on Build GRCh38
rs898099415 Chr.1: 180975912-180975912 on Build GRCh38
rs546479342 Chr.1: 180975781-180975781 on Build GRCh38
rs139674833 Chr.1: 180975816-180975816 on Build GRCh38
rs1036607029 Chr.1: 180975691-180975691 on Build GRCh38
rs909293839 Chr.1: 180975901-180975901 on Build GRCh38
rs79356721 Chr.1: 180975875-180975875 on Build GRCh38
rs939178531 Chr.1: 180975783-180975783 on Build GRCh38
rs562537078 Chr.1: 180975720-180975720 on Build GRCh38
rs994261420 Chr.1: 180975756-180975756 on Build GRCh38
rs914935499 Chr.1: 180975958-180975958 on Build GRCh38
rs568191059 Chr.1: 180975955-180975955 on Build GRCh38
rs1018121500 Chr.1: 180975796-180975796 on Build GRCh38
rs752522302 Chr.1: 180975908-180975908 on Build GRCh38
rs41268450 Chr.1: 180975916-180975916 on Build GRCh38
rs1036469441 Chr.1: 180975903-180975903 on Build GRCh38
rs531562089 Chr.1: 180975902-180975902 on Build GRCh38
rs771759269 Chr.1: 180975669-180975669 on Build GRCh38
rs1031260726 Chr.1: 180975922-180975922 on Build GRCh38
rs764216498 Chr.1: 180975954-180975954 on Build GRCh38

View Full Product Details