Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 182397914-182398460 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEDDM1
Entrez Gene ID
127670
Gene Name
transmembrane epididymal protein 1
Gene Aliases
EDDM9, Epdd1, HE9, HEL-S-45e, TMEM45C
UniGene
Hs.156977
Gene Chromosome Location
Chr.1: 182398117-182400616 on Build GRCh38
Transcripts
NM_172000.3

SNP Detail

SNP ID Chromosome Location
rs928900491 Chr.1: 182398210-182398210 on Build GRCh38
rs866744806 Chr.1: 182398109-182398109 on Build GRCh38
rs191497195 Chr.1: 182397976-182397976 on Build GRCh38
rs544650399 Chr.1: 182398158-182398158 on Build GRCh38
rs749566340 Chr.1: 182398308-182398308 on Build GRCh38
rs916273391 Chr.1: 182398289-182398289 on Build GRCh38
rs1043932648 Chr.1: 182397960-182397961 on Build GRCh38
rs79931242 Chr.1: 182398043-182398043 on Build GRCh38
rs909339718 Chr.1: 182398399-182398399 on Build GRCh38
rs935159574 Chr.1: 182398380-182398380 on Build GRCh38
rs1036112858 Chr.1: 182398350-182398350 on Build GRCh38
rs371389226 Chr.1: 182397975-182397976 on Build GRCh38
rs183732391 Chr.1: 182398202-182398202 on Build GRCh38
rs530177523 Chr.1: 182398172-182398172 on Build GRCh38
rs376358899 Chr.1: 182398064-182398064 on Build GRCh38
rs371484921 Chr.1: 182397972-182397972 on Build GRCh38
rs972252311 Chr.1: 182398195-182398195 on Build GRCh38
rs991536560 Chr.1: 182398366-182398366 on Build GRCh38
rs938991710 Chr.1: 182398239-182398239 on Build GRCh38
rs1029055997 Chr.1: 182398030-182398030 on Build GRCh38
rs901776853 Chr.1: 182398409-182398409 on Build GRCh38
rs947782871 Chr.1: 182398306-182398306 on Build GRCh38
rs370651726 Chr.1: 182398402-182398402 on Build GRCh38
rs991588905 Chr.1: 182397951-182397951 on Build GRCh38
rs777578814 Chr.1: 182398041-182398041 on Build GRCh38
rs919499659 Chr.1: 182398432-182398432 on Build GRCh38
rs2038891 Chr.1: 182398413-182398413 on Build GRCh38
rs144958151 Chr.1: 182398338-182398338 on Build GRCh38
rs141306254 Chr.1: 182398250-182398250 on Build GRCh38
rs540963654 Chr.1: 182397984-182397984 on Build GRCh38
rs77451966 Chr.1: 182398170-182398170 on Build GRCh38
rs138828215 Chr.1: 182398435-182398435 on Build GRCh38
rs958783807 Chr.1: 182398352-182398352 on Build GRCh38
rs1020867828 Chr.1: 182398200-182398200 on Build GRCh38
rs758416272 Chr.1: 182398015-182398015 on Build GRCh38
rs903718238 Chr.1: 182397965-182397965 on Build GRCh38
rs901810910 Chr.1: 182398003-182398003 on Build GRCh38
rs1007737111 Chr.1: 182398075-182398075 on Build GRCh38
rs991804372 Chr.1: 182398262-182398262 on Build GRCh38
rs909391979 Chr.1: 182397975-182397975 on Build GRCh38

View Full Product Details