Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 183567075-183567572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCF2
Entrez Gene ID
4688
Gene Name
neutrophil cytosolic factor 2
Gene Aliases
NCF-2, NOXA2, P67-PHOX, P67PHOX
UniGene
Hs.587558
Gene Chromosome Location
Chr.1: 183555562-183590921 on Build GRCh38
Transcripts
NM_000433.3 NM_001127651.2 NM_001190789.1 NM_001190794.1 XM_005245207.1 XM_011509580.1 XM_011509581.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5965733 c.736C>T p.R246C Chr.1: 183567323-183567323 on Build GRCh38
5936908 c.823G>A p.D275N Chr.1: 183567236-183567236 on Build GRCh38
6453682 c.837G>A p.T279T Chr.1: 183567222-183567222 on Build GRCh38
4838378 c.813G>A p.K271K Chr.1: 183567246-183567246 on Build GRCh38
6889557 c.815A>C p.K272T Chr.1: 183567244-183567244 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763690773 Chr.1: 183567194-183567194 on Build GRCh38
rs774251057 Chr.1: 183567171-183567171 on Build GRCh38
rs772674498 Chr.1: 183567408-183567408 on Build GRCh38
rs13306581 Chr.1: 183567223-183567223 on Build GRCh38
rs377217293 Chr.1: 183567358-183567358 on Build GRCh38
rs765026266 Chr.1: 183567451-183567451 on Build GRCh38
rs761796267 Chr.1: 183567209-183567209 on Build GRCh38
rs988863738 Chr.1: 183567337-183567337 on Build GRCh38
rs773016794 Chr.1: 183567181-183567181 on Build GRCh38
rs777953876 Chr.1: 183567375-183567375 on Build GRCh38
rs771349846 Chr.1: 183567282-183567282 on Build GRCh38
rs778431195 Chr.1: 183567264-183567264 on Build GRCh38
rs770127634 Chr.1: 183567395-183567395 on Build GRCh38
rs756393041 Chr.1: 183567157-183567157 on Build GRCh38
rs762465566 Chr.1: 183567189-183567189 on Build GRCh38
rs747834710 Chr.1: 183567527-183567527 on Build GRCh38
rs865973319 Chr.1: 183567333-183567333 on Build GRCh38
rs750380443 Chr.1: 183567326-183567326 on Build GRCh38
rs776684859 Chr.1: 183567369-183567369 on Build GRCh38
rs112923747 Chr.1: 183567399-183567399 on Build GRCh38
rs760718996 Chr.1: 183567409-183567409 on Build GRCh38
rs768181039 Chr.1: 183567322-183567322 on Build GRCh38
rs373262281 Chr.1: 183567184-183567184 on Build GRCh38
rs760561718 Chr.1: 183567335-183567335 on Build GRCh38
rs988183241 Chr.1: 183567487-183567487 on Build GRCh38
rs1049314807 Chr.1: 183567124-183567124 on Build GRCh38
rs770409907 Chr.1: 183567548-183567548 on Build GRCh38
rs771688022 Chr.1: 183567534-183567534 on Build GRCh38
rs764311226 Chr.1: 183567473-183567473 on Build GRCh38
rs781553916 Chr.1: 183567490-183567490 on Build GRCh38
rs6668645 Chr.1: 183567126-183567126 on Build GRCh38
rs759330761 Chr.1: 183567400-183567400 on Build GRCh38
rs980038998 Chr.1: 183567407-183567407 on Build GRCh38
rs766952838 Chr.1: 183567324-183567324 on Build GRCh38
rs539632630 Chr.1: 183567239-183567239 on Build GRCh38
rs766217153 Chr.1: 183567425-183567425 on Build GRCh38
rs201901727 Chr.1: 183567177-183567177 on Build GRCh38
rs199788941 Chr.1: 183567166-183567166 on Build GRCh38
rs751011909 Chr.1: 183567196-183567196 on Build GRCh38
rs772770694 Chr.1: 183567535-183567535 on Build GRCh38
rs762714261 Chr.1: 183567317-183567317 on Build GRCh38
rs780354190 Chr.1: 183567512-183567512 on Build GRCh38
rs370075659 Chr.1: 183567390-183567390 on Build GRCh38
rs768800076 Chr.1: 183567170-183567170 on Build GRCh38
rs752991966 Chr.1: 183567462-183567462 on Build GRCh38
rs933376245 Chr.1: 183567181-183567184 on Build GRCh38
rs879287555 Chr.1: 183567330-183567330 on Build GRCh38
rs776822572 Chr.1: 183567300-183567300 on Build GRCh38
rs774599642 Chr.1: 183567348-183567348 on Build GRCh38
rs576111074 Chr.1: 183567365-183567365 on Build GRCh38
rs758654941 Chr.1: 183567472-183567472 on Build GRCh38
rs956803431 Chr.1: 183567172-183567172 on Build GRCh38
rs761119292 Chr.1: 183567184-183567184 on Build GRCh38
rs753556861 Chr.1: 183567343-183567343 on Build GRCh38
rs774004164 Chr.1: 183567398-183567398 on Build GRCh38
rs759257176 Chr.1: 183567446-183567446 on Build GRCh38
rs757849374 Chr.1: 183567481-183567481 on Build GRCh38
rs746214657 Chr.1: 183567391-183567391 on Build GRCh38
rs755290534 Chr.1: 183567351-183567351 on Build GRCh38
rs371493437 Chr.1: 183567252-183567252 on Build GRCh38
rs778751639 Chr.1: 183567243-183567245 on Build GRCh38
rs747462402 Chr.1: 183567275-183567275 on Build GRCh38
rs776559383 Chr.1: 183567435-183567435 on Build GRCh38
rs993917599 Chr.1: 183567316-183567316 on Build GRCh38
rs772766707 Chr.1: 183567518-183567518 on Build GRCh38
rs746326365 Chr.1: 183567492-183567492 on Build GRCh38
rs745648528 Chr.1: 183567156-183567156 on Build GRCh38
rs748228594 Chr.1: 183567254-183567254 on Build GRCh38
rs767437169 Chr.1: 183567210-183567210 on Build GRCh38
rs775224104 Chr.1: 183567188-183567188 on Build GRCh38
rs751675908 Chr.1: 183567477-183567477 on Build GRCh38
rs1030451234 Chr.1: 183567506-183567506 on Build GRCh38
rs747550232 Chr.1: 183567387-183567387 on Build GRCh38
rs769538131 Chr.1: 183567446-183567447 on Build GRCh38
rs761315970 Chr.1: 183567323-183567323 on Build GRCh38
rs986166767 Chr.1: 183567532-183567532 on Build GRCh38
rs907491789 Chr.1: 183567499-183567499 on Build GRCh38
rs554005946 Chr.1: 183567219-183567219 on Build GRCh38
rs749410546 Chr.1: 183567168-183567168 on Build GRCh38
rs771356784 Chr.1: 183567185-183567185 on Build GRCh38
rs1000185935 Chr.1: 183567302-183567302 on Build GRCh38
rs374822007 Chr.1: 183567160-183567160 on Build GRCh38
rs923268186 Chr.1: 183567178-183567178 on Build GRCh38
rs772775720 Chr.1: 183567262-183567262 on Build GRCh38
rs942486247 Chr.1: 183567222-183567222 on Build GRCh38
rs557535861 Chr.1: 183567243-183567243 on Build GRCh38
rs756453916 Chr.1: 183567501-183567501 on Build GRCh38
rs766232480 Chr.1: 183567341-183567341 on Build GRCh38
rs754118806 Chr.1: 183567238-183567238 on Build GRCh38
rs940452486 Chr.1: 183567523-183567523 on Build GRCh38
rs746501134 Chr.1: 183567541-183567541 on Build GRCh38

View Full Product Details