Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 184072140-184072680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSEN15
Entrez Gene ID
116461
Gene Name
tRNA splicing endonuclease subunit 15
Gene Aliases
C1orf19, PCH2F, sen15
UniGene
Hs.548197
Gene Chromosome Location
Chr.1: 184050207-184095846 on Build GRCh38
Transcripts
NM_001127394.3 NM_001300764.1 NM_001300766.1 NM_052965.3 NR_023349.2 NR_125335.1 XM_006711148.1 XM_011509139.2 XM_017000228.1 XM_017000229.1 XM_017000230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
165205 c.400G>C p.D134H Chr.1: 184072203-184072203 on Build GRCh38
4576671 c.379C>T p.R127* Chr.1: 184072182-184072182 on Build GRCh38
1601372 c.434A>T p.E145V Chr.1: 184072237-184072237 on Build GRCh38
4432827 c.435G>T p.E145D Chr.1: 184072238-184072238 on Build GRCh38
1337059 c.467C>T p.T156I Chr.1: 184072270-184072270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs910638610 Chr.1: 184072577-184072577 on Build GRCh38
rs946476056 Chr.1: 184072498-184072498 on Build GRCh38
rs762141345 Chr.1: 184072182-184072182 on Build GRCh38
rs573629773 Chr.1: 184072493-184072493 on Build GRCh38
rs544059998 Chr.1: 184072442-184072442 on Build GRCh38
rs895490937 Chr.1: 184072369-184072369 on Build GRCh38
rs184488794 Chr.1: 184072183-184072183 on Build GRCh38
rs542288432 Chr.1: 184072233-184072233 on Build GRCh38
rs55827728 Chr.1: 184072204-184072204 on Build GRCh38
rs57880810 Chr.1: 184072592-184072592 on Build GRCh38
rs369212777 Chr.1: 184072313-184072313 on Build GRCh38
rs923338506 Chr.1: 184072232-184072232 on Build GRCh38
rs942197905 Chr.1: 184072579-184072579 on Build GRCh38
rs759310152 Chr.1: 184072192-184072192 on Build GRCh38
rs751142686 Chr.1: 184072229-184072229 on Build GRCh38
rs771497560 Chr.1: 184072362-184072362 on Build GRCh38
rs145403047 Chr.1: 184072634-184072634 on Build GRCh38
rs914988012 Chr.1: 184072469-184072469 on Build GRCh38
rs749782700 Chr.1: 184072255-184072255 on Build GRCh38
rs138873313 Chr.1: 184072626-184072627 on Build GRCh38
rs1031999445 Chr.1: 184072496-184072496 on Build GRCh38
rs543212100 Chr.1: 184072418-184072419 on Build GRCh38
rs988530083 Chr.1: 184072506-184072506 on Build GRCh38
rs373043340 Chr.1: 184072322-184072322 on Build GRCh38
rs551218468 Chr.1: 184072609-184072609 on Build GRCh38
rs398039072 Chr.1: 184072505-184072506 on Build GRCh38
rs555398865 Chr.1: 184072256-184072256 on Build GRCh38
rs372868359 Chr.1: 184072168-184072168 on Build GRCh38
rs764974917 Chr.1: 184072319-184072319 on Build GRCh38
rs146745937 Chr.1: 184072352-184072352 on Build GRCh38
rs572205936 Chr.1: 184072350-184072350 on Build GRCh38
rs377553264 Chr.1: 184072291-184072291 on Build GRCh38
rs199657425 Chr.1: 184072211-184072211 on Build GRCh38
rs879253779 Chr.1: 184072258-184072258 on Build GRCh38
rs777274418 Chr.1: 184072221-184072221 on Build GRCh38
rs767166549 Chr.1: 184072342-184072342 on Build GRCh38
rs770094658 Chr.1: 184072246-184072246 on Build GRCh38
rs375946603 Chr.1: 184072214-184072214 on Build GRCh38
rs986143788 Chr.1: 184072544-184072544 on Build GRCh38
rs778507833 Chr.1: 184072244-184072244 on Build GRCh38
rs34615524 Chr.1: 184072496-184072497 on Build GRCh38
rs562251971 Chr.1: 184072453-184072453 on Build GRCh38
rs535467457 Chr.1: 184072325-184072325 on Build GRCh38
rs200328966 Chr.1: 184072196-184072196 on Build GRCh38
rs993560808 Chr.1: 184072351-184072351 on Build GRCh38
rs374264447 Chr.1: 184072251-184072251 on Build GRCh38
rs956012520 Chr.1: 184072505-184072505 on Build GRCh38
rs765202343 Chr.1: 184072210-184072210 on Build GRCh38
rs954721848 Chr.1: 184072520-184072520 on Build GRCh38
rs140346547 Chr.1: 184072439-184072439 on Build GRCh38
rs746182575 Chr.1: 184072531-184072531 on Build GRCh38
rs576634243 Chr.1: 184072495-184072496 on Build GRCh38
rs753665502 Chr.1: 184072187-184072187 on Build GRCh38
rs1028472365 Chr.1: 184072293-184072293 on Build GRCh38
rs369732365 Chr.1: 184072241-184072241 on Build GRCh38
rs939218819 Chr.1: 184072597-184072597 on Build GRCh38
rs756895353 Chr.1: 184072230-184072230 on Build GRCh38
rs768269293 Chr.1: 184072321-184072321 on Build GRCh38
rs1030307446 Chr.1: 184072507-184072507 on Build GRCh38
rs759611164 Chr.1: 184072481-184072481 on Build GRCh38
rs1050488037 Chr.1: 184072243-184072243 on Build GRCh38
rs576582464 Chr.1: 184072441-184072441 on Build GRCh38
rs773357701 Chr.1: 184072306-184072306 on Build GRCh38
rs780879464 Chr.1: 184072235-184072235 on Build GRCh38
rs541022812 Chr.1: 184072496-184072496 on Build GRCh38
rs190588086 Chr.1: 184072589-184072589 on Build GRCh38
rs762053126 Chr.1: 184072274-184072274 on Build GRCh38
rs988251198 Chr.1: 184072468-184072468 on Build GRCh38
rs1000069664 Chr.1: 184072484-184072484 on Build GRCh38
rs887066287 Chr.1: 184072628-184072628 on Build GRCh38
rs895451326 Chr.1: 184072635-184072635 on Build GRCh38
rs143508231 Chr.1: 184072338-184072338 on Build GRCh38

View Full Product Details