Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 109536752-109537255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NXT2
Entrez Gene ID
55916
Gene Name
nuclear transport factor 2 like export factor 2
Gene Aliases
P15-2
UniGene
Hs.25010
Gene Chromosome Location
Chr.X: 109535781-109544698 on Build GRCh38
Transcripts
NM_001242617.1 NM_001242618.1 NM_018698.4 XM_011530989.2

SNP Detail

SNP ID Chromosome Location
rs148682193 Chr.X: 109536945-109536945 on Build GRCh38
rs765141125 Chr.X: 109536967-109536967 on Build GRCh38
rs759117283 Chr.X: 109537198-109537198 on Build GRCh38
rs896886033 Chr.X: 109537192-109537192 on Build GRCh38
rs371138421 Chr.X: 109537020-109537020 on Build GRCh38
rs1049724067 Chr.X: 109537169-109537169 on Build GRCh38
rs774785226 Chr.X: 109536927-109536927 on Build GRCh38
rs188446532 Chr.X: 109537088-109537088 on Build GRCh38
rs747679339 Chr.X: 109537022-109537023 on Build GRCh38
rs998421153 Chr.X: 109536920-109536920 on Build GRCh38
rs776564437 Chr.X: 109536961-109536961 on Build GRCh38
rs751075998 Chr.X: 109536976-109536976 on Build GRCh38
rs941001921 Chr.X: 109537155-109537155 on Build GRCh38
rs369385435 Chr.X: 109537234-109537234 on Build GRCh38
rs73639133 Chr.X: 109537035-109537035 on Build GRCh38
rs373934009 Chr.X: 109537060-109537060 on Build GRCh38
rs776374806 Chr.X: 109537143-109537143 on Build GRCh38
rs763624983 Chr.X: 109536975-109536975 on Build GRCh38
rs752689038 Chr.X: 109536904-109536904 on Build GRCh38
rs771028580 Chr.X: 109536848-109536848 on Build GRCh38
rs770525699 Chr.X: 109536915-109536915 on Build GRCh38
rs761004672 Chr.X: 109537047-109537047 on Build GRCh38
rs948436764 Chr.X: 109536833-109536833 on Build GRCh38
rs1036817929 Chr.X: 109537190-109537190 on Build GRCh38
rs1005311978 Chr.X: 109536874-109536874 on Build GRCh38
rs749772750 Chr.X: 109537024-109537024 on Build GRCh38
rs1008208361 Chr.X: 109537183-109537183 on Build GRCh38
rs370503612 Chr.X: 109537058-109537058 on Build GRCh38
rs956191057 Chr.X: 109536851-109536851 on Build GRCh38
rs772200308 Chr.X: 109537040-109537040 on Build GRCh38
rs779107277 Chr.X: 109536906-109536906 on Build GRCh38
rs1008451947 Chr.X: 109536828-109536828 on Build GRCh38
rs776127712 Chr.X: 109537148-109537148 on Build GRCh38
rs142278164 Chr.X: 109536991-109536991 on Build GRCh38
rs745522079 Chr.X: 109537006-109537006 on Build GRCh38
rs770279889 Chr.X: 109536926-109536926 on Build GRCh38
rs758302732 Chr.X: 109536973-109536973 on Build GRCh38
rs762074597 Chr.X: 109536951-109536951 on Build GRCh38
rs768732444 Chr.X: 109536941-109536941 on Build GRCh38
rs781512474 Chr.X: 109536919-109536919 on Build GRCh38
rs1053299711 Chr.X: 109537185-109537185 on Build GRCh38
rs956260726 Chr.X: 109537041-109537041 on Build GRCh38
rs1011711643 Chr.X: 109537201-109537201 on Build GRCh38
rs773512751 Chr.X: 109537044-109537044 on Build GRCh38
rs775907051 Chr.X: 109536928-109536928 on Build GRCh38
rs961293367 Chr.X: 109536819-109536827 on Build GRCh38
rs954083660 Chr.X: 109536980-109536980 on Build GRCh38
rs759340088 Chr.X: 109536963-109536963 on Build GRCh38
rs376162548 Chr.X: 109537042-109537042 on Build GRCh38
rs775347147 Chr.X: 109537052-109537052 on Build GRCh38
rs1022213740 Chr.X: 109537224-109537224 on Build GRCh38
rs932728404 Chr.X: 109537159-109537159 on Build GRCh38
rs374060059 Chr.X: 109536895-109536895 on Build GRCh38
rs745324299 Chr.X: 109537218-109537218 on Build GRCh38
rs780887533 Chr.X: 109536999-109536999 on Build GRCh38
rs777623807 Chr.X: 109536982-109536982 on Build GRCh38
rs770738124 Chr.X: 109537132-109537132 on Build GRCh38
rs752636225 Chr.X: 109536969-109536969 on Build GRCh38
rs892083703 Chr.X: 109537194-109537194 on Build GRCh38
rs374876254 Chr.X: 109537039-109537039 on Build GRCh38
rs765088351 Chr.X: 109537056-109537056 on Build GRCh38
rs767692077 Chr.X: 109536952-109536952 on Build GRCh38
rs199510422 Chr.X: 109537061-109537061 on Build GRCh38
rs1044099873 Chr.X: 109536840-109536840 on Build GRCh38
rs1006738874 Chr.X: 109537028-109537028 on Build GRCh38
rs377649567 Chr.X: 109536909-109536909 on Build GRCh38
rs746286020 Chr.X: 109536922-109536922 on Build GRCh38
rs769093033 Chr.X: 109537033-109537033 on Build GRCh38
rs891114919 Chr.X: 109537182-109537182 on Build GRCh38
rs775461119 Chr.X: 109537057-109537057 on Build GRCh38
rs896042681 Chr.X: 109536801-109536801 on Build GRCh38
rs746628863 Chr.X: 109536914-109536914 on Build GRCh38
rs756411347 Chr.X: 109537012-109537012 on Build GRCh38
rs779063051 Chr.X: 109536902-109536902 on Build GRCh38
rs749808445 Chr.X: 109536933-109536933 on Build GRCh38

View Full Product Details