Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 110173966-110174475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM164
Entrez Gene ID
84187
Gene Name
transmembrane protein 164
Gene Aliases
bB360B22.3
UniGene
Hs.496572
Gene Chromosome Location
Chr.X: 110002413-110184438 on Build GRCh38
Transcripts
NM_017698.2 NM_032227.3 XM_005262205.4 XM_005262206.3 XM_005262208.4 XM_011531051.2 XM_011531054.1 XM_011531055.2 XM_017029894.1 XM_017029896.1 XM_017029897.1 XM_017029899.1 XM_017029900.1

SNP Detail

SNP ID Chromosome Location
rs915109557 Chr.X: 110174207-110174207 on Build GRCh38
rs186423425 Chr.X: 110174427-110174427 on Build GRCh38
rs866502414 Chr.X: 110174383-110174383 on Build GRCh38
rs1028167000 Chr.X: 110174227-110174227 on Build GRCh38
rs1002662593 Chr.X: 110174030-110174030 on Build GRCh38
rs1035347393 Chr.X: 110174040-110174040 on Build GRCh38
rs866318701 Chr.X: 110174365-110174365 on Build GRCh38
rs937379760 Chr.X: 110174451-110174451 on Build GRCh38
rs866735212 Chr.X: 110174387-110174387 on Build GRCh38
rs1002712652 Chr.X: 110174001-110174001 on Build GRCh38
rs111943015 Chr.X: 110174357-110174357 on Build GRCh38
rs946535309 Chr.X: 110174228-110174228 on Build GRCh38
rs867025287 Chr.X: 110174370-110174370 on Build GRCh38
rs988447521 Chr.X: 110174079-110174079 on Build GRCh38
rs976735559 Chr.X: 110174349-110174349 on Build GRCh38
rs894298016 Chr.X: 110174066-110174066 on Build GRCh38
rs753408976 Chr.X: 110174156-110174156 on Build GRCh38
rs1012361064 Chr.X: 110174077-110174077 on Build GRCh38
rs966315362 Chr.X: 110174340-110174340 on Build GRCh38
rs1004888249 Chr.X: 110174293-110174293 on Build GRCh38
rs1014897658 Chr.X: 110174301-110174301 on Build GRCh38
rs767946603 Chr.X: 110174246-110174246 on Build GRCh38
rs1055666399 Chr.X: 110174048-110174048 on Build GRCh38
rs927364695 Chr.X: 110174364-110174364 on Build GRCh38
rs867278508 Chr.X: 110174034-110174034 on Build GRCh38
rs867610661 Chr.X: 110174376-110174376 on Build GRCh38
rs952085282 Chr.X: 110174229-110174229 on Build GRCh38

View Full Product Details