Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 111298010-111298509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCX
Entrez Gene ID
1641
Gene Name
doublecortin
Gene Aliases
DBCN, DC, LISX, SCLH, XLIS
UniGene
Hs.34780
Gene Chromosome Location
Chr.X: 111293779-111412232 on Build GRCh38
Transcripts
NM_000555.3 NM_001195553.1 NM_178151.2 NM_178152.2 NM_178153.2 XM_011530878.2 XM_011530879.2 XM_011530880.2 XM_017029312.1

SNP Detail

SNP ID Chromosome Location
rs934822149 Chr.X: 111298464-111298464 on Build GRCh38
rs115396024 Chr.X: 111298201-111298201 on Build GRCh38
rs948742222 Chr.X: 111298065-111298065 on Build GRCh38
rs781483602 Chr.X: 111298096-111298096 on Build GRCh38
rs1027549173 Chr.X: 111298267-111298267 on Build GRCh38
rs893166632 Chr.X: 111298051-111298051 on Build GRCh38
rs932944017 Chr.X: 111298334-111298334 on Build GRCh38
rs907626156 Chr.X: 111298292-111298292 on Build GRCh38
rs773249937 Chr.X: 111298115-111298115 on Build GRCh38
rs924748491 Chr.X: 111298393-111298393 on Build GRCh38
rs182066939 Chr.X: 111298081-111298081 on Build GRCh38
rs112645866 Chr.X: 111298108-111298108 on Build GRCh38
rs902646225 Chr.X: 111298432-111298432 on Build GRCh38
rs1011009404 Chr.X: 111298045-111298045 on Build GRCh38
rs1002718368 Chr.X: 111298102-111298102 on Build GRCh38
rs1003503156 Chr.X: 111298181-111298181 on Build GRCh38
rs1021008138 Chr.X: 111298088-111298088 on Build GRCh38
rs780161075 Chr.X: 111298323-111298323 on Build GRCh38
rs902573109 Chr.X: 111298124-111298124 on Build GRCh38
rs944435241 Chr.X: 111298314-111298314 on Build GRCh38
rs867833022 Chr.X: 111298371-111298371 on Build GRCh38
rs1050452624 Chr.X: 111298363-111298363 on Build GRCh38
rs145443853 Chr.X: 111298266-111298266 on Build GRCh38
rs991125590 Chr.X: 111298261-111298261 on Build GRCh38
rs964758123 Chr.X: 111298294-111298294 on Build GRCh38
rs915467894 Chr.X: 111298366-111298366 on Build GRCh38
rs1016014441 Chr.X: 111298290-111298290 on Build GRCh38
rs1042495871 Chr.X: 111298381-111298381 on Build GRCh38
rs749322250 Chr.X: 111298369-111298369 on Build GRCh38
rs959346329 Chr.X: 111298241-111298241 on Build GRCh38
rs73545251 Chr.X: 111298276-111298276 on Build GRCh38
rs983247358 Chr.X: 111298289-111298289 on Build GRCh38
rs142522253 Chr.X: 111298328-111298328 on Build GRCh38
rs765456325 Chr.X: 111298467-111298467 on Build GRCh38

View Full Product Details