Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 119080778-119081324 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1210
Entrez Gene ID
57481
Gene Name
KIAA1210
Gene Aliases
-
UniGene
Hs.97594
Gene Chromosome Location
Chr.X: 119078635-119151082 on Build GRCh38
Transcripts
NM_020721.1 XM_006724774.3 XM_017029688.1 XM_017029689.1

SNP Detail

SNP ID Chromosome Location
rs879237701 Chr.X: 119081290-119081290 on Build GRCh38
rs766149150 Chr.X: 119081282-119081282 on Build GRCh38
rs1031047477 Chr.X: 119080916-119080916 on Build GRCh38
rs192696948 Chr.X: 119081254-119081254 on Build GRCh38
rs761688867 Chr.X: 119081281-119081282 on Build GRCh38
rs755756915 Chr.X: 119081302-119081303 on Build GRCh38
rs759817111 Chr.X: 119081222-119081222 on Build GRCh38
rs780693097 Chr.X: 119081291-119081291 on Build GRCh38
rs947917748 Chr.X: 119080978-119080978 on Build GRCh38
rs771640744 Chr.X: 119081269-119081280 on Build GRCh38
rs749412145 Chr.X: 119081144-119081144 on Build GRCh38
rs927754355 Chr.X: 119081199-119081199 on Build GRCh38
rs12391835 Chr.X: 119080989-119080989 on Build GRCh38
rs745438522 Chr.X: 119081291-119081292 on Build GRCh38
rs957723790 Chr.X: 119081173-119081173 on Build GRCh38
rs969252518 Chr.X: 119081191-119081191 on Build GRCh38
rs755879239 Chr.X: 119081280-119081280 on Build GRCh38
rs376072164 Chr.X: 119081192-119081192 on Build GRCh38
rs956556646 Chr.X: 119080908-119080908 on Build GRCh38
rs747072312 Chr.X: 119081300-119081300 on Build GRCh38
rs1007074466 Chr.X: 119081117-119081117 on Build GRCh38
rs772987230 Chr.X: 119081125-119081125 on Build GRCh38
rs766416107 Chr.X: 119081284-119081285 on Build GRCh38
rs769775913 Chr.X: 119081107-119081107 on Build GRCh38
rs34785358 Chr.X: 119081288-119081289 on Build GRCh38
rs765625208 Chr.X: 119081289-119081290 on Build GRCh38
rs1030703749 Chr.X: 119081172-119081172 on Build GRCh38
rs916479785 Chr.X: 119080966-119080966 on Build GRCh38
rs762564343 Chr.X: 119080987-119080987 on Build GRCh38
rs866139825 Chr.X: 119081294-119081294 on Build GRCh38
rs770897459 Chr.X: 119081159-119081171 on Build GRCh38
rs941733011 Chr.X: 119081024-119081024 on Build GRCh38
rs1057337657 Chr.X: 119081096-119081096 on Build GRCh38
rs931302005 Chr.X: 119081077-119081077 on Build GRCh38
rs965473705 Chr.X: 119080814-119080814 on Build GRCh38
rs759821109 Chr.X: 119081289-119081291 on Build GRCh38
rs796660599 Chr.X: 119081289-119081290 on Build GRCh38
rs938726363 Chr.X: 119081016-119081016 on Build GRCh38
rs989093279 Chr.X: 119081183-119081183 on Build GRCh38
rs748661545 Chr.X: 119081295-119081295 on Build GRCh38
rs772360075 Chr.X: 119081223-119081223 on Build GRCh38
rs904543306 Chr.X: 119081131-119081131 on Build GRCh38
rs10714793 Chr.X: 119081289-119081289 on Build GRCh38
rs753502463 Chr.X: 119081289-119081289 on Build GRCh38
rs1037043213 Chr.X: 119081017-119081017 on Build GRCh38
rs182306116 Chr.X: 119081160-119081160 on Build GRCh38
rs776589669 Chr.X: 119081288-119081290 on Build GRCh38
rs1023638503 Chr.X: 119081143-119081143 on Build GRCh38
rs1023519710 Chr.X: 119081189-119081189 on Build GRCh38
rs1037473329 Chr.X: 119081033-119081033 on Build GRCh38
rs753083591 Chr.X: 119081290-119081295 on Build GRCh38
rs1013169231 Chr.X: 119081123-119081124 on Build GRCh38
rs1000213295 Chr.X: 119081140-119081140 on Build GRCh38
rs372881706 Chr.X: 119081157-119081157 on Build GRCh38
rs754836474 Chr.X: 119081002-119081002 on Build GRCh38
rs755304007 Chr.X: 119081291-119081291 on Build GRCh38
rs986162411 Chr.X: 119081003-119081003 on Build GRCh38
rs1048399460 Chr.X: 119081288-119081288 on Build GRCh38
rs910228293 Chr.X: 119081012-119081012 on Build GRCh38
rs918658982 Chr.X: 119081237-119081237 on Build GRCh38
rs977785894 Chr.X: 119080873-119080873 on Build GRCh38
rs1032104537 Chr.X: 119080872-119080872 on Build GRCh38
rs1020159452 Chr.X: 119081124-119081124 on Build GRCh38
rs762562695 Chr.X: 119081299-119081299 on Build GRCh38
rs957691519 Chr.X: 119080924-119080924 on Build GRCh38
rs187736216 Chr.X: 119081238-119081238 on Build GRCh38
rs183062841 Chr.X: 119080984-119080984 on Build GRCh38
rs187750732 Chr.X: 119081053-119081053 on Build GRCh38
rs190683766 Chr.X: 119081089-119081089 on Build GRCh38
rs749921112 Chr.X: 119081279-119081279 on Build GRCh38
rs199760859 Chr.X: 119081258-119081259 on Build GRCh38
rs988015164 Chr.X: 119080933-119080933 on Build GRCh38
rs367944230 Chr.X: 119081139-119081139 on Build GRCh38
rs897603894 Chr.X: 119081046-119081046 on Build GRCh38
rs376218437 Chr.X: 119081290-119081291 on Build GRCh38
rs112292937 Chr.X: 119080961-119080961 on Build GRCh38
rs369101504 Chr.X: 119081146-119081146 on Build GRCh38
rs757705673 Chr.X: 119081291-119081294 on Build GRCh38
rs774882310 Chr.X: 119081267-119081268 on Build GRCh38
rs764392260 Chr.X: 119081290-119081290 on Build GRCh38
rs774997388 Chr.X: 119080973-119080973 on Build GRCh38

View Full Product Details