Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 120526145-120526672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL4B
Entrez Gene ID
8450
Gene Name
cullin 4B
Gene Aliases
CUL-4B, MRXHF2, MRXS15, MRXSC, SFM2
UniGene
Hs.102914
Gene Chromosome Location
Chr.X: 120524589-120575829 on Build GRCh38
Transcripts
NM_001079872.1 NM_003588.3 XM_005262481.1 XM_006724784.1 XM_006724785.2 XM_011531399.2 XM_011531400.2 XM_011531401.1

SNP Detail

SNP ID Chromosome Location
rs918787272 Chr.X: 120526302-120526302 on Build GRCh38
rs953747450 Chr.X: 120526173-120526173 on Build GRCh38
rs939488410 Chr.X: 120526313-120526313 on Build GRCh38
rs766008370 Chr.X: 120526446-120526446 on Build GRCh38
rs35267132 Chr.X: 120526446-120526447 on Build GRCh38
rs764820525 Chr.X: 120526327-120526330 on Build GRCh38
rs973371951 Chr.X: 120526296-120526296 on Build GRCh38
rs749994745 Chr.X: 120526334-120526334 on Build GRCh38
rs776630950 Chr.X: 120526185-120526185 on Build GRCh38
rs988160940 Chr.X: 120526548-120526548 on Build GRCh38
rs1017275858 Chr.X: 120526442-120526442 on Build GRCh38
rs963000601 Chr.X: 120526192-120526192 on Build GRCh38
rs761902157 Chr.X: 120526286-120526286 on Build GRCh38
rs1056598656 Chr.X: 120526377-120526377 on Build GRCh38
rs916666838 Chr.X: 120526446-120526447 on Build GRCh38
rs878948978 Chr.X: 120526324-120526324 on Build GRCh38
rs199865454 Chr.X: 120526426-120526429 on Build GRCh38
rs915191963 Chr.X: 120526582-120526582 on Build GRCh38
rs1043743337 Chr.X: 120526563-120526563 on Build GRCh38
rs185867059 Chr.X: 120526597-120526597 on Build GRCh38

View Full Product Details