Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 133216587-133217083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TFDP3
Entrez Gene ID
51270
Gene Name
transcription factor Dp family member 3
Gene Aliases
CT30, DP4, HCA661
UniGene
Hs.142908
Gene Chromosome Location
Chr.X: 133216669-133218348 on Build GRCh38
Transcripts
NM_016521.2

SNP Detail

SNP ID Chromosome Location
rs930360372 Chr.X: 133216876-133216876 on Build GRCh38
rs747706717 Chr.X: 133216994-133216994 on Build GRCh38
rs757845711 Chr.X: 133217033-133217033 on Build GRCh38
rs869108913 Chr.X: 133216975-133216976 on Build GRCh38
rs767721939 Chr.X: 133216795-133216795 on Build GRCh38
rs765232764 Chr.X: 133217007-133217008 on Build GRCh38
rs761922289 Chr.X: 133216780-133216780 on Build GRCh38
rs911802909 Chr.X: 133216835-133216835 on Build GRCh38
rs138184941 Chr.X: 133216863-133216863 on Build GRCh38
rs771826932 Chr.X: 133216995-133216995 on Build GRCh38
rs888627436 Chr.X: 133216622-133216622 on Build GRCh38
rs964218626 Chr.X: 133216914-133216914 on Build GRCh38
rs759947075 Chr.X: 133217040-133217040 on Build GRCh38
rs888575472 Chr.X: 133216924-133216924 on Build GRCh38
rs1001551006 Chr.X: 133216625-133216625 on Build GRCh38
rs901900707 Chr.X: 133216955-133216955 on Build GRCh38
rs1039089140 Chr.X: 133216868-133216868 on Build GRCh38
rs1018428492 Chr.X: 133216901-133216901 on Build GRCh38
rs775801098 Chr.X: 133217063-133217063 on Build GRCh38
rs1010637067 Chr.X: 133216677-133216677 on Build GRCh38
rs779984500 Chr.X: 133216649-133216649 on Build GRCh38
rs753555290 Chr.X: 133216916-133216916 on Build GRCh38
rs752635426 Chr.X: 133216975-133216977 on Build GRCh38
rs879103347 Chr.X: 133216974-133216975 on Build GRCh38
rs756224748 Chr.X: 133216821-133216821 on Build GRCh38
rs960319866 Chr.X: 133216657-133216657 on Build GRCh38
rs776942014 Chr.X: 133217038-133217038 on Build GRCh38
rs997485080 Chr.X: 133216968-133216968 on Build GRCh38
rs190448749 Chr.X: 133216875-133216875 on Build GRCh38
rs766665780 Chr.X: 133216893-133216893 on Build GRCh38
rs145651503 Chr.X: 133216612-133216612 on Build GRCh38
rs896044070 Chr.X: 133217057-133217057 on Build GRCh38
rs895935555 Chr.X: 133216666-133216666 on Build GRCh38
rs754791329 Chr.X: 133216928-133216928 on Build GRCh38
rs111884715 Chr.X: 133216975-133216975 on Build GRCh38
rs765474569 Chr.X: 133217053-133217053 on Build GRCh38
rs925749975 Chr.X: 133216793-133216793 on Build GRCh38
rs751468810 Chr.X: 133216688-133216688 on Build GRCh38
rs751525612 Chr.X: 133217039-133217040 on Build GRCh38
rs922328541 Chr.X: 133217015-133217015 on Build GRCh38
rs978888984 Chr.X: 133216788-133216788 on Build GRCh38
rs772888267 Chr.X: 133217007-133217007 on Build GRCh38
rs368919287 Chr.X: 133216988-133216989 on Build GRCh38
rs746511755 Chr.X: 133217012-133217012 on Build GRCh38
rs1022995202 Chr.X: 133216744-133216744 on Build GRCh38
rs970528898 Chr.X: 133216751-133216751 on Build GRCh38
rs775016802 Chr.X: 133216650-133216650 on Build GRCh38

View Full Product Details