Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 139726627-139727136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP11C
Entrez Gene ID
286410
Gene Name
ATPase phospholipid transporting 11C
Gene Aliases
ATPIG, ATPIQ
UniGene
Hs.88252
Gene Chromosome Location
Chr.X: 139726346-139933083 on Build GRCh38
Transcripts
NM_001010986.2 NM_173694.4 XM_017029439.1 XM_017029440.1 XM_017029441.1 XM_017029442.1 XM_017029443.1 XM_017029444.1 XM_017029446.1 XM_017029447.1 XM_017029448.1 XM_017029449.1

SNP Detail

SNP ID Chromosome Location
rs900097823 Chr.X: 139726811-139726811 on Build GRCh38
rs1011494699 Chr.X: 139726769-139726769 on Build GRCh38
rs1052558526 Chr.X: 139726762-139726762 on Build GRCh38
rs774362140 Chr.X: 139726935-139726936 on Build GRCh38
rs762422169 Chr.X: 139727050-139727050 on Build GRCh38
rs977065041 Chr.X: 139726734-139726734 on Build GRCh38
rs1014628324 Chr.X: 139726862-139726862 on Build GRCh38
rs987146680 Chr.X: 139726844-139726844 on Build GRCh38
rs753163402 Chr.X: 139726940-139726940 on Build GRCh38
rs1045461077 Chr.X: 139726712-139726712 on Build GRCh38
rs1003628887 Chr.X: 139726746-139726746 on Build GRCh38
rs920466537 Chr.X: 139726897-139726897 on Build GRCh38
rs938982097 Chr.X: 139726969-139726969 on Build GRCh38
rs942020124 Chr.X: 139726827-139726827 on Build GRCh38
rs948050019 Chr.X: 139726658-139726658 on Build GRCh38
rs1032908175 Chr.X: 139727016-139727016 on Build GRCh38
rs763764506 Chr.X: 139727072-139727072 on Build GRCh38
rs767489333 Chr.X: 139727003-139727003 on Build GRCh38
rs557806667 Chr.X: 139727105-139727105 on Build GRCh38
rs980754145 Chr.X: 139726910-139726910 on Build GRCh38
rs906557626 Chr.X: 139726713-139726713 on Build GRCh38
rs973656427 Chr.X: 139726875-139726875 on Build GRCh38
rs144918632 Chr.X: 139727010-139727010 on Build GRCh38
rs1010790968 Chr.X: 139727087-139727087 on Build GRCh38
rs748269830 Chr.X: 139726838-139726838 on Build GRCh38
rs184990901 Chr.X: 139727051-139727051 on Build GRCh38
rs146438992 Chr.X: 139726822-139726822 on Build GRCh38
rs961878216 Chr.X: 139726873-139726873 on Build GRCh38
rs983574897 Chr.X: 139726785-139726785 on Build GRCh38
rs892604567 Chr.X: 139726767-139726767 on Build GRCh38
rs747367234 Chr.X: 139726778-139726778 on Build GRCh38
rs142539677 Chr.X: 139726793-139726793 on Build GRCh38
rs951268795 Chr.X: 139726784-139726784 on Build GRCh38
rs927944998 Chr.X: 139726935-139726935 on Build GRCh38
rs947948845 Chr.X: 139726907-139726907 on Build GRCh38
rs760190619 Chr.X: 139726726-139726726 on Build GRCh38
rs997131025 Chr.X: 139726817-139726817 on Build GRCh38
rs759453266 Chr.X: 139726951-139726951 on Build GRCh38
rs2805901 Chr.X: 139726843-139726843 on Build GRCh38
rs892571992 Chr.X: 139727019-139727019 on Build GRCh38
rs965302789 Chr.X: 139726728-139726729 on Build GRCh38

View Full Product Details