Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 192810280-192810793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RGS2
Entrez Gene ID
5997
Gene Name
regulator of G-protein signaling 2
Gene Aliases
G0S8
UniGene
Hs.78944
Gene Chromosome Location
Chr.1: 192809039-192812277 on Build GRCh38
Transcripts
NM_002923.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4842859 c.232C>T p.Q78* Chr.1: 192810389-192810389 on Build GRCh38
3418533 c.247G>T p.A83S Chr.1: 192810404-192810404 on Build GRCh38
4576690 c.274+2_274+6delTAAGT p.? Chr.1: 192810433-192810437 on Build GRCh38
6765864 c.229G>A p.A77T Chr.1: 192810386-192810386 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs866910811 Chr.1: 192810567-192810567 on Build GRCh38
rs958492888 Chr.1: 192810420-192810420 on Build GRCh38
rs908884402 Chr.1: 192810524-192810524 on Build GRCh38
rs757964706 Chr.1: 192810358-192810358 on Build GRCh38
rs1030477176 Chr.1: 192810502-192810502 on Build GRCh38
rs996363183 Chr.1: 192810305-192810305 on Build GRCh38
rs767838827 Chr.1: 192810488-192810488 on Build GRCh38
rs537691552 Chr.1: 192810520-192810520 on Build GRCh38
rs768656623 Chr.1: 192810416-192810416 on Build GRCh38
rs1015378090 Chr.1: 192810339-192810339 on Build GRCh38
rs529917908 Chr.1: 192810336-192810336 on Build GRCh38
rs759569562 Chr.1: 192810340-192810340 on Build GRCh38
rs772262559 Chr.1: 192810441-192810441 on Build GRCh38
rs753159597 Chr.1: 192810631-192810631 on Build GRCh38
rs186294942 Chr.1: 192810733-192810733 on Build GRCh38
rs752316198 Chr.1: 192810353-192810353 on Build GRCh38
rs12565735 Chr.1: 192810664-192810664 on Build GRCh38
rs757727614 Chr.1: 192810373-192810373 on Build GRCh38
rs1057339364 Chr.1: 192810690-192810690 on Build GRCh38
rs1002879466 Chr.1: 192810756-192810756 on Build GRCh38
rs547204431 Chr.1: 192810432-192810432 on Build GRCh38
rs781569392 Chr.1: 192810374-192810374 on Build GRCh38
rs570476212 Chr.1: 192810580-192810580 on Build GRCh38
rs372850701 Chr.1: 192810470-192810470 on Build GRCh38
rs200208332 Chr.1: 192810395-192810395 on Build GRCh38
rs980333429 Chr.1: 192810600-192810600 on Build GRCh38
rs371653540 Chr.1: 192810326-192810326 on Build GRCh38
rs751831501 Chr.1: 192810466-192810466 on Build GRCh38
rs374873881 Chr.1: 192810468-192810468 on Build GRCh38
rs775228976 Chr.1: 192810483-192810483 on Build GRCh38
rs780414391 Chr.1: 192810408-192810408 on Build GRCh38
rs552200895 Chr.1: 192810490-192810490 on Build GRCh38
rs1027866716 Chr.1: 192810306-192810306 on Build GRCh38
rs1039446967 Chr.1: 192810514-192810514 on Build GRCh38
rs1004877458 Chr.1: 192810325-192810325 on Build GRCh38
rs927577317 Chr.1: 192810601-192810601 on Build GRCh38
rs45577436 Chr.1: 192810738-192810738 on Build GRCh38
rs146292708 Chr.1: 192810510-192810510 on Build GRCh38
rs368471160 Chr.1: 192810323-192810323 on Build GRCh38
rs1005569036 Chr.1: 192810660-192810660 on Build GRCh38
rs963573097 Chr.1: 192810552-192810552 on Build GRCh38
rs781296778 Chr.1: 192810350-192810351 on Build GRCh38
rs141407357 Chr.1: 192810485-192810485 on Build GRCh38
rs762939759 Chr.1: 192810316-192810316 on Build GRCh38
rs912244790 Chr.1: 192810494-192810494 on Build GRCh38
rs775686830 Chr.1: 192810310-192810310 on Build GRCh38
rs1034415952 Chr.1: 192810381-192810381 on Build GRCh38
rs375755008 Chr.1: 192810554-192810554 on Build GRCh38
rs1000790169 Chr.1: 192810459-192810459 on Build GRCh38
rs756711570 Chr.1: 192810393-192810393 on Build GRCh38
rs45594334 Chr.1: 192810341-192810341 on Build GRCh38
rs749383315 Chr.1: 192810412-192810412 on Build GRCh38
rs754344037 Chr.1: 192810375-192810377 on Build GRCh38
rs971487102 Chr.1: 192810363-192810363 on Build GRCh38
rs765692586 Chr.1: 192810465-192810466 on Build GRCh38
rs774590409 Chr.1: 192810436-192810436 on Build GRCh38
rs1043111918 Chr.1: 192810431-192810431 on Build GRCh38
rs762304757 Chr.1: 192810434-192810434 on Build GRCh38
rs764528890 Chr.1: 192810349-192810349 on Build GRCh38
rs768158249 Chr.1: 192810642-192810642 on Build GRCh38
rs766126102 Chr.1: 192810335-192810335 on Build GRCh38
rs763847693 Chr.1: 192810359-192810359 on Build GRCh38
rs183691150 Chr.1: 192810384-192810384 on Build GRCh38
rs751189088 Chr.1: 192810361-192810361 on Build GRCh38
rs765273898 Chr.1: 192810476-192810476 on Build GRCh38
rs907175557 Chr.1: 192810680-192810680 on Build GRCh38
rs766308836 Chr.1: 192810438-192810438 on Build GRCh38
rs1036351196 Chr.1: 192810661-192810661 on Build GRCh38
rs888035075 Chr.1: 192810320-192810320 on Build GRCh38

View Full Product Details