Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 193023791-193024306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UCHL5
Entrez Gene ID
51377
Gene Name
ubiquitin C-terminal hydrolase L5
Gene Aliases
CGI-70, INO80R, UCH-L5, UCH37
UniGene
Hs.145469
Gene Chromosome Location
Chr.1: 193012366-193060140 on Build GRCh38
Transcripts
NM_001199261.1 NM_001199262.1 NM_001199263.1 NM_015984.3 NR_037607.1 XM_005245244.4 XM_005245246.4 XM_005245247.4 XM_005245248.4 XM_005245249.4 XM_006711366.3 XM_006711367.3 XM_006711368.3 XM_006711369.2 XM_006711370.3 XM_006711371.3 XM_011509604.1 XM_011509607.2 XM_011509608.2 XM_011509609.2 XM_017001429.1 XM_017001430.1 XM_017001431.1 XM_017001432.1 XM_017001433.1 XM_017001434.1 XM_017001435.1 XM_017001436.1 XM_017001437.1 XM_017001438.1 XM_017001439.1 XM_017001440.1 XM_017001441.1 XM_017001442.1 XM_017001443.1 XM_017001444.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7421529 c.664T>C p.Y222H Chr.1: 193023885-193023885 on Build GRCh38
4340890 c.632A>G p.Y211C Chr.1: 193023944-193023944 on Build GRCh38
5686902 c.666C>T p.A222A Chr.1: 193023910-193023910 on Build GRCh38
7421525 c.691T>C p.Y231H Chr.1: 193023885-193023885 on Build GRCh38
4227901 c.605A>G p.Y202C Chr.1: 193023944-193023944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368644197 Chr.1: 193023976-193023976 on Build GRCh38
rs376746404 Chr.1: 193023986-193023986 on Build GRCh38
rs746827300 Chr.1: 193023815-193023815 on Build GRCh38
rs978410228 Chr.1: 193024067-193024067 on Build GRCh38
rs1014092855 Chr.1: 193024235-193024235 on Build GRCh38
rs1002616319 Chr.1: 193024213-193024213 on Build GRCh38
rs752172700 Chr.1: 193023958-193023961 on Build GRCh38
rs147440473 Chr.1: 193024088-193024088 on Build GRCh38
rs145003421 Chr.1: 193024179-193024179 on Build GRCh38
rs769820397 Chr.1: 193023853-193023853 on Build GRCh38
rs1034304983 Chr.1: 193024218-193024218 on Build GRCh38
rs1020052898 Chr.1: 193024244-193024244 on Build GRCh38
rs546690492 Chr.1: 193024018-193024018 on Build GRCh38
rs942351629 Chr.1: 193024109-193024109 on Build GRCh38
rs925650035 Chr.1: 193024103-193024103 on Build GRCh38
rs1033744755 Chr.1: 193024154-193024154 on Build GRCh38
rs887207446 Chr.1: 193024194-193024194 on Build GRCh38
rs975470257 Chr.1: 193024251-193024251 on Build GRCh38
rs553433201 Chr.1: 193024238-193024238 on Build GRCh38
rs369646878 Chr.1: 193023991-193023991 on Build GRCh38
rs750768618 Chr.1: 193023970-193023970 on Build GRCh38
rs963780025 Chr.1: 193024269-193024269 on Build GRCh38
rs759436892 Chr.1: 193023852-193023852 on Build GRCh38
rs775430311 Chr.1: 193023941-193023941 on Build GRCh38
rs1047632859 Chr.1: 193024193-193024193 on Build GRCh38
rs775204683 Chr.1: 193023877-193023877 on Build GRCh38
rs762535915 Chr.1: 193023886-193023886 on Build GRCh38
rs116048538 Chr.1: 193024199-193024199 on Build GRCh38
rs529629917 Chr.1: 193024002-193024002 on Build GRCh38
rs574003199 Chr.1: 193024200-193024200 on Build GRCh38
rs149924193 Chr.1: 193024148-193024148 on Build GRCh38
rs905220101 Chr.1: 193024203-193024203 on Build GRCh38
rs1034665237 Chr.1: 193023898-193023898 on Build GRCh38
rs139783848 Chr.1: 193024147-193024147 on Build GRCh38
rs566405050 Chr.1: 193024085-193024085 on Build GRCh38
rs1040100266 Chr.1: 193024013-193024013 on Build GRCh38
rs762268546 Chr.1: 193023940-193023940 on Build GRCh38
rs779230859 Chr.1: 193023989-193023989 on Build GRCh38
rs543490313 Chr.1: 193024243-193024244 on Build GRCh38
rs886962555 Chr.1: 193024112-193024112 on Build GRCh38
rs6672649 Chr.1: 193024223-193024223 on Build GRCh38
rs767724987 Chr.1: 193023958-193023958 on Build GRCh38
rs1010673748 Chr.1: 193024160-193024160 on Build GRCh38
rs912777974 Chr.1: 193023974-193023974 on Build GRCh38
rs922834862 Chr.1: 193024140-193024140 on Build GRCh38
rs977687194 Chr.1: 193023934-193023934 on Build GRCh38
rs967539526 Chr.1: 193024185-193024185 on Build GRCh38
rs931376635 Chr.1: 193024055-193024055 on Build GRCh38
rs200881380 Chr.1: 193023848-193023848 on Build GRCh38
rs1048438241 Chr.1: 193024062-193024062 on Build GRCh38
rs780527861 Chr.1: 193023980-193023980 on Build GRCh38
rs753929634 Chr.1: 193023985-193023985 on Build GRCh38
rs1032719791 Chr.1: 193024255-193024256 on Build GRCh38
rs371150012 Chr.1: 193024028-193024028 on Build GRCh38
rs1003963407 Chr.1: 193024119-193024119 on Build GRCh38
rs990357427 Chr.1: 193023835-193023835 on Build GRCh38
rs941398313 Chr.1: 193024195-193024195 on Build GRCh38
rs1020692603 Chr.1: 193024170-193024170 on Build GRCh38
rs374277946 Chr.1: 193024130-193024130 on Build GRCh38
rs751104681 Chr.1: 193023935-193023935 on Build GRCh38
rs967170754 Chr.1: 193024250-193024250 on Build GRCh38
rs921384180 Chr.1: 193024022-193024022 on Build GRCh38
rs975730078 Chr.1: 193024116-193024116 on Build GRCh38
rs957839333 Chr.1: 193024156-193024156 on Build GRCh38
rs571650883 Chr.1: 193024174-193024174 on Build GRCh38
rs375856662 Chr.1: 193023927-193023927 on Build GRCh38
rs201888142 Chr.1: 193024244-193024245 on Build GRCh38
rs975448289 Chr.1: 193024275-193024275 on Build GRCh38
rs112462491 Chr.1: 193023840-193023840 on Build GRCh38
rs967509934 Chr.1: 193023882-193023882 on Build GRCh38

View Full Product Details