Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 196672720-196673265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFH
Entrez Gene ID
3075
Gene Name
complement factor H
Gene Aliases
AHUS1, AMBP1, ARMD4, ARMS1, CFHL3, FH, FHL1, HF, HF1, HF2, HUS
UniGene
Hs.363396
Gene Chromosome Location
Chr.1: 196651878-196747504 on Build GRCh38
Transcripts
NM_000186.3 NM_001014975.2 XM_017001108.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6060125 c.243G>T p.Q81H Chr.1: 196673162-196673162 on Build GRCh38
463659 c.81A>T p.R27S Chr.1: 196673000-196673000 on Build GRCh38
145229 c.214G>A p.V72I Chr.1: 196673133-196673133 on Build GRCh38
463660 c.193G>A p.V65I Chr.1: 196673112-196673112 on Build GRCh38
146716 c.184G>A p.V62I Chr.1: 196673103-196673103 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781060260 Chr.1: 196673027-196673027 on Build GRCh38
rs1007759235 Chr.1: 196672990-196672990 on Build GRCh38
rs758081247 Chr.1: 196673152-196673152 on Build GRCh38
rs982559296 Chr.1: 196672790-196672790 on Build GRCh38
rs34731535 Chr.1: 196672841-196672842 on Build GRCh38
rs751825151 Chr.1: 196672957-196672957 on Build GRCh38
rs775851384 Chr.1: 196672845-196672846 on Build GRCh38
rs765558706 Chr.1: 196672958-196672958 on Build GRCh38
rs551397 Chr.1: 196672942-196672942 on Build GRCh38
rs1041787372 Chr.1: 196672752-196672752 on Build GRCh38
rs770000678 Chr.1: 196672967-196672967 on Build GRCh38
rs75064175 Chr.1: 196672810-196672810 on Build GRCh38
rs760566051 Chr.1: 196672997-196672997 on Build GRCh38
rs768809638 Chr.1: 196673212-196673212 on Build GRCh38
rs990343171 Chr.1: 196672954-196672954 on Build GRCh38
rs1015343796 Chr.1: 196672915-196672915 on Build GRCh38
rs778461886 Chr.1: 196673055-196673055 on Build GRCh38
rs938599130 Chr.1: 196672824-196672824 on Build GRCh38
rs976333015 Chr.1: 196673077-196673077 on Build GRCh38
rs764571169 Chr.1: 196673158-196673158 on Build GRCh38
rs753468508 Chr.1: 196672937-196672937 on Build GRCh38
rs755601507 Chr.1: 196672931-196672931 on Build GRCh38
rs941222340 Chr.1: 196672893-196672893 on Build GRCh38
rs781144436 Chr.1: 196673078-196673078 on Build GRCh38
rs764941928 Chr.1: 196673046-196673046 on Build GRCh38
rs751914625 Chr.1: 196673173-196673173 on Build GRCh38
rs757785149 Chr.1: 196673076-196673076 on Build GRCh38
rs372383350 Chr.1: 196673203-196673203 on Build GRCh38
rs754615067 Chr.1: 196672938-196672938 on Build GRCh38
rs929675494 Chr.1: 196672757-196672757 on Build GRCh38
rs767373625 Chr.1: 196672928-196672928 on Build GRCh38
rs977158597 Chr.1: 196672975-196672975 on Build GRCh38
rs538559848 Chr.1: 196672941-196672941 on Build GRCh38
rs368835171 Chr.1: 196673139-196673139 on Build GRCh38
rs761662067 Chr.1: 196672983-196672983 on Build GRCh38
rs773104581 Chr.1: 196672987-196672987 on Build GRCh38
rs886045742 Chr.1: 196673022-196673022 on Build GRCh38
rs770964312 Chr.1: 196672839-196672839 on Build GRCh38
rs755165508 Chr.1: 196673180-196673189 on Build GRCh38
rs760619101 Chr.1: 196673124-196673124 on Build GRCh38
rs1010034860 Chr.1: 196672948-196672948 on Build GRCh38
rs1004946508 Chr.1: 196672906-196672906 on Build GRCh38
rs754932801 Chr.1: 196672986-196672986 on Build GRCh38
rs776602859 Chr.1: 196672897-196672897 on Build GRCh38
rs370778401 Chr.1: 196672936-196672936 on Build GRCh38
rs146954818 Chr.1: 196672844-196672845 on Build GRCh38
rs752361141 Chr.1: 196673149-196673149 on Build GRCh38
rs756189687 Chr.1: 196673202-196673202 on Build GRCh38
rs931171935 Chr.1: 196672878-196672878 on Build GRCh38
rs747978546 Chr.1: 196673112-196673112 on Build GRCh38
rs759042534 Chr.1: 196672950-196672950 on Build GRCh38
rs928296341 Chr.1: 196672989-196672989 on Build GRCh38
rs1048202103 Chr.1: 196672887-196672887 on Build GRCh38
rs772943168 Chr.1: 196673120-196673120 on Build GRCh38
rs34895813 Chr.1: 196673219-196673219 on Build GRCh38
rs774019952 Chr.1: 196672974-196672974 on Build GRCh38
rs945801612 Chr.1: 196672746-196672746 on Build GRCh38
rs750524702 Chr.1: 196673190-196673190 on Build GRCh38
rs749999830 Chr.1: 196672930-196672930 on Build GRCh38
rs796052137 Chr.1: 196673002-196673005 on Build GRCh38
rs537084841 Chr.1: 196672837-196672837 on Build GRCh38
rs1055603171 Chr.1: 196672862-196672862 on Build GRCh38
rs140560017 Chr.1: 196672977-196672977 on Build GRCh38
rs1029289866 Chr.1: 196672944-196672944 on Build GRCh38
rs550027371 Chr.1: 196672924-196672924 on Build GRCh38
rs766241195 Chr.1: 196673133-196673133 on Build GRCh38
rs749536139 Chr.1: 196673209-196673209 on Build GRCh38
rs905147492 Chr.1: 196672922-196672922 on Build GRCh38
rs894388955 Chr.1: 196672864-196672864 on Build GRCh38
rs771886580 Chr.1: 196673114-196673114 on Build GRCh38
rs766487897 Chr.1: 196672953-196672953 on Build GRCh38
rs182750499 Chr.1: 196673044-196673044 on Build GRCh38
rs376689220 Chr.1: 196672843-196672844 on Build GRCh38
rs141336681 Chr.1: 196673091-196673091 on Build GRCh38
rs758810701 Chr.1: 196673053-196673053 on Build GRCh38
rs201067374 Chr.1: 196672962-196672962 on Build GRCh38
rs55759441 Chr.1: 196672865-196672866 on Build GRCh38
rs769975480 Chr.1: 196673100-196673100 on Build GRCh38
rs776075595 Chr.1: 196673138-196673138 on Build GRCh38
rs800292 Chr.1: 196673103-196673103 on Build GRCh38
rs866627729 Chr.1: 196672995-196672995 on Build GRCh38
rs767494411 Chr.1: 196672984-196672984 on Build GRCh38
rs541476028 Chr.1: 196672835-196672835 on Build GRCh38
rs757842424 Chr.1: 196673178-196673178 on Build GRCh38
rs778413105 Chr.1: 196672951-196672951 on Build GRCh38
rs764849497 Chr.1: 196673144-196673144 on Build GRCh38
rs747546121 Chr.1: 196673064-196673064 on Build GRCh38
rs765897045 Chr.1: 196673020-196673020 on Build GRCh38
rs781719657 Chr.1: 196673182-196673182 on Build GRCh38
rs771102987 Chr.1: 196672956-196672956 on Build GRCh38
rs796760458 Chr.1: 196672844-196672845 on Build GRCh38

View Full Product Details