Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 198540786-198541285 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1G3
Entrez Gene ID
127124
Gene Name
ATPase H+ transporting V1 subunit G3
Gene Aliases
ATP6G3, Vma10
UniGene
Hs.127743
Gene Chromosome Location
Chr.1: 198523222-198540945 on Build GRCh38
Transcripts
NM_001320218.1 NM_133262.2 NM_133326.1

SNP Detail

SNP ID Chromosome Location
rs77836571 Chr.1: 198540809-198540809 on Build GRCh38
rs919851633 Chr.1: 198541078-198541078 on Build GRCh38
rs995748339 Chr.1: 198541245-198541245 on Build GRCh38
rs915881598 Chr.1: 198541096-198541096 on Build GRCh38
rs143103047 Chr.1: 198541009-198541009 on Build GRCh38
rs1045815089 Chr.1: 198541108-198541108 on Build GRCh38
rs919621680 Chr.1: 198541185-198541185 on Build GRCh38
rs548863940 Chr.1: 198541112-198541112 on Build GRCh38
rs1033706982 Chr.1: 198540861-198540861 on Build GRCh38
rs1027951792 Chr.1: 198540942-198540942 on Build GRCh38
rs955115616 Chr.1: 198540954-198540954 on Build GRCh38
rs543538599 Chr.1: 198541124-198541124 on Build GRCh38
rs762515693 Chr.1: 198541098-198541098 on Build GRCh38
rs751157396 Chr.1: 198541142-198541142 on Build GRCh38
rs74721994 Chr.1: 198541044-198541044 on Build GRCh38
rs553753331 Chr.1: 198540883-198540883 on Build GRCh38
rs777192720 Chr.1: 198541085-198541085 on Build GRCh38
rs924693142 Chr.1: 198541111-198541111 on Build GRCh38
rs950040214 Chr.1: 198541100-198541100 on Build GRCh38
rs557596639 Chr.1: 198541243-198541243 on Build GRCh38
rs558726124 Chr.1: 198541153-198541153 on Build GRCh38
rs529911008 Chr.1: 198541253-198541253 on Build GRCh38
rs372296692 Chr.1: 198541242-198541242 on Build GRCh38
rs997080341 Chr.1: 198541141-198541141 on Build GRCh38
rs973921014 Chr.1: 198541057-198541057 on Build GRCh38
rs944269217 Chr.1: 198540941-198540941 on Build GRCh38
rs761078981 Chr.1: 198540991-198540991 on Build GRCh38
rs982512205 Chr.1: 198541228-198541228 on Build GRCh38
rs991903499 Chr.1: 198541094-198541094 on Build GRCh38
rs943132 Chr.1: 198540915-198540915 on Build GRCh38
rs1015349167 Chr.1: 198540971-198540971 on Build GRCh38
rs893398688 Chr.1: 198541132-198541132 on Build GRCh38
rs887288945 Chr.1: 198540934-198540934 on Build GRCh38
rs1041694630 Chr.1: 198540959-198540959 on Build GRCh38
rs138400846 Chr.1: 198541226-198541226 on Build GRCh38
rs746548640 Chr.1: 198540977-198540977 on Build GRCh38
rs991543949 Chr.1: 198541154-198541154 on Build GRCh38
rs559319335 Chr.1: 198541241-198541241 on Build GRCh38
rs959295841 Chr.1: 198541089-198541089 on Build GRCh38
rs772094121 Chr.1: 198540886-198540886 on Build GRCh38
rs1008014240 Chr.1: 198540960-198540960 on Build GRCh38
rs959563549 Chr.1: 198541261-198541261 on Build GRCh38
rs147052323 Chr.1: 198541196-198541196 on Build GRCh38
rs890819555 Chr.1: 198541188-198541188 on Build GRCh38
rs538609971 Chr.1: 198540829-198540829 on Build GRCh38
rs931087145 Chr.1: 198540899-198540899 on Build GRCh38

View Full Product Details