Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 201386937-201387433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LAD1
Entrez Gene ID
3898
Gene Name
ladinin 1
Gene Aliases
LadA
UniGene
Hs.519035
Gene Chromosome Location
Chr.1: 201380838-201399541 on Build GRCh38
Transcripts
NM_005558.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4906012 c.451G>A p.E151K Chr.1: 201386952-201386952 on Build GRCh38
4904217 c.286G>A p.D96N Chr.1: 201387117-201387117 on Build GRCh38
4787397 c.388G>T p.E130* Chr.1: 201387015-201387015 on Build GRCh38
6698951 c.318G>T p.Q106H Chr.1: 201387043-201387043 on Build GRCh38
4899392 c.326G>A p.R109Q Chr.1: 201387077-201387077 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143280381 Chr.1: 201387170-201387170 on Build GRCh38
rs779873997 Chr.1: 201387108-201387109 on Build GRCh38
rs200089805 Chr.1: 201387136-201387136 on Build GRCh38
rs368638299 Chr.1: 201387141-201387141 on Build GRCh38
rs747024773 Chr.1: 201387172-201387172 on Build GRCh38
rs774353289 Chr.1: 201387015-201387015 on Build GRCh38
rs770999018 Chr.1: 201387034-201387034 on Build GRCh38
rs781678996 Chr.1: 201387066-201387066 on Build GRCh38
rs768544493 Chr.1: 201387187-201387187 on Build GRCh38
rs4915230 Chr.1: 201387300-201387300 on Build GRCh38
rs181261425 Chr.1: 201387079-201387079 on Build GRCh38
rs1001717756 Chr.1: 201387060-201387060 on Build GRCh38
rs1023480839 Chr.1: 201387401-201387401 on Build GRCh38
rs112981585 Chr.1: 201387354-201387354 on Build GRCh38
rs775810941 Chr.1: 201386995-201386995 on Build GRCh38
rs372084515 Chr.1: 201387115-201387115 on Build GRCh38
rs760114057 Chr.1: 201387227-201387227 on Build GRCh38
rs770094076 Chr.1: 201386985-201386985 on Build GRCh38
rs34525025 Chr.1: 201387065-201387065 on Build GRCh38
rs780464600 Chr.1: 201387077-201387077 on Build GRCh38
rs759430005 Chr.1: 201387038-201387038 on Build GRCh38
rs933175528 Chr.1: 201387248-201387248 on Build GRCh38
rs930393495 Chr.1: 201387246-201387252 on Build GRCh38
rs773391681 Chr.1: 201387096-201387096 on Build GRCh38
rs534710597 Chr.1: 201387120-201387120 on Build GRCh38
rs759609920 Chr.1: 201387102-201387102 on Build GRCh38
rs74662114 Chr.1: 201387265-201387265 on Build GRCh38
rs766829657 Chr.1: 201387228-201387228 on Build GRCh38
rs201050623 Chr.1: 201387086-201387086 on Build GRCh38
rs769838822 Chr.1: 201387114-201387114 on Build GRCh38
rs749058067 Chr.1: 201387186-201387186 on Build GRCh38
rs147613262 Chr.1: 201387154-201387154 on Build GRCh38
rs201607259 Chr.1: 201387071-201387071 on Build GRCh38
rs1033430409 Chr.1: 201387412-201387412 on Build GRCh38
rs762567945 Chr.1: 201387047-201387047 on Build GRCh38
rs775140152 Chr.1: 201387030-201387030 on Build GRCh38
rs41302137 Chr.1: 201387173-201387173 on Build GRCh38
rs767144810 Chr.1: 201387213-201387213 on Build GRCh38
rs369509551 Chr.1: 201387266-201387266 on Build GRCh38
rs772233623 Chr.1: 201387087-201387087 on Build GRCh38
rs1020439969 Chr.1: 201387342-201387342 on Build GRCh38
rs1051586406 Chr.1: 201387263-201387263 on Build GRCh38
rs200604690 Chr.1: 201387125-201387125 on Build GRCh38
rs754160423 Chr.1: 201387151-201387151 on Build GRCh38
rs1056004993 Chr.1: 201387318-201387318 on Build GRCh38
rs898793549 Chr.1: 201387306-201387306 on Build GRCh38
rs375408134 Chr.1: 201387131-201387131 on Build GRCh38
rs776674635 Chr.1: 201387036-201387036 on Build GRCh38
rs940423564 Chr.1: 201387284-201387284 on Build GRCh38
rs547836419 Chr.1: 201387239-201387239 on Build GRCh38
rs373145430 Chr.1: 201387057-201387057 on Build GRCh38
rs944379282 Chr.1: 201387270-201387272 on Build GRCh38
rs779029242 Chr.1: 201387168-201387168 on Build GRCh38
rs139455501 Chr.1: 201387005-201387005 on Build GRCh38
rs749512948 Chr.1: 201387001-201387001 on Build GRCh38
rs188007352 Chr.1: 201387078-201387078 on Build GRCh38
rs570680572 Chr.1: 201387366-201387366 on Build GRCh38
rs1001747256 Chr.1: 201387317-201387317 on Build GRCh38
rs746263240 Chr.1: 201386974-201386974 on Build GRCh38
rs751033233 Chr.1: 201387059-201387059 on Build GRCh38
rs957488659 Chr.1: 201387123-201387123 on Build GRCh38
rs550481230 Chr.1: 201387331-201387331 on Build GRCh38
rs750976101 Chr.1: 201387072-201387072 on Build GRCh38
rs780554157 Chr.1: 201387148-201387148 on Build GRCh38
rs200025654 Chr.1: 201387169-201387169 on Build GRCh38
rs1036075765 Chr.1: 201386965-201386965 on Build GRCh38
rs769283990 Chr.1: 201387175-201387175 on Build GRCh38
rs768111267 Chr.1: 201387392-201387392 on Build GRCh38
rs974252866 Chr.1: 201387218-201387218 on Build GRCh38
rs765159869 Chr.1: 201387039-201387039 on Build GRCh38
rs756582952 Chr.1: 201387074-201387074 on Build GRCh38
rs757635955 Chr.1: 201387008-201387009 on Build GRCh38
rs1050640339 Chr.1: 201387247-201387247 on Build GRCh38
rs1054203588 Chr.1: 201387330-201387330 on Build GRCh38
rs199686862 Chr.1: 201387042-201387042 on Build GRCh38
rs773985312 Chr.1: 201387209-201387209 on Build GRCh38
rs749932257 Chr.1: 201387134-201387134 on Build GRCh38
rs925949426 Chr.1: 201387138-201387138 on Build GRCh38
rs761432152 Chr.1: 201387130-201387130 on Build GRCh38
rs902989823 Chr.1: 201387092-201387092 on Build GRCh38
rs1000264306 Chr.1: 201387388-201387388 on Build GRCh38
rs528222926 Chr.1: 201387219-201387219 on Build GRCh38
rs1020825611 Chr.1: 201387083-201387083 on Build GRCh38
rs1014862661 Chr.1: 201387336-201387336 on Build GRCh38
rs961516869 Chr.1: 201387216-201387216 on Build GRCh38
rs761520509 Chr.1: 201387212-201387212 on Build GRCh38

View Full Product Details